Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87407916-87408440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB4
Entrez Gene ID
5244
Gene Name
ATP binding cassette subfamily B member 4
Gene Aliases
ABC21, GBD1, ICP3, MDR2, MDR2/3, MDR3, PFIC-3, PGY3
UniGene
Hs.654403
Gene Chromosome Location
Chr.7: 87399461-87475864 on Build GRCh38
Transcripts
NM_000443.3 NM_018849.2 NM_018850.2 XM_011516308.2 XM_011516309.2 XM_011516310.2 XM_011516311.2 XM_011516312.2 XM_011516313.2 XM_011516315.2 XM_017012323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422439 c.3159G>C p.Q1053H Chr.7: 87408157-87408157 on Build GRCh38
5557609 c.3225G>T p.K1075N Chr.7: 87408091-87408091 on Build GRCh38
5589497 c.3095G>A p.G1032E Chr.7: 87408221-87408221 on Build GRCh38
6873086 c.3209G>A p.S1070N Chr.7: 87408107-87408107 on Build GRCh38
158796 c.3258C>T p.Y1086Y Chr.7: 87408058-87408058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779141589 Chr.7: 87408123-87408123 on Build GRCh38
rs1045923356 Chr.7: 87407963-87407963 on Build GRCh38
rs371486074 Chr.7: 87408146-87408146 on Build GRCh38
rs772948456 Chr.7: 87408075-87408075 on Build GRCh38
rs757571888 Chr.7: 87408113-87408113 on Build GRCh38
rs774151533 Chr.7: 87408179-87408179 on Build GRCh38
rs376152562 Chr.7: 87407985-87407985 on Build GRCh38
rs868403254 Chr.7: 87408148-87408148 on Build GRCh38
rs758615467 Chr.7: 87408206-87408206 on Build GRCh38
rs907407119 Chr.7: 87407980-87407980 on Build GRCh38
rs770859693 Chr.7: 87408271-87408271 on Build GRCh38
rs542917895 Chr.7: 87408105-87408105 on Build GRCh38
rs371152960 Chr.7: 87408185-87408185 on Build GRCh38
rs575403204 Chr.7: 87408030-87408030 on Build GRCh38
rs769661643 Chr.7: 87408165-87408165 on Build GRCh38
rs147171162 Chr.7: 87408065-87408065 on Build GRCh38
rs868333171 Chr.7: 87408045-87408045 on Build GRCh38
rs781769968 Chr.7: 87408005-87408005 on Build GRCh38
rs749666244 Chr.7: 87408109-87408109 on Build GRCh38
rs8187808 Chr.7: 87408205-87408205 on Build GRCh38
rs754565782 Chr.7: 87408086-87408086 on Build GRCh38
rs754513436 Chr.7: 87408002-87408002 on Build GRCh38
rs745972631 Chr.7: 87408059-87408059 on Build GRCh38
rs55690313 Chr.7: 87408057-87408057 on Build GRCh38
rs759202962 Chr.7: 87408180-87408180 on Build GRCh38
rs941333716 Chr.7: 87408252-87408252 on Build GRCh38
rs140387595 Chr.7: 87408144-87408144 on Build GRCh38
rs767182556 Chr.7: 87408183-87408183 on Build GRCh38
rs772408988 Chr.7: 87408320-87408320 on Build GRCh38
rs147929636 Chr.7: 87408201-87408201 on Build GRCh38
rs376825608 Chr.7: 87408085-87408085 on Build GRCh38
rs761716829 Chr.7: 87408004-87408004 on Build GRCh38
rs753258408 Chr.7: 87408097-87408097 on Build GRCh38
rs142980157 Chr.7: 87408226-87408226 on Build GRCh38
rs772103147 Chr.7: 87408061-87408061 on Build GRCh38
rs747464582 Chr.7: 87408394-87408394 on Build GRCh38
rs939911320 Chr.7: 87408248-87408248 on Build GRCh38
rs143340462 Chr.7: 87408296-87408296 on Build GRCh38
rs947646201 Chr.7: 87408380-87408380 on Build GRCh38
rs756502124 Chr.7: 87408023-87408023 on Build GRCh38
rs370897670 Chr.7: 87408282-87408282 on Build GRCh38
rs766868231 Chr.7: 87408093-87408093 on Build GRCh38
rs765208250 Chr.7: 87408074-87408074 on Build GRCh38
rs757626981 Chr.7: 87408198-87408198 on Build GRCh38
rs748563314 Chr.7: 87408011-87408011 on Build GRCh38
rs777601533 Chr.7: 87408238-87408238 on Build GRCh38
rs983123635 Chr.7: 87408122-87408122 on Build GRCh38
rs534519842 Chr.7: 87408276-87408276 on Build GRCh38
rs368167333 Chr.7: 87408115-87408115 on Build GRCh38
rs773177743 Chr.7: 87408170-87408170 on Build GRCh38
rs754306589 Chr.7: 87408196-87408196 on Build GRCh38
rs1032368464 Chr.7: 87407983-87407983 on Build GRCh38
rs774294440 Chr.7: 87407996-87407996 on Build GRCh38
rs907354691 Chr.7: 87408398-87408398 on Build GRCh38
rs1036411641 Chr.7: 87407974-87407974 on Build GRCh38
rs974079968 Chr.7: 87408288-87408290 on Build GRCh38
rs903426338 Chr.7: 87408405-87408405 on Build GRCh38
rs760137938 Chr.7: 87408286-87408286 on Build GRCh38
rs762862613 Chr.7: 87408171-87408171 on Build GRCh38
rs772154065 Chr.7: 87408142-87408142 on Build GRCh38
rs940780683 Chr.7: 87407968-87407968 on Build GRCh38
rs761598845 Chr.7: 87408069-87408069 on Build GRCh38
rs45484901 Chr.7: 87408353-87408353 on Build GRCh38
rs901920014 Chr.7: 87407990-87407990 on Build GRCh38
rs976982008 Chr.7: 87408277-87408277 on Build GRCh38
rs1045870823 Chr.7: 87408381-87408381 on Build GRCh38
rs768730681 Chr.7: 87408232-87408232 on Build GRCh38
rs377364410 Chr.7: 87408261-87408261 on Build GRCh38
rs143392573 Chr.7: 87408047-87408047 on Build GRCh38
rs766180923 Chr.7: 87408081-87408081 on Build GRCh38
rs45526441 Chr.7: 87408413-87408413 on Build GRCh38
rs55867538 Chr.7: 87408279-87408279 on Build GRCh38
rs774536983 Chr.7: 87408054-87408054 on Build GRCh38
rs533510070 Chr.7: 87408281-87408281 on Build GRCh38
rs762812271 Chr.7: 87408076-87408076 on Build GRCh38
rs549672794 Chr.7: 87408366-87408366 on Build GRCh38
rs8187807 Chr.7: 87408172-87408172 on Build GRCh38
rs779194886 Chr.7: 87408199-87408199 on Build GRCh38
rs759397269 Chr.7: 87408278-87408278 on Build GRCh38
rs780292654 Chr.7: 87408215-87408215 on Build GRCh38
rs184992589 Chr.7: 87408316-87408316 on Build GRCh38
rs776811744 Chr.7: 87407999-87407999 on Build GRCh38
rs999096003 Chr.7: 87407981-87407981 on Build GRCh38
rs778336813 Chr.7: 87408106-87408106 on Build GRCh38
rs144852385 Chr.7: 87408186-87408186 on Build GRCh38
rs749479443 Chr.7: 87407955-87407955 on Build GRCh38
rs749647874 Chr.7: 87408043-87408043 on Build GRCh38
rs373011908 Chr.7: 87408137-87408137 on Build GRCh38

View Full Product Details