Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90268045-90268544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP69
Entrez Gene ID
79846
Gene Name
cilia and flagella associated protein 69
Gene Aliases
C7orf63, FAP69
UniGene
Hs.521014
Gene Chromosome Location
Chr.7: 90245174-90320964 on Build GRCh38
Transcripts
NM_001039706.2 NM_001160138.1 XM_005250600.3 XM_011516577.2 XM_011516578.2 XM_011516579.1 XM_017012624.1 XM_017012625.1 XM_017012626.1 XM_017012627.1 XM_017012628.1 XM_017012629.1 XM_017012630.1 XM_017012631.1 XM_017012632.1 XM_017012633.1 XM_017012634.1 XM_017012635.1 XM_017012636.1 XM_017012637.1 XM_017012638.1 XM_017012639.1 XM_017012641.1 XM_017012642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721089 c.467G>T p.R156M Chr.7: 90268319-90268319 on Build GRCh38
1635463 c.512C>T p.P171L Chr.7: 90268364-90268364 on Build GRCh38
3642129 c.493G>T p.D165Y Chr.7: 90268345-90268345 on Build GRCh38
5408229 c.439T>C p.L147L Chr.7: 90268291-90268291 on Build GRCh38
3431841 c.517A>G p.K173E Chr.7: 90268369-90268369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010527727 Chr.7: 90268439-90268439 on Build GRCh38
rs186778385 Chr.7: 90268083-90268083 on Build GRCh38
rs914202498 Chr.7: 90268441-90268441 on Build GRCh38
rs183530931 Chr.7: 90268231-90268231 on Build GRCh38
rs532739354 Chr.7: 90268181-90268181 on Build GRCh38
rs907492796 Chr.7: 90268464-90268464 on Build GRCh38
rs558797982 Chr.7: 90268496-90268496 on Build GRCh38
rs745864940 Chr.7: 90268283-90268283 on Build GRCh38
rs1048175982 Chr.7: 90268381-90268381 on Build GRCh38
rs537665078 Chr.7: 90268248-90268248 on Build GRCh38
rs979882681 Chr.7: 90268322-90268322 on Build GRCh38
rs756788449 Chr.7: 90268479-90268479 on Build GRCh38
rs776348740 Chr.7: 90268342-90268342 on Build GRCh38
rs938525295 Chr.7: 90268230-90268230 on Build GRCh38
rs1042356460 Chr.7: 90268463-90268463 on Build GRCh38
rs778795185 Chr.7: 90268317-90268317 on Build GRCh38
rs956328580 Chr.7: 90268232-90268232 on Build GRCh38
rs748254646 Chr.7: 90268255-90268255 on Build GRCh38
rs1006238657 Chr.7: 90268488-90268488 on Build GRCh38
rs1017718251 Chr.7: 90268219-90268219 on Build GRCh38
rs371357343 Chr.7: 90268434-90268434 on Build GRCh38
rs17867605 Chr.7: 90268515-90268515 on Build GRCh38
rs190622414 Chr.7: 90268221-90268221 on Build GRCh38
rs1009020596 Chr.7: 90268180-90268180 on Build GRCh38
rs373747438 Chr.7: 90268425-90268425 on Build GRCh38
rs780170968 Chr.7: 90268422-90268422 on Build GRCh38
rs1039053774 Chr.7: 90268427-90268427 on Build GRCh38
rs772100468 Chr.7: 90268288-90268288 on Build GRCh38
rs1026330181 Chr.7: 90268107-90268107 on Build GRCh38
rs1034396047 Chr.7: 90268206-90268206 on Build GRCh38
rs979768156 Chr.7: 90268475-90268475 on Build GRCh38
rs1033168556 Chr.7: 90268436-90268436 on Build GRCh38
rs764795583 Chr.7: 90268346-90268346 on Build GRCh38
rs978901018 Chr.7: 90268223-90268223 on Build GRCh38
rs376739951 Chr.7: 90268426-90268426 on Build GRCh38
rs574843478 Chr.7: 90268443-90268443 on Build GRCh38
rs778034979 Chr.7: 90268402-90268402 on Build GRCh38
rs368604033 Chr.7: 90268395-90268395 on Build GRCh38
rs1054222402 Chr.7: 90268440-90268440 on Build GRCh38
rs971055429 Chr.7: 90268444-90268444 on Build GRCh38
rs746843204 Chr.7: 90268301-90268301 on Build GRCh38
rs1003033086 Chr.7: 90268157-90268157 on Build GRCh38
rs777984596 Chr.7: 90268282-90268282 on Build GRCh38
rs772680500 Chr.7: 90268349-90268349 on Build GRCh38
rs751901045 Chr.7: 90268244-90268244 on Build GRCh38
rs755378721 Chr.7: 90268362-90268362 on Build GRCh38
rs767718862 Chr.7: 90268379-90268379 on Build GRCh38
rs1016650925 Chr.7: 90268269-90268269 on Build GRCh38
rs751974341 Chr.7: 90268356-90268356 on Build GRCh38
rs1041993506 Chr.7: 90268101-90268101 on Build GRCh38
rs565508025 Chr.7: 90268148-90268148 on Build GRCh38
rs764950014 Chr.7: 90268180-90268181 on Build GRCh38
rs368893731 Chr.7: 90268266-90268266 on Build GRCh38
rs897079594 Chr.7: 90268070-90268070 on Build GRCh38
rs371422121 Chr.7: 90268431-90268431 on Build GRCh38
rs749353784 Chr.7: 90268411-90268411 on Build GRCh38
rs781512331 Chr.7: 90268252-90268252 on Build GRCh38
rs17863057 Chr.7: 90268389-90268389 on Build GRCh38
rs376199817 Chr.7: 90268344-90268344 on Build GRCh38
rs1016373690 Chr.7: 90268490-90268490 on Build GRCh38
rs200834034 Chr.7: 90268352-90268352 on Build GRCh38
rs201631766 Chr.7: 90268354-90268354 on Build GRCh38
rs372422328 Chr.7: 90268315-90268315 on Build GRCh38
rs1003083963 Chr.7: 90268472-90268472 on Build GRCh38
rs927039317 Chr.7: 90268323-90268323 on Build GRCh38
rs17869450 Chr.7: 90268437-90268437 on Build GRCh38
rs530113668 Chr.7: 90268331-90268331 on Build GRCh38
rs775471186 Chr.7: 90268295-90268295 on Build GRCh38
rs17867345 Chr.7: 90268409-90268409 on Build GRCh38
rs757363115 Chr.7: 90268415-90268415 on Build GRCh38
rs894661992 Chr.7: 90268481-90268481 on Build GRCh38
rs755252982 Chr.7: 90268250-90268250 on Build GRCh38

View Full Product Details