Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91265014-91265506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD1
Entrez Gene ID
8321
Gene Name
frizzled class receptor 1
Gene Aliases
-
UniGene
Hs.94234
Gene Chromosome Location
Chr.7: 91264468-91268817 on Build GRCh38
Transcripts
NM_003505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668114 c.422C>G p.T141R Chr.7: 91265302-91265302 on Build GRCh38
6419454 c.277C>A p.P93T Chr.7: 91265157-91265157 on Build GRCh38
6668112 c.449T>G p.V150G Chr.7: 91265329-91265329 on Build GRCh38
4877264 c.497A>G p.K166R Chr.7: 91265377-91265377 on Build GRCh38
3374686 c.618C>A p.G206G Chr.7: 91265498-91265498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781188913 Chr.7: 91265366-91265366 on Build GRCh38
rs531115231 Chr.7: 91265430-91265430 on Build GRCh38
rs1021665247 Chr.7: 91265439-91265439 on Build GRCh38
rs759080221 Chr.7: 91265108-91265108 on Build GRCh38
rs780272926 Chr.7: 91265147-91265148 on Build GRCh38
rs139480179 Chr.7: 91265144-91265145 on Build GRCh38
rs1014656110 Chr.7: 91265134-91265134 on Build GRCh38
rs377459455 Chr.7: 91265212-91265212 on Build GRCh38
rs770398025 Chr.7: 91265411-91265411 on Build GRCh38
rs529145118 Chr.7: 91265162-91265162 on Build GRCh38
rs767964823 Chr.7: 91265118-91265118 on Build GRCh38
rs769565425 Chr.7: 91265167-91265167 on Build GRCh38
rs751725868 Chr.7: 91265346-91265346 on Build GRCh38
rs898110856 Chr.7: 91265273-91265273 on Build GRCh38
rs370896736 Chr.7: 91265483-91265483 on Build GRCh38
rs866903038 Chr.7: 91265064-91265064 on Build GRCh38
rs749138157 Chr.7: 91265236-91265236 on Build GRCh38
rs755245569 Chr.7: 91265160-91265162 on Build GRCh38
rs769346857 Chr.7: 91265296-91265296 on Build GRCh38
rs776244417 Chr.7: 91265405-91265405 on Build GRCh38
rs765641770 Chr.7: 91265210-91265210 on Build GRCh38
rs563575931 Chr.7: 91265161-91265161 on Build GRCh38
rs577757561 Chr.7: 91265147-91265147 on Build GRCh38
rs766475224 Chr.7: 91265344-91265344 on Build GRCh38
rs746687511 Chr.7: 91265457-91265457 on Build GRCh38
rs992945609 Chr.7: 91265205-91265205 on Build GRCh38
rs759052167 Chr.7: 91265196-91265196 on Build GRCh38
rs757146464 Chr.7: 91265369-91265369 on Build GRCh38
rs771602779 Chr.7: 91265264-91265264 on Build GRCh38
rs149357966 Chr.7: 91265263-91265263 on Build GRCh38
rs1028239110 Chr.7: 91265104-91265104 on Build GRCh38
rs71292991 Chr.7: 91265158-91265159 on Build GRCh38
rs777680747 Chr.7: 91265235-91265235 on Build GRCh38
rs375051170 Chr.7: 91265188-91265188 on Build GRCh38
rs756511034 Chr.7: 91265136-91265136 on Build GRCh38
rs988874488 Chr.7: 91265224-91265224 on Build GRCh38
rs757226244 Chr.7: 91265443-91265443 on Build GRCh38
rs757520423 Chr.7: 91265042-91265044 on Build GRCh38
rs931865188 Chr.7: 91265045-91265045 on Build GRCh38
rs999129541 Chr.7: 91265462-91265462 on Build GRCh38
rs763226057 Chr.7: 91265417-91265417 on Build GRCh38
rs1040309734 Chr.7: 91265104-91265115 on Build GRCh38
rs779934443 Chr.7: 91265164-91265164 on Build GRCh38
rs770493888 Chr.7: 91265461-91265461 on Build GRCh38
rs745620349 Chr.7: 91265377-91265377 on Build GRCh38
rs202070404 Chr.7: 91265159-91265159 on Build GRCh38
rs764541069 Chr.7: 91265144-91265144 on Build GRCh38
rs142085794 Chr.7: 91265070-91265070 on Build GRCh38
rs749748342 Chr.7: 91265480-91265480 on Build GRCh38
rs777293881 Chr.7: 91265304-91265304 on Build GRCh38
rs200651234 Chr.7: 91265445-91265445 on Build GRCh38
rs778728528 Chr.7: 91265372-91265372 on Build GRCh38
rs767616876 Chr.7: 91265435-91265435 on Build GRCh38
rs773582345 Chr.7: 91265471-91265471 on Build GRCh38
rs754322513 Chr.7: 91265209-91265209 on Build GRCh38
rs775107631 Chr.7: 91265289-91265289 on Build GRCh38
rs368968709 Chr.7: 91265119-91265119 on Build GRCh38
rs779829428 Chr.7: 91265447-91265447 on Build GRCh38
rs541996570 Chr.7: 91265428-91265428 on Build GRCh38
rs887909297 Chr.7: 91265117-91265117 on Build GRCh38
rs749049302 Chr.7: 91265185-91265185 on Build GRCh38
rs779561649 Chr.7: 91265387-91265387 on Build GRCh38
rs370651906 Chr.7: 91265429-91265429 on Build GRCh38
rs754232711 Chr.7: 91265146-91265146 on Build GRCh38
rs758139174 Chr.7: 91265444-91265444 on Build GRCh38
rs377247017 Chr.7: 91265255-91265255 on Build GRCh38
rs959977879 Chr.7: 91265193-91265193 on Build GRCh38
rs891811319 Chr.7: 91265086-91265086 on Build GRCh38
rs770635483 Chr.7: 91265187-91265187 on Build GRCh38
rs772947349 Chr.7: 91265055-91265055 on Build GRCh38
rs749576541 Chr.7: 91265155-91265156 on Build GRCh38
rs752725437 Chr.7: 91265438-91265438 on Build GRCh38
rs375790061 Chr.7: 91265207-91265207 on Build GRCh38
rs748151772 Chr.7: 91265165-91265165 on Build GRCh38
rs773662622 Chr.7: 91265415-91265415 on Build GRCh38
rs551898695 Chr.7: 91265327-91265327 on Build GRCh38
rs760101096 Chr.7: 91265291-91265291 on Build GRCh38
rs979669443 Chr.7: 91265061-91265061 on Build GRCh38
rs767058121 Chr.7: 91265200-91265200 on Build GRCh38
rs762437358 Chr.7: 91265409-91265409 on Build GRCh38
rs913305628 Chr.7: 91265246-91265246 on Build GRCh38
rs751566731 Chr.7: 91265221-91265221 on Build GRCh38
rs965728003 Chr.7: 91265138-91265138 on Build GRCh38
rs774814084 Chr.7: 91265203-91265203 on Build GRCh38
rs753260530 Chr.7: 91265126-91265126 on Build GRCh38
rs574691354 Chr.7: 91265091-91265091 on Build GRCh38
rs780894315 Chr.7: 91265458-91265458 on Build GRCh38
rs746616682 Chr.7: 91265401-91265401 on Build GRCh38
rs755035659 Chr.7: 91265355-91265355 on Build GRCh38
rs747634105 Chr.7: 91265459-91265459 on Build GRCh38
rs762347513 Chr.7: 91265318-91265318 on Build GRCh38
rs765731647 Chr.7: 91265321-91265321 on Build GRCh38
rs138935038 Chr.7: 91265237-91265237 on Build GRCh38
rs773037088 Chr.7: 91265175-91265175 on Build GRCh38
rs977241948 Chr.7: 91265092-91265092 on Build GRCh38
rs771826054 Chr.7: 91265381-91265381 on Build GRCh38
rs113491382 Chr.7: 91265420-91265420 on Build GRCh38
rs150690083 Chr.7: 91265213-91265213 on Build GRCh38
rs201693580 Chr.7: 91265219-91265219 on Build GRCh38
rs562020756 Chr.7: 91265106-91265117 on Build GRCh38
rs937596409 Chr.7: 91265407-91265407 on Build GRCh38
rs750709517 Chr.7: 91265322-91265322 on Build GRCh38
rs1046009111 Chr.7: 91265099-91265099 on Build GRCh38
rs1053100439 Chr.7: 91265081-91265081 on Build GRCh38
rs756113937 Chr.7: 91265222-91265222 on Build GRCh38
rs768288689 Chr.7: 91265402-91265402 on Build GRCh38
rs942183361 Chr.7: 91265233-91265233 on Build GRCh38
rs1052225120 Chr.7: 91265416-91265416 on Build GRCh38
rs557713242 Chr.7: 91265101-91265101 on Build GRCh38
rs1004645902 Chr.7: 91265127-91265127 on Build GRCh38

View Full Product Details