Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91267287-91267797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD1
Entrez Gene ID
8321
Gene Name
frizzled class receptor 1
Gene Aliases
-
UniGene
Hs.94234
Gene Chromosome Location
Chr.7: 91264468-91268817 on Build GRCh38
Transcripts
NM_003505.1

SNP Detail

SNP ID Chromosome Location
rs188223046 Chr.7: 91267486-91267486 on Build GRCh38
rs918440988 Chr.7: 91267742-91267743 on Build GRCh38
rs918405279 Chr.7: 91267498-91267498 on Build GRCh38
rs971388126 Chr.7: 91267427-91267427 on Build GRCh38
rs529508992 Chr.7: 91267473-91267473 on Build GRCh38
rs879887652 Chr.7: 91267625-91267625 on Build GRCh38
rs903653463 Chr.7: 91267667-91267667 on Build GRCh38
rs942377923 Chr.7: 91267620-91267620 on Build GRCh38
rs941265687 Chr.7: 91267694-91267694 on Build GRCh38
rs537195277 Chr.7: 91267645-91267645 on Build GRCh38
rs1017843267 Chr.7: 91267530-91267530 on Build GRCh38
rs1007827898 Chr.7: 91267308-91267308 on Build GRCh38
rs571106547 Chr.7: 91267551-91267551 on Build GRCh38
rs941208562 Chr.7: 91267436-91267436 on Build GRCh38
rs1051884116 Chr.7: 91267744-91267744 on Build GRCh38
rs980063277 Chr.7: 91267440-91267440 on Build GRCh38
rs1036868463 Chr.7: 91267452-91267452 on Build GRCh38
rs751867283 Chr.7: 91267696-91267696 on Build GRCh38
rs933799510 Chr.7: 91267742-91267742 on Build GRCh38
rs927394000 Chr.7: 91267441-91267441 on Build GRCh38
rs1038231521 Chr.7: 91267477-91267477 on Build GRCh38
rs981904989 Chr.7: 91267613-91267613 on Build GRCh38
rs909787196 Chr.7: 91267408-91267408 on Build GRCh38
rs777135522 Chr.7: 91267529-91267529 on Build GRCh38
rs1021089661 Chr.7: 91267564-91267564 on Build GRCh38
rs993608702 Chr.7: 91267470-91267470 on Build GRCh38
rs917484039 Chr.7: 91267399-91267399 on Build GRCh38
rs1011076632 Chr.7: 91267563-91267563 on Build GRCh38
rs894056876 Chr.7: 91267542-91267542 on Build GRCh38
rs989479956 Chr.7: 91267378-91267378 on Build GRCh38
rs966491608 Chr.7: 91267379-91267379 on Build GRCh38
rs1030721151 Chr.7: 91267741-91267741 on Build GRCh38
rs546915330 Chr.7: 91267345-91267345 on Build GRCh38
rs990477392 Chr.7: 91267657-91267657 on Build GRCh38
rs958227072 Chr.7: 91267336-91267336 on Build GRCh38
rs3750145 Chr.7: 91267516-91267516 on Build GRCh38
rs967252522 Chr.7: 91267586-91267586 on Build GRCh38
rs941422557 Chr.7: 91267454-91267454 on Build GRCh38
rs192284995 Chr.7: 91267543-91267543 on Build GRCh38
rs79504063 Chr.7: 91267647-91267647 on Build GRCh38
rs927666836 Chr.7: 91267394-91267394 on Build GRCh38
rs891977090 Chr.7: 91267771-91267771 on Build GRCh38
rs183134538 Chr.7: 91267330-91267330 on Build GRCh38
rs1055074876 Chr.7: 91267405-91267405 on Build GRCh38
rs1035164854 Chr.7: 91267617-91267617 on Build GRCh38
rs1051473679 Chr.7: 91267522-91267522 on Build GRCh38
rs1039337689 Chr.7: 91267631-91267631 on Build GRCh38
rs956343548 Chr.7: 91267361-91267361 on Build GRCh38
rs775468853 Chr.7: 91267402-91267402 on Build GRCh38
rs891003302 Chr.7: 91267532-91267532 on Build GRCh38
rs561188251 Chr.7: 91267325-91267325 on Build GRCh38
rs972617395 Chr.7: 91267721-91267721 on Build GRCh38

View Full Product Details