Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92111884-92112416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP51A1
Entrez Gene ID
1595
Gene Name
cytochrome P450 family 51 subfamily A member 1
Gene Aliases
CP51, CYP51, CYPL1, LDM, P450-14DM, P450L1
UniGene
Hs.417077
Gene Chromosome Location
Chr.7: 92112149-92134745 on Build GRCh38
Transcripts
NM_000786.3 NM_001146152.1

SNP Detail

SNP ID Chromosome Location
rs945732473 Chr.7: 92112072-92112072 on Build GRCh38
rs535595810 Chr.7: 92111970-92111970 on Build GRCh38
rs886165754 Chr.7: 92112108-92112108 on Build GRCh38
rs182366870 Chr.7: 92111907-92111907 on Build GRCh38
rs537438608 Chr.7: 92112352-92112352 on Build GRCh38
rs535068396 Chr.7: 92112314-92112314 on Build GRCh38
rs921413944 Chr.7: 92112019-92112019 on Build GRCh38
rs192593756 Chr.7: 92112350-92112350 on Build GRCh38
rs770055639 Chr.7: 92111985-92111985 on Build GRCh38
rs529515441 Chr.7: 92111952-92111953 on Build GRCh38
rs898778081 Chr.7: 92112083-92112083 on Build GRCh38
rs528139567 Chr.7: 92112248-92112249 on Build GRCh38
rs188361781 Chr.7: 92112336-92112336 on Build GRCh38
rs1004602070 Chr.7: 92112168-92112168 on Build GRCh38
rs951426837 Chr.7: 92111948-92111948 on Build GRCh38
rs67387939 Chr.7: 92111999-92112000 on Build GRCh38
rs148296793 Chr.7: 92111997-92112001 on Build GRCh38
rs1024620683 Chr.7: 92111911-92111911 on Build GRCh38
rs992248889 Chr.7: 92112343-92112343 on Build GRCh38
rs370839064 Chr.7: 92111997-92111997 on Build GRCh38
rs368341883 Chr.7: 92111997-92111997 on Build GRCh38
rs568787469 Chr.7: 92112055-92112055 on Build GRCh38
rs992687546 Chr.7: 92111908-92111908 on Build GRCh38
rs545459859 Chr.7: 92112121-92112122 on Build GRCh38
rs770698483 Chr.7: 92112296-92112296 on Build GRCh38
rs375348381 Chr.7: 92112065-92112066 on Build GRCh38
rs773564673 Chr.7: 92112131-92112131 on Build GRCh38
rs1028899847 Chr.7: 92112303-92112303 on Build GRCh38
rs1040149158 Chr.7: 92112181-92112182 on Build GRCh38
rs901677507 Chr.7: 92112233-92112234 on Build GRCh38
rs537419740 Chr.7: 92112122-92112122 on Build GRCh38
rs143809493 Chr.7: 92111998-92111999 on Build GRCh38
rs368397442 Chr.7: 92111966-92111966 on Build GRCh38
rs143034052 Chr.7: 92112015-92112015 on Build GRCh38
rs368415258 Chr.7: 92112021-92112022 on Build GRCh38
rs566160969 Chr.7: 92112084-92112084 on Build GRCh38
rs372503575 Chr.7: 92112106-92112107 on Build GRCh38
rs948267156 Chr.7: 92112071-92112072 on Build GRCh38
rs941417953 Chr.7: 92112003-92112003 on Build GRCh38
rs55776212 Chr.7: 92112204-92112204 on Build GRCh38
rs367999786 Chr.7: 92112002-92112002 on Build GRCh38
rs918102012 Chr.7: 92112070-92112071 on Build GRCh38
rs748947706 Chr.7: 92112250-92112250 on Build GRCh38
rs1051362807 Chr.7: 92112045-92112045 on Build GRCh38
rs751270903 Chr.7: 92112341-92112342 on Build GRCh38
rs1031024130 Chr.7: 92112338-92112339 on Build GRCh38
rs934122439 Chr.7: 92112039-92112039 on Build GRCh38
rs969577521 Chr.7: 92112365-92112366 on Build GRCh38
rs1048740341 Chr.7: 92112085-92112086 on Build GRCh38
rs956744599 Chr.7: 92112339-92112339 on Build GRCh38
rs750814001 Chr.7: 92112048-92112049 on Build GRCh38
rs759494067 Chr.7: 92112330-92112330 on Build GRCh38
rs375581416 Chr.7: 92112249-92112250 on Build GRCh38

View Full Product Details