Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92458445-92458944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD1
Entrez Gene ID
57798
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
CMD2B, ODAG, RG083M05.2
UniGene
Hs.21145
Gene Chromosome Location
Chr.7: 92447448-92494631 on Build GRCh38
Transcripts
NM_021167.4 NR_052016.1

SNP Detail

SNP ID Chromosome Location
rs1053833041 Chr.7: 92458774-92458774 on Build GRCh38
rs548525500 Chr.7: 92458727-92458727 on Build GRCh38
rs142251156 Chr.7: 92458855-92458855 on Build GRCh38
rs961944811 Chr.7: 92458776-92458776 on Build GRCh38
rs1012578543 Chr.7: 92458835-92458835 on Build GRCh38
rs1023089561 Chr.7: 92458756-92458756 on Build GRCh38
rs930415634 Chr.7: 92458538-92458538 on Build GRCh38
rs540316918 Chr.7: 92458831-92458831 on Build GRCh38
rs893052326 Chr.7: 92458879-92458879 on Build GRCh38
rs549731604 Chr.7: 92458674-92458674 on Build GRCh38
rs945588452 Chr.7: 92458781-92458784 on Build GRCh38
rs776577594 Chr.7: 92458529-92458529 on Build GRCh38
rs1055343923 Chr.7: 92458832-92458832 on Build GRCh38
rs368448866 Chr.7: 92458897-92458897 on Build GRCh38
rs535394734 Chr.7: 92458689-92458689 on Build GRCh38
rs1013015200 Chr.7: 92458741-92458741 on Build GRCh38
rs998768784 Chr.7: 92458701-92458701 on Build GRCh38
rs371632143 Chr.7: 92458591-92458591 on Build GRCh38
rs1054347235 Chr.7: 92458869-92458869 on Build GRCh38
rs77607170 Chr.7: 92458828-92458828 on Build GRCh38
rs745459455 Chr.7: 92458551-92458551 on Build GRCh38
rs888787863 Chr.7: 92458641-92458641 on Build GRCh38
rs565335378 Chr.7: 92458755-92458755 on Build GRCh38
rs1012796414 Chr.7: 92458900-92458900 on Build GRCh38
rs183702741 Chr.7: 92458601-92458601 on Build GRCh38
rs957189336 Chr.7: 92458726-92458726 on Build GRCh38
rs919212143 Chr.7: 92458630-92458630 on Build GRCh38
rs1022786670 Chr.7: 92458918-92458918 on Build GRCh38
rs954530115 Chr.7: 92458883-92458884 on Build GRCh38
rs552759747 Chr.7: 92458508-92458508 on Build GRCh38
rs908528284 Chr.7: 92458773-92458773 on Build GRCh38
rs551892949 Chr.7: 92458528-92458528 on Build GRCh38
rs981466488 Chr.7: 92458759-92458759 on Build GRCh38
rs920089051 Chr.7: 92458808-92458808 on Build GRCh38
rs1050095487 Chr.7: 92458547-92458547 on Build GRCh38
rs944441460 Chr.7: 92458834-92458834 on Build GRCh38
rs1033270772 Chr.7: 92458713-92458713 on Build GRCh38
rs191480624 Chr.7: 92458507-92458507 on Build GRCh38
rs1026586352 Chr.7: 92458880-92458880 on Build GRCh38
rs573181080 Chr.7: 92458652-92458652 on Build GRCh38
rs971468493 Chr.7: 92458758-92458758 on Build GRCh38
rs376068569 Chr.7: 92458751-92458751 on Build GRCh38
rs999196305 Chr.7: 92458867-92458867 on Build GRCh38
rs974329921 Chr.7: 92458795-92458795 on Build GRCh38
rs1038973862 Chr.7: 92458484-92458484 on Build GRCh38
rs773865764 Chr.7: 92458472-92458472 on Build GRCh38
rs188250836 Chr.7: 92458686-92458686 on Build GRCh38
rs1045701 Chr.7: 92458782-92458782 on Build GRCh38
rs34606827 Chr.7: 92458550-92458551 on Build GRCh38
rs796333236 Chr.7: 92458829-92458829 on Build GRCh38
rs915309687 Chr.7: 92458778-92458778 on Build GRCh38
rs557418474 Chr.7: 92458852-92458852 on Build GRCh38

View Full Product Details