Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92565928-92566337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133B
Entrez Gene ID
257415
Gene Name
family with sequence similarity 133 member B
Gene Aliases
-
UniGene
Hs.18564
Gene Chromosome Location
Chr.7: 92560758-92590394 on Build GRCh38
Transcripts
NM_001040057.2 NM_001288584.1 NM_152789.3 NR_109929.1 XM_011516004.2 XM_011516005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5175090 c.628A>G p.K210E Chr.7: 92566043-92566043 on Build GRCh38
4682551 c.623delA p.K208fs*>40 Chr.7: 92566048-92566048 on Build GRCh38
1093074 c.613C>T p.R205* Chr.7: 92566058-92566058 on Build GRCh38
86648 c.644G>A p.R215Q Chr.7: 92566027-92566027 on Build GRCh38
1452853 c.651delA p.A218fs*>30 Chr.7: 92566020-92566020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371337331 Chr.7: 92565998-92565998 on Build GRCh38
rs907482366 Chr.7: 92566120-92566120 on Build GRCh38
rs780942723 Chr.7: 92566013-92566013 on Build GRCh38
rs759015531 Chr.7: 92565981-92565981 on Build GRCh38
rs773930695 Chr.7: 92565979-92565979 on Build GRCh38
rs763516747 Chr.7: 92565967-92565967 on Build GRCh38
rs369564208 Chr.7: 92566286-92566286 on Build GRCh38
rs753651950 Chr.7: 92566039-92566039 on Build GRCh38
rs560707662 Chr.7: 92566062-92566062 on Build GRCh38
rs913224319 Chr.7: 92566046-92566046 on Build GRCh38
rs1038112027 Chr.7: 92566244-92566244 on Build GRCh38
rs548929405 Chr.7: 92566247-92566247 on Build GRCh38
rs769551351 Chr.7: 92566276-92566276 on Build GRCh38
rs1007201778 Chr.7: 92566273-92566273 on Build GRCh38
rs924451317 Chr.7: 92565997-92565997 on Build GRCh38
rs773647103 Chr.7: 92565960-92565960 on Build GRCh38
rs771444019 Chr.7: 92566087-92566087 on Build GRCh38
rs770073370 Chr.7: 92566066-92566066 on Build GRCh38
rs770214453 Chr.7: 92565955-92565955 on Build GRCh38
rs957467424 Chr.7: 92566027-92566027 on Build GRCh38
rs995654371 Chr.7: 92566211-92566211 on Build GRCh38
rs981669149 Chr.7: 92566096-92566096 on Build GRCh38
rs749705254 Chr.7: 92566084-92566084 on Build GRCh38
rs763994795 Chr.7: 92566009-92566009 on Build GRCh38
rs202240462 Chr.7: 92565983-92565983 on Build GRCh38
rs575929837 Chr.7: 92566259-92566259 on Build GRCh38
rs182615762 Chr.7: 92566058-92566058 on Build GRCh38
rs190768063 Chr.7: 92565984-92565984 on Build GRCh38
rs938946581 Chr.7: 92566138-92566138 on Build GRCh38
rs887982508 Chr.7: 92566177-92566177 on Build GRCh38
rs779950514 Chr.7: 92566057-92566057 on Build GRCh38
rs748485225 Chr.7: 92565953-92565953 on Build GRCh38
rs781392045 Chr.7: 92566061-92566061 on Build GRCh38
rs755959190 Chr.7: 92565999-92565999 on Build GRCh38
rs1037263593 Chr.7: 92566158-92566158 on Build GRCh38
rs188012495 Chr.7: 92566161-92566161 on Build GRCh38
rs757252043 Chr.7: 92566053-92566053 on Build GRCh38
rs77651911 Chr.7: 92566264-92566264 on Build GRCh38
rs941095462 Chr.7: 92566180-92566180 on Build GRCh38
rs115025261 Chr.7: 92565973-92565973 on Build GRCh38
rs780573741 Chr.7: 92566065-92566066 on Build GRCh38
rs1007251668 Chr.7: 92566299-92566299 on Build GRCh38
rs774758734 Chr.7: 92566102-92566102 on Build GRCh38
rs749616641 Chr.7: 92566101-92566102 on Build GRCh38
rs369266600 Chr.7: 92566073-92566073 on Build GRCh38
rs763822144 Chr.7: 92566179-92566179 on Build GRCh38

View Full Product Details