Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92607964-92608477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK6
Entrez Gene ID
1021
Gene Name
cyclin dependent kinase 6
Gene Aliases
MCPH12, PLSTIRE
UniGene
Hs.119882
Gene Chromosome Location
Chr.7: 92604921-92836627 on Build GRCh38
Transcripts
NM_001145306.1 NM_001259.6 XM_006715835.2

SNP Detail

SNP ID Chromosome Location
rs42033 Chr.7: 92608219-92608219 on Build GRCh38
rs760890420 Chr.7: 92608439-92608439 on Build GRCh38
rs191304750 Chr.7: 92608050-92608050 on Build GRCh38
rs961245275 Chr.7: 92608270-92608270 on Build GRCh38
rs42031 Chr.7: 92608082-92608082 on Build GRCh38
rs944213213 Chr.7: 92608130-92608130 on Build GRCh38
rs940980577 Chr.7: 92608104-92608104 on Build GRCh38
rs980782868 Chr.7: 92608025-92608025 on Build GRCh38
rs34956279 Chr.7: 92608292-92608293 on Build GRCh38
rs901602420 Chr.7: 92608174-92608174 on Build GRCh38
rs535913719 Chr.7: 92608011-92608011 on Build GRCh38
rs982545588 Chr.7: 92608357-92608357 on Build GRCh38
rs575640163 Chr.7: 92608330-92608330 on Build GRCh38
rs903565500 Chr.7: 92608226-92608226 on Build GRCh38
rs950820420 Chr.7: 92608331-92608331 on Build GRCh38
rs1002146592 Chr.7: 92608261-92608261 on Build GRCh38
rs1026984790 Chr.7: 92608400-92608400 on Build GRCh38
rs570070010 Chr.7: 92608009-92608009 on Build GRCh38
rs187310427 Chr.7: 92608194-92608194 on Build GRCh38
rs909605486 Chr.7: 92608360-92608360 on Build GRCh38
rs777499353 Chr.7: 92608098-92608098 on Build GRCh38
rs568236889 Chr.7: 92608055-92608055 on Build GRCh38
rs113665429 Chr.7: 92608155-92608155 on Build GRCh38
rs1051369878 Chr.7: 92608156-92608156 on Build GRCh38
rs1022298079 Chr.7: 92608230-92608230 on Build GRCh38
rs376837031 Chr.7: 92608309-92608309 on Build GRCh38
rs42032 Chr.7: 92608112-92608112 on Build GRCh38
rs921232214 Chr.7: 92608419-92608419 on Build GRCh38
rs1026904131 Chr.7: 92608328-92608328 on Build GRCh38
rs908703482 Chr.7: 92608117-92608117 on Build GRCh38
rs1013856795 Chr.7: 92608321-92608321 on Build GRCh38
rs892586115 Chr.7: 92608198-92608198 on Build GRCh38
rs975469345 Chr.7: 92608416-92608416 on Build GRCh38
rs561467795 Chr.7: 92608423-92608423 on Build GRCh38
rs192660194 Chr.7: 92608392-92608392 on Build GRCh38
rs188032819 Chr.7: 92608005-92608005 on Build GRCh38
rs1033958471 Chr.7: 92608262-92608262 on Build GRCh38
rs560489680 Chr.7: 92608226-92608226 on Build GRCh38
rs3814100 Chr.7: 92608062-92608062 on Build GRCh38
rs899736165 Chr.7: 92608151-92608151 on Build GRCh38
rs968040450 Chr.7: 92608237-92608237 on Build GRCh38
rs969450628 Chr.7: 92608376-92608377 on Build GRCh38

View Full Product Details