Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92611580-92612082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK6
Entrez Gene ID
1021
Gene Name
cyclin dependent kinase 6
Gene Aliases
MCPH12, PLSTIRE
UniGene
Hs.119882
Gene Chromosome Location
Chr.7: 92604921-92836627 on Build GRCh38
Transcripts
NM_001145306.1 NM_001259.6 XM_006715835.2

SNP Detail

SNP ID Chromosome Location
rs972630410 Chr.7: 92611697-92611697 on Build GRCh38
rs942479379 Chr.7: 92611973-92611973 on Build GRCh38
rs1033250148 Chr.7: 92611746-92611746 on Build GRCh38
rs972383782 Chr.7: 92611927-92611927 on Build GRCh38
rs1009400470 Chr.7: 92611691-92611691 on Build GRCh38
rs537099893 Chr.7: 92612028-92612028 on Build GRCh38
rs1041937925 Chr.7: 92611808-92611808 on Build GRCh38
rs563109834 Chr.7: 92611804-92611804 on Build GRCh38
rs947282943 Chr.7: 92612035-92612035 on Build GRCh38
rs920556373 Chr.7: 92611918-92611918 on Build GRCh38
rs971720318 Chr.7: 92611905-92611905 on Build GRCh38
rs571221962 Chr.7: 92612013-92612013 on Build GRCh38
rs1050324012 Chr.7: 92611987-92611987 on Build GRCh38
rs940362115 Chr.7: 92611874-92611874 on Build GRCh38
rs991707352 Chr.7: 92611791-92611791 on Build GRCh38
rs180945892 Chr.7: 92611840-92611840 on Build GRCh38
rs927682771 Chr.7: 92611846-92611846 on Build GRCh38
rs528429629 Chr.7: 92612005-92612005 on Build GRCh38
rs909482683 Chr.7: 92611671-92611671 on Build GRCh38
rs1023556042 Chr.7: 92611799-92611799 on Build GRCh38
rs998802493 Chr.7: 92611735-92611735 on Build GRCh38
rs74980371 Chr.7: 92611948-92611948 on Build GRCh38
rs893029141 Chr.7: 92611783-92611783 on Build GRCh38
rs766236563 Chr.7: 92611854-92611854 on Build GRCh38
rs544309707 Chr.7: 92611700-92611700 on Build GRCh38
rs958672942 Chr.7: 92611865-92611865 on Build GRCh38
rs888919204 Chr.7: 92611683-92611683 on Build GRCh38
rs1039565529 Chr.7: 92611755-92611755 on Build GRCh38
rs971564092 Chr.7: 92611817-92611817 on Build GRCh38
rs929714659 Chr.7: 92611727-92611727 on Build GRCh38
rs957856703 Chr.7: 92611752-92611752 on Build GRCh38
rs912322450 Chr.7: 92611961-92611961 on Build GRCh38
rs912189430 Chr.7: 92611748-92611748 on Build GRCh38
rs765456689 Chr.7: 92611800-92611800 on Build GRCh38
rs762653269 Chr.7: 92611853-92611853 on Build GRCh38
rs1041642769 Chr.7: 92612037-92612037 on Build GRCh38
rs773483142 Chr.7: 92611831-92611831 on Build GRCh38
rs954877014 Chr.7: 92611932-92611932 on Build GRCh38
rs1016297655 Chr.7: 92611890-92611890 on Build GRCh38
rs34855746 Chr.7: 92611621-92611622 on Build GRCh38
rs549190618 Chr.7: 92611866-92611866 on Build GRCh38
rs551083861 Chr.7: 92612007-92612007 on Build GRCh38
rs991701675 Chr.7: 92611668-92611668 on Build GRCh38
rs1018962149 Chr.7: 92611694-92611694 on Build GRCh38

View Full Product Details