Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93258230-93258775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS50
Entrez Gene ID
55610
Gene Name
VPS50, EARP/GARPII complex subunit
Gene Aliases
CCDC132, VPS54L
UniGene
Hs.202424
Gene Chromosome Location
Chr.7: 93232340-93361123 on Build GRCh38
Transcripts
NM_001257998.1 NM_017667.3 NM_024553.2 XM_011516395.2 XM_011516396.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3431876 c.545G>T p.R182I Chr.7: 93258361-93258361 on Build GRCh38
5671863 c.501G>C p.L167L Chr.7: 93258237-93258237 on Build GRCh38
6621498 c.413T>C p.I138T Chr.7: 93258239-93258239 on Build GRCh38
6621459 c.565G>T p.E189* Chr.7: 93258381-93258381 on Build GRCh38
6621458 c.565G>T p.E189* Chr.7: 93258381-93258381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900790701 Chr.7: 93258550-93258550 on Build GRCh38
rs756724528 Chr.7: 93258348-93258348 on Build GRCh38
rs768773422 Chr.7: 93258399-93258410 on Build GRCh38
rs1024780406 Chr.7: 93258690-93258690 on Build GRCh38
rs753230941 Chr.7: 93258347-93258347 on Build GRCh38
rs373803205 Chr.7: 93258551-93258551 on Build GRCh38
rs1041810858 Chr.7: 93258628-93258628 on Build GRCh38
rs771909828 Chr.7: 93258270-93258270 on Build GRCh38
rs370687593 Chr.7: 93258432-93258432 on Build GRCh38
rs932235014 Chr.7: 93258560-93258560 on Build GRCh38
rs781727232 Chr.7: 93258319-93258319 on Build GRCh38
rs1037957422 Chr.7: 93258529-93258529 on Build GRCh38
rs367917813 Chr.7: 93258349-93258349 on Build GRCh38
rs766462487 Chr.7: 93258307-93258307 on Build GRCh38
rs542617378 Chr.7: 93258468-93258468 on Build GRCh38
rs868435151 Chr.7: 93258632-93258632 on Build GRCh38
rs775404430 Chr.7: 93258276-93258276 on Build GRCh38
rs145621721 Chr.7: 93258372-93258372 on Build GRCh38
rs758681295 Chr.7: 93258400-93258402 on Build GRCh38
rs545089795 Chr.7: 93258572-93258572 on Build GRCh38
rs28461604 Chr.7: 93258301-93258301 on Build GRCh38
rs773550941 Chr.7: 93258528-93258528 on Build GRCh38
rs765280769 Chr.7: 93258286-93258286 on Build GRCh38
rs1053013799 Chr.7: 93258625-93258625 on Build GRCh38
rs961944275 Chr.7: 93258423-93258423 on Build GRCh38
rs563288707 Chr.7: 93258629-93258629 on Build GRCh38
rs763441201 Chr.7: 93258653-93258653 on Build GRCh38
rs751809989 Chr.7: 93258309-93258309 on Build GRCh38
rs1012597561 Chr.7: 93258685-93258685 on Build GRCh38
rs137864226 Chr.7: 93258656-93258656 on Build GRCh38
rs917745896 Chr.7: 93258385-93258385 on Build GRCh38
rs529585863 Chr.7: 93258693-93258693 on Build GRCh38
rs770491697 Chr.7: 93258420-93258420 on Build GRCh38
rs763125093 Chr.7: 93258294-93258294 on Build GRCh38
rs774070732 Chr.7: 93258259-93258259 on Build GRCh38
rs573596063 Chr.7: 93258433-93258433 on Build GRCh38
rs983619673 Chr.7: 93258453-93258453 on Build GRCh38
rs755291881 Chr.7: 93258318-93258318 on Build GRCh38
rs1010290822 Chr.7: 93258608-93258608 on Build GRCh38
rs759198008 Chr.7: 93258268-93258268 on Build GRCh38
rs762995862 Chr.7: 93258294-93258294 on Build GRCh38
rs552956843 Chr.7: 93258504-93258504 on Build GRCh38
rs760666012 Chr.7: 93258282-93258282 on Build GRCh38
rs561614840 Chr.7: 93258734-93258734 on Build GRCh38
rs750543565 Chr.7: 93258287-93258287 on Build GRCh38
rs1000191947 Chr.7: 93258652-93258652 on Build GRCh38
rs890635316 Chr.7: 93258575-93258575 on Build GRCh38
rs184715558 Chr.7: 93258512-93258512 on Build GRCh38
rs959280982 Chr.7: 93258666-93258666 on Build GRCh38
rs748754141 Chr.7: 93258390-93258390 on Build GRCh38
rs951887037 Chr.7: 93258298-93258298 on Build GRCh38
rs776115213 Chr.7: 93258293-93258294 on Build GRCh38
rs925455807 Chr.7: 93258565-93258565 on Build GRCh38
rs544623059 Chr.7: 93258663-93258663 on Build GRCh38
rs904513903 Chr.7: 93258634-93258634 on Build GRCh38

View Full Product Details