Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93906368-93906909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNGT1
Entrez Gene ID
2792
Gene Name
G protein subunit gamma transducin 1
Gene Aliases
GNG1
UniGene
Hs.732212
Gene Chromosome Location
Chr.7: 93906508-93911173 on Build GRCh38
Transcripts
XM_017012009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4412307 c.96+4A>C p.? Chr.7: 93906846-93906846 on Build GRCh38
1635495 c.48G>A p.K16K Chr.7: 93906794-93906794 on Build GRCh38
1635496 c.49A>T p.M17L Chr.7: 93906795-93906795 on Build GRCh38
747465 c.12C>T p.I4I Chr.7: 93906758-93906758 on Build GRCh38
1673307 c.89G>T p.R30I Chr.7: 93906835-93906835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048252567 Chr.7: 93906715-93906715 on Build GRCh38
rs1037993267 Chr.7: 93906782-93906782 on Build GRCh38
rs1047732307 Chr.7: 93906577-93906580 on Build GRCh38
rs900753143 Chr.7: 93906800-93906800 on Build GRCh38
rs989082533 Chr.7: 93906514-93906514 on Build GRCh38
rs778843372 Chr.7: 93906746-93906746 on Build GRCh38
rs373250765 Chr.7: 93906456-93906456 on Build GRCh38
rs917947375 Chr.7: 93906501-93906501 on Build GRCh38
rs191685430 Chr.7: 93906755-93906755 on Build GRCh38
rs142704200 Chr.7: 93906425-93906425 on Build GRCh38
rs781074304 Chr.7: 93906809-93906809 on Build GRCh38
rs150956934 Chr.7: 93906803-93906803 on Build GRCh38
rs866854789 Chr.7: 93906556-93906556 on Build GRCh38
rs866397447 Chr.7: 93906663-93906663 on Build GRCh38
rs779917603 Chr.7: 93906464-93906464 on Build GRCh38
rs763396965 Chr.7: 93906884-93906884 on Build GRCh38
rs571963650 Chr.7: 93906725-93906725 on Build GRCh38
rs913079471 Chr.7: 93906550-93906550 on Build GRCh38
rs769341718 Chr.7: 93906721-93906723 on Build GRCh38
rs755999687 Chr.7: 93906823-93906823 on Build GRCh38
rs376703142 Chr.7: 93906751-93906751 on Build GRCh38
rs1029987313 Chr.7: 93906821-93906821 on Build GRCh38
rs752594162 Chr.7: 93906822-93906822 on Build GRCh38
rs768944512 Chr.7: 93906761-93906761 on Build GRCh38
rs746027091 Chr.7: 93906750-93906750 on Build GRCh38
rs751213457 Chr.7: 93906805-93906805 on Build GRCh38
rs193921133 Chr.7: 93906843-93906843 on Build GRCh38
rs375007032 Chr.7: 93906698-93906698 on Build GRCh38
rs768926345 Chr.7: 93906846-93906846 on Build GRCh38
rs772100965 Chr.7: 93906837-93906837 on Build GRCh38
rs147350641 Chr.7: 93906587-93906587 on Build GRCh38
rs77942026 Chr.7: 93906433-93906433 on Build GRCh38
rs558030488 Chr.7: 93906420-93906420 on Build GRCh38
rs978655456 Chr.7: 93906583-93906583 on Build GRCh38
rs200035274 Chr.7: 93906828-93906828 on Build GRCh38
rs542579365 Chr.7: 93906615-93906615 on Build GRCh38
rs954469962 Chr.7: 93906512-93906512 on Build GRCh38
rs772250512 Chr.7: 93906752-93906752 on Build GRCh38
rs947186509 Chr.7: 93906568-93906568 on Build GRCh38
rs775641362 Chr.7: 93906754-93906754 on Build GRCh38
rs773715853 Chr.7: 93906881-93906881 on Build GRCh38
rs762577190 Chr.7: 93906801-93906801 on Build GRCh38
rs780114005 Chr.7: 93906838-93906838 on Build GRCh38
rs1006361572 Chr.7: 93906774-93906774 on Build GRCh38
rs564346446 Chr.7: 93906546-93906546 on Build GRCh38
rs747966528 Chr.7: 93906738-93906738 on Build GRCh38
rs1016486956 Chr.7: 93906392-93906392 on Build GRCh38
rs1055030712 Chr.7: 93906627-93906627 on Build GRCh38
rs964382506 Chr.7: 93906395-93906395 on Build GRCh38
rs765842422 Chr.7: 93906783-93906783 on Build GRCh38
rs781161398 Chr.7: 93906708-93906708 on Build GRCh38
rs369246838 Chr.7: 93906877-93906877 on Build GRCh38
rs141065761 Chr.7: 93906807-93906807 on Build GRCh38
rs540836125 Chr.7: 93906741-93906741 on Build GRCh38
rs573732840 Chr.7: 93906598-93906598 on Build GRCh38
rs561669621 Chr.7: 93906695-93906695 on Build GRCh38
rs980863228 Chr.7: 93906536-93906536 on Build GRCh38
rs1030534539 Chr.7: 93906492-93906492 on Build GRCh38
rs202009903 Chr.7: 93906842-93906842 on Build GRCh38
rs944294670 Chr.7: 93906654-93906654 on Build GRCh38
rs776858019 Chr.7: 93906859-93906859 on Build GRCh38
rs948133655 Chr.7: 93906511-93906512 on Build GRCh38
rs904326580 Chr.7: 93906784-93906784 on Build GRCh38
rs762282560 Chr.7: 93906863-93906863 on Build GRCh38
rs938050205 Chr.7: 93906606-93906606 on Build GRCh38
rs1040718977 Chr.7: 93906679-93906679 on Build GRCh38
rs776996322 Chr.7: 93906775-93906775 on Build GRCh38

View Full Product Details