Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97009654-97010177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX6
Entrez Gene ID
1750
Gene Name
distal-less homeobox 6
Gene Aliases
-
UniGene
Hs.249196
Gene Chromosome Location
Chr.7: 97005978-97011040 on Build GRCh38
Transcripts
NM_005222.3 NR_015448.1

Gene Symbol
DLX6-AS1
Entrez Gene ID
285987
Gene Name
DLX6 antisense RNA 1
Gene Aliases
DLX6-AS, DLX6AS, Evf-2, NCRNA00212
UniGene
Hs.34969
Gene Chromosome Location
Chr.7: 96968515-97014065 on Build GRCh38
Transcripts
NM_005222.3 NR_015448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919163 c.649A>T p.N217Y Chr.7: 97009814-97009814 on Build GRCh38
6292328 c.672T>A p.P224P Chr.7: 97009921-97009921 on Build GRCh38
2865817 c.858G>A p.T286T Chr.7: 97010023-97010023 on Build GRCh38
4677948 c.739C>T p.R247C Chr.7: 97009904-97009904 on Build GRCh38
3698616 c.740G>A p.R247H Chr.7: 97009905-97009905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376564037 Chr.7: 97010023-97010023 on Build GRCh38
rs575323334 Chr.7: 97010100-97010100 on Build GRCh38
rs185805944 Chr.7: 97009887-97009887 on Build GRCh38
rs771847061 Chr.7: 97009682-97009688 on Build GRCh38
rs1031229299 Chr.7: 97010117-97010117 on Build GRCh38
rs756327965 Chr.7: 97010049-97010049 on Build GRCh38
rs916681741 Chr.7: 97009955-97009955 on Build GRCh38
rs868516782 Chr.7: 97009994-97009994 on Build GRCh38
rs557570706 Chr.7: 97010119-97010119 on Build GRCh38
rs368469391 Chr.7: 97010076-97010076 on Build GRCh38
rs565262452 Chr.7: 97009876-97009876 on Build GRCh38
rs189277896 Chr.7: 97009685-97009685 on Build GRCh38
rs201772433 Chr.7: 97009914-97009914 on Build GRCh38
rs756020861 Chr.7: 97010148-97010148 on Build GRCh38
rs757114145 Chr.7: 97009908-97009908 on Build GRCh38
rs745808846 Chr.7: 97009913-97009913 on Build GRCh38
rs781159910 Chr.7: 97009915-97009915 on Build GRCh38
rs377688666 Chr.7: 97009945-97009945 on Build GRCh38
rs180982495 Chr.7: 97009770-97009770 on Build GRCh38
rs369370543 Chr.7: 97009888-97009888 on Build GRCh38
rs760073542 Chr.7: 97009900-97009900 on Build GRCh38
rs1002017487 Chr.7: 97010113-97010113 on Build GRCh38
rs373065383 Chr.7: 97010011-97010011 on Build GRCh38
rs780748008 Chr.7: 97009822-97009822 on Build GRCh38
rs776717617 Chr.7: 97009882-97009882 on Build GRCh38
rs766818524 Chr.7: 97009891-97009891 on Build GRCh38
rs778865296 Chr.7: 97009871-97009871 on Build GRCh38
rs373835215 Chr.7: 97009740-97009740 on Build GRCh38
rs763593085 Chr.7: 97009989-97009989 on Build GRCh38
rs768678104 Chr.7: 97009873-97009873 on Build GRCh38
rs3213654 Chr.7: 97010056-97010056 on Build GRCh38
rs752083204 Chr.7: 97009766-97009766 on Build GRCh38
rs746293523 Chr.7: 97010060-97010060 on Build GRCh38
rs1053806008 Chr.7: 97009692-97009695 on Build GRCh38
rs772295690 Chr.7: 97010075-97010075 on Build GRCh38
rs567341724 Chr.7: 97009925-97009925 on Build GRCh38
rs759993113 Chr.7: 97009950-97009950 on Build GRCh38
rs530888638 Chr.7: 97009899-97009899 on Build GRCh38
rs201287967 Chr.7: 97009691-97009691 on Build GRCh38
rs370326007 Chr.7: 97009965-97009965 on Build GRCh38
rs754487236 Chr.7: 97009773-97009773 on Build GRCh38
rs753549738 Chr.7: 97009996-97009996 on Build GRCh38
rs771116990 Chr.7: 97010057-97010057 on Build GRCh38
rs760229000 Chr.7: 97010088-97010089 on Build GRCh38
rs538651211 Chr.7: 97010050-97010050 on Build GRCh38
rs752212160 Chr.7: 97009904-97009904 on Build GRCh38
rs559368037 Chr.7: 97009864-97009864 on Build GRCh38
rs772656443 Chr.7: 97009784-97009784 on Build GRCh38
rs948164608 Chr.7: 97009960-97009960 on Build GRCh38
rs755697691 Chr.7: 97009853-97009853 on Build GRCh38
rs750189411 Chr.7: 97010022-97010022 on Build GRCh38
rs376417071 Chr.7: 97009906-97009906 on Build GRCh38
rs895180273 Chr.7: 97009699-97009699 on Build GRCh38
rs886308619 Chr.7: 97009881-97009881 on Build GRCh38
rs984670364 Chr.7: 97009987-97009987 on Build GRCh38
rs769967866 Chr.7: 97009933-97009933 on Build GRCh38
rs762709913 Chr.7: 97009730-97009730 on Build GRCh38
rs571735734 Chr.7: 97009892-97009892 on Build GRCh38
rs763456945 Chr.7: 97009890-97009890 on Build GRCh38
rs749033297 Chr.7: 97009867-97009867 on Build GRCh38
rs74517213 Chr.7: 97009824-97009824 on Build GRCh38
rs766940860 Chr.7: 97009750-97009750 on Build GRCh38
rs1050431898 Chr.7: 97010117-97010134 on Build GRCh38
rs73404534 Chr.7: 97010118-97010118 on Build GRCh38
rs747812566 Chr.7: 97009828-97009828 on Build GRCh38
rs149451437 Chr.7: 97009698-97009698 on Build GRCh38
rs373223835 Chr.7: 97009905-97009905 on Build GRCh38
rs774601592 Chr.7: 97010058-97010058 on Build GRCh38
rs1014639142 Chr.7: 97010139-97010139 on Build GRCh38
rs997883699 Chr.7: 97009875-97009875 on Build GRCh38
rs759003436 Chr.7: 97009748-97009748 on Build GRCh38
rs1002091017 Chr.7: 97009910-97009910 on Build GRCh38
rs761942448 Chr.7: 97009885-97009885 on Build GRCh38
rs948095634 Chr.7: 97009721-97009721 on Build GRCh38
rs906520840 Chr.7: 97009741-97009741 on Build GRCh38
rs573892022 Chr.7: 97010133-97010133 on Build GRCh38
rs370659738 Chr.7: 97010029-97010029 on Build GRCh38
rs778760066 Chr.7: 97009909-97009909 on Build GRCh38
rs563706652 Chr.7: 97009689-97009689 on Build GRCh38
rs768021315 Chr.7: 97009964-97009964 on Build GRCh38
rs536599566 Chr.7: 97009968-97009968 on Build GRCh38
rs371198246 Chr.7: 97009870-97009870 on Build GRCh38
rs1043512895 Chr.7: 97010099-97010099 on Build GRCh38
rs773608335 Chr.7: 97009934-97009934 on Build GRCh38
rs766948280 Chr.7: 97009902-97009902 on Build GRCh38
rs746908528 Chr.7: 97010043-97010043 on Build GRCh38
rs916757731 Chr.7: 97010134-97010134 on Build GRCh38
rs763651356 Chr.7: 97009869-97009869 on Build GRCh38
rs575211886 Chr.7: 97009676-97009676 on Build GRCh38
rs374453064 Chr.7: 97009944-97009944 on Build GRCh38
rs978302427 Chr.7: 97009682-97009695 on Build GRCh38
rs553310103 Chr.7: 97010006-97010006 on Build GRCh38
rs954384692 Chr.7: 97010105-97010105 on Build GRCh38
rs924136972 Chr.7: 97009683-97009683 on Build GRCh38
rs1052114929 Chr.7: 97010074-97010074 on Build GRCh38

View Full Product Details