Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98291420-98291918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
UniGene
Hs.656063
Gene Chromosome Location
Chr.7: 98291650-98401115 on Build GRCh38
Transcripts
NM_018842.4 NM_001159491.1 NM_015379.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
UniGene
Hs.567438
Gene Chromosome Location
Chr.7: 98281667-98322398 on Build GRCh38
Transcripts
NM_018842.4 NM_001159491.1 NM_015379.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

SNP Detail

SNP ID Chromosome Location
rs574550552 Chr.7: 98291875-98291875 on Build GRCh38
rs184893730 Chr.7: 98291808-98291808 on Build GRCh38
rs924842633 Chr.7: 98291799-98291799 on Build GRCh38
rs890831865 Chr.7: 98291589-98291589 on Build GRCh38
rs941247740 Chr.7: 98291551-98291551 on Build GRCh38
rs1002746941 Chr.7: 98291699-98291699 on Build GRCh38
rs998953618 Chr.7: 98291468-98291470 on Build GRCh38
rs934122520 Chr.7: 98291766-98291766 on Build GRCh38
rs925936861 Chr.7: 98291466-98291466 on Build GRCh38
rs576242986 Chr.7: 98291599-98291599 on Build GRCh38
rs568197007 Chr.7: 98291591-98291591 on Build GRCh38
rs1033193101 Chr.7: 98291668-98291668 on Build GRCh38
rs909001436 Chr.7: 98291719-98291719 on Build GRCh38
rs543253710 Chr.7: 98291827-98291827 on Build GRCh38
rs941152688 Chr.7: 98291778-98291778 on Build GRCh38
rs1140916 Chr.7: 98291756-98291756 on Build GRCh38
rs982642350 Chr.7: 98291726-98291726 on Build GRCh38
rs1044421919 Chr.7: 98291833-98291833 on Build GRCh38
rs1022743776 Chr.7: 98291686-98291686 on Build GRCh38
rs763430907 Chr.7: 98291579-98291579 on Build GRCh38
rs981105249 Chr.7: 98291536-98291536 on Build GRCh38
rs772783029 Chr.7: 98291675-98291675 on Build GRCh38
rs950172595 Chr.7: 98291713-98291713 on Build GRCh38
rs1022858224 Chr.7: 98291509-98291510 on Build GRCh38
rs967627713 Chr.7: 98291632-98291632 on Build GRCh38
rs899518337 Chr.7: 98291572-98291572 on Build GRCh38
rs1012964185 Chr.7: 98291833-98291834 on Build GRCh38
rs764990846 Chr.7: 98291593-98291594 on Build GRCh38
rs950674994 Chr.7: 98291881-98291882 on Build GRCh38
rs186701941 Chr.7: 98291590-98291590 on Build GRCh38
rs557095284 Chr.7: 98291640-98291640 on Build GRCh38
rs991720823 Chr.7: 98291488-98291488 on Build GRCh38
rs191511048 Chr.7: 98291690-98291690 on Build GRCh38
rs190991514 Chr.7: 98291546-98291546 on Build GRCh38
rs903152431 Chr.7: 98291465-98291465 on Build GRCh38
rs182105881 Chr.7: 98291547-98291547 on Build GRCh38
rs949915577 Chr.7: 98291525-98291525 on Build GRCh38
rs1051357549 Chr.7: 98291767-98291767 on Build GRCh38
rs558163252 Chr.7: 98291736-98291736 on Build GRCh38
rs1026164905 Chr.7: 98291702-98291702 on Build GRCh38
rs540076335 Chr.7: 98291714-98291714 on Build GRCh38
rs967594943 Chr.7: 98291456-98291456 on Build GRCh38
rs1038308549 Chr.7: 98291786-98291786 on Build GRCh38
rs115393163 Chr.7: 98291578-98291578 on Build GRCh38
rs563450266 Chr.7: 98291464-98291464 on Build GRCh38
rs890906346 Chr.7: 98291823-98291823 on Build GRCh38
rs1051310293 Chr.7: 98291612-98291612 on Build GRCh38
rs979000517 Chr.7: 98291633-98291633 on Build GRCh38
rs576811626 Chr.7: 98291738-98291738 on Build GRCh38
rs548377152 Chr.7: 98291458-98291458 on Build GRCh38
rs903204421 Chr.7: 98291635-98291635 on Build GRCh38
rs893194423 Chr.7: 98291666-98291666 on Build GRCh38
rs957243173 Chr.7: 98291474-98291474 on Build GRCh38
rs1020396010 Chr.7: 98291614-98291614 on Build GRCh38
rs899713419 Chr.7: 98291795-98291795 on Build GRCh38
rs1009712030 Chr.7: 98291828-98291828 on Build GRCh38
rs974457184 Chr.7: 98291737-98291737 on Build GRCh38
rs553132300 Chr.7: 98291850-98291850 on Build GRCh38
rs917285386 Chr.7: 98291517-98291517 on Build GRCh38
rs917377786 Chr.7: 98291695-98291695 on Build GRCh38
rs932338635 Chr.7: 98291805-98291805 on Build GRCh38
rs1054576457 Chr.7: 98291645-98291645 on Build GRCh38
rs754616183 Chr.7: 98291495-98291495 on Build GRCh38
rs1033489475 Chr.7: 98291470-98291470 on Build GRCh38
rs753394385 Chr.7: 98291848-98291848 on Build GRCh38
rs776617849 Chr.7: 98291555-98291555 on Build GRCh38
rs34534766 Chr.7: 98291559-98291559 on Build GRCh38
rs991599992 Chr.7: 98291502-98291502 on Build GRCh38

View Full Product Details