Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98293962-98294474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
UniGene
Hs.656063
Gene Chromosome Location
Chr.7: 98291650-98401115 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
UniGene
Hs.567438
Gene Chromosome Location
Chr.7: 98281667-98322398 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1245940 c.1449G>A p.P483P Chr.7: 98294085-98294085 on Build GRCh38
4951143 c.1459A>C p.S487R Chr.7: 98294075-98294075 on Build GRCh38
3394968 c.1460+5G>A p.? Chr.7: 98294069-98294069 on Build GRCh38
6328521 c.1423-7C>T p.? Chr.7: 98294118-98294118 on Build GRCh38
6261387 c.1460+6C>A p.? Chr.7: 98294068-98294068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780808006 Chr.7: 98294150-98294150 on Build GRCh38
rs189766591 Chr.7: 98294057-98294057 on Build GRCh38
rs984603715 Chr.7: 98294173-98294173 on Build GRCh38
rs757685013 Chr.7: 98294118-98294118 on Build GRCh38
rs753902717 Chr.7: 98294228-98294228 on Build GRCh38
rs769545835 Chr.7: 98294157-98294157 on Build GRCh38
rs914287237 Chr.7: 98294314-98294314 on Build GRCh38
rs993465095 Chr.7: 98294400-98294400 on Build GRCh38
rs116936580 Chr.7: 98294439-98294439 on Build GRCh38
rs776358441 Chr.7: 98294046-98294046 on Build GRCh38
rs769779241 Chr.7: 98294093-98294093 on Build GRCh38
rs943067220 Chr.7: 98294015-98294015 on Build GRCh38
rs1006494984 Chr.7: 98294433-98294433 on Build GRCh38
rs374663231 Chr.7: 98293997-98293997 on Build GRCh38
rs565379340 Chr.7: 98294340-98294340 on Build GRCh38
rs535654668 Chr.7: 98293992-98293992 on Build GRCh38
rs373011560 Chr.7: 98294116-98294116 on Build GRCh38
rs751704840 Chr.7: 98294053-98294053 on Build GRCh38
rs918381069 Chr.7: 98294268-98294268 on Build GRCh38
rs765088736 Chr.7: 98294051-98294051 on Build GRCh38
rs768361429 Chr.7: 98294139-98294139 on Build GRCh38
rs181707306 Chr.7: 98294212-98294212 on Build GRCh38
rs559672707 Chr.7: 98294318-98294318 on Build GRCh38
rs544614541 Chr.7: 98294297-98294297 on Build GRCh38
rs200251950 Chr.7: 98294125-98294125 on Build GRCh38
rs747280967 Chr.7: 98294367-98294367 on Build GRCh38
rs905471637 Chr.7: 98294332-98294332 on Build GRCh38
rs367683409 Chr.7: 98294052-98294052 on Build GRCh38
rs368175991 Chr.7: 98294021-98294021 on Build GRCh38
rs186843214 Chr.7: 98294322-98294322 on Build GRCh38
rs565462844 Chr.7: 98294260-98294260 on Build GRCh38
rs140846982 Chr.7: 98294076-98294076 on Build GRCh38
rs367914437 Chr.7: 98294034-98294034 on Build GRCh38
rs779573408 Chr.7: 98294077-98294077 on Build GRCh38
rs148701332 Chr.7: 98294155-98294155 on Build GRCh38
rs543975802 Chr.7: 98294216-98294216 on Build GRCh38
rs985127889 Chr.7: 98293999-98293999 on Build GRCh38
rs758886875 Chr.7: 98294129-98294129 on Build GRCh38
rs538027916 Chr.7: 98294036-98294036 on Build GRCh38
rs539047916 Chr.7: 98294086-98294086 on Build GRCh38
rs371780461 Chr.7: 98294056-98294056 on Build GRCh38
rs759992240 Chr.7: 98294126-98294128 on Build GRCh38
rs150825031 Chr.7: 98294447-98294447 on Build GRCh38
rs975741247 Chr.7: 98294243-98294243 on Build GRCh38
rs548702194 Chr.7: 98294344-98294344 on Build GRCh38
rs773018564 Chr.7: 98294158-98294158 on Build GRCh38
rs779560885 Chr.7: 98294068-98294068 on Build GRCh38
rs73147337 Chr.7: 98294000-98294000 on Build GRCh38
rs760822063 Chr.7: 98294108-98294108 on Build GRCh38
rs770700624 Chr.7: 98294125-98294127 on Build GRCh38
rs765307405 Chr.7: 98294255-98294255 on Build GRCh38
rs369999863 Chr.7: 98294067-98294067 on Build GRCh38
rs773251871 Chr.7: 98294088-98294088 on Build GRCh38
rs759700340 Chr.7: 98294099-98294099 on Build GRCh38
rs1043893939 Chr.7: 98294323-98294323 on Build GRCh38
rs966813155 Chr.7: 98294422-98294422 on Build GRCh38
rs768295005 Chr.7: 98294084-98294084 on Build GRCh38
rs971920027 Chr.7: 98293986-98293986 on Build GRCh38
rs776411998 Chr.7: 98294085-98294085 on Build GRCh38
rs879749388 Chr.7: 98294386-98294386 on Build GRCh38
rs374428368 Chr.7: 98294032-98294032 on Build GRCh38
rs896504548 Chr.7: 98294370-98294370 on Build GRCh38
rs949282388 Chr.7: 98294253-98294253 on Build GRCh38
rs1048281451 Chr.7: 98294411-98294411 on Build GRCh38
rs776353868 Chr.7: 98294125-98294128 on Build GRCh38
rs779616120 Chr.7: 98294122-98294122 on Build GRCh38
rs577089075 Chr.7: 98294285-98294285 on Build GRCh38
rs900920188 Chr.7: 98294368-98294368 on Build GRCh38
rs752773992 Chr.7: 98294105-98294105 on Build GRCh38
rs988527774 Chr.7: 98294283-98294283 on Build GRCh38
rs373700529 Chr.7: 98294017-98294017 on Build GRCh38
rs746660259 Chr.7: 98294083-98294083 on Build GRCh38
rs754199617 Chr.7: 98294060-98294060 on Build GRCh38
rs1048316747 Chr.7: 98294284-98294284 on Build GRCh38
rs766575393 Chr.7: 98294224-98294224 on Build GRCh38
rs760919152 Chr.7: 98294148-98294148 on Build GRCh38
rs371507314 Chr.7: 98294011-98294011 on Build GRCh38
rs572571991 Chr.7: 98294109-98294109 on Build GRCh38
rs995205332 Chr.7: 98294023-98294023 on Build GRCh38
rs528720522 Chr.7: 98294012-98294012 on Build GRCh38
rs114306991 Chr.7: 98294035-98294035 on Build GRCh38
rs959395804 Chr.7: 98294450-98294450 on Build GRCh38
rs754993054 Chr.7: 98294245-98294245 on Build GRCh38
rs750867155 Chr.7: 98294123-98294123 on Build GRCh38
rs990878932 Chr.7: 98294206-98294206 on Build GRCh38
rs934298543 Chr.7: 98294280-98294280 on Build GRCh38
rs998128328 Chr.7: 98294407-98294407 on Build GRCh38
rs1056709567 Chr.7: 98294345-98294345 on Build GRCh38
rs760454552 Chr.7: 98294039-98294040 on Build GRCh38
rs149794421 Chr.7: 98294096-98294096 on Build GRCh38
rs548665195 Chr.7: 98294019-98294019 on Build GRCh38
rs10237767 Chr.7: 98294421-98294421 on Build GRCh38
rs761749054 Chr.7: 98294049-98294049 on Build GRCh38
rs765470173 Chr.7: 98294149-98294150 on Build GRCh38
rs951980801 Chr.7: 98294172-98294172 on Build GRCh38
rs746459981 Chr.7: 98294137-98294137 on Build GRCh38
rs547173009 Chr.7: 98294100-98294100 on Build GRCh38
rs745443802 Chr.7: 98294072-98294072 on Build GRCh38

View Full Product Details