Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127591122-127591617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
-
UniGene
Hs.653202
Gene Chromosome Location
Chr.7: 127588352-127591705 on Build GRCh38
Transcripts
NM_001662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1447853 c.539G>A p.R180H Chr.7: 127591295-127591295 on Build GRCh38
6108737 c.474C>T p.T158T Chr.7: 127591230-127591230 on Build GRCh38
1447852 c.517C>T p.L173L Chr.7: 127591273-127591273 on Build GRCh38
744675 c.515G>T p.W172L Chr.7: 127591271-127591271 on Build GRCh38
1085151 c.538C>T p.R180C Chr.7: 127591294-127591294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539228447 Chr.7: 127591428-127591428 on Build GRCh38
rs1001291992 Chr.7: 127591448-127591448 on Build GRCh38
rs760153827 Chr.7: 127591313-127591313 on Build GRCh38
rs780694657 Chr.7: 127591326-127591326 on Build GRCh38
rs911339328 Chr.7: 127591443-127591443 on Build GRCh38
rs777906580 Chr.7: 127591203-127591203 on Build GRCh38
rs752982209 Chr.7: 127591319-127591319 on Build GRCh38
rs779907940 Chr.7: 127591257-127591257 on Build GRCh38
rs578033918 Chr.7: 127591538-127591538 on Build GRCh38
rs779568172 Chr.7: 127591335-127591335 on Build GRCh38
rs969831020 Chr.7: 127591157-127591157 on Build GRCh38
rs1024014570 Chr.7: 127591423-127591428 on Build GRCh38
rs558006789 Chr.7: 127591449-127591449 on Build GRCh38
rs77229708 Chr.7: 127591248-127591248 on Build GRCh38
rs556705397 Chr.7: 127591405-127591405 on Build GRCh38
rs115245503 Chr.7: 127591381-127591381 on Build GRCh38
rs762514943 Chr.7: 127591183-127591183 on Build GRCh38
rs562589316 Chr.7: 127591252-127591252 on Build GRCh38
rs749170977 Chr.7: 127591338-127591338 on Build GRCh38
rs566998061 Chr.7: 127591375-127591375 on Build GRCh38
rs368493966 Chr.7: 127591172-127591172 on Build GRCh38
rs767581189 Chr.7: 127591162-127591162 on Build GRCh38
rs957156299 Chr.7: 127591269-127591269 on Build GRCh38
rs1020523070 Chr.7: 127591520-127591520 on Build GRCh38
rs753178005 Chr.7: 127591173-127591173 on Build GRCh38
rs11539552 Chr.7: 127591327-127591327 on Build GRCh38
rs924973215 Chr.7: 127591379-127591379 on Build GRCh38
rs747082944 Chr.7: 127591242-127591242 on Build GRCh38
rs756882514 Chr.7: 127591227-127591227 on Build GRCh38
rs770878160 Chr.7: 127591287-127591287 on Build GRCh38
rs551339342 Chr.7: 127591320-127591320 on Build GRCh38
rs898510483 Chr.7: 127591464-127591464 on Build GRCh38
rs750443569 Chr.7: 127591164-127591164 on Build GRCh38
rs376927315 Chr.7: 127591318-127591318 on Build GRCh38
rs377612947 Chr.7: 127591591-127591592 on Build GRCh38
rs374060875 Chr.7: 127591312-127591312 on Build GRCh38
rs775435205 Chr.7: 127591318-127591319 on Build GRCh38
rs755339407 Chr.7: 127591200-127591200 on Build GRCh38
rs749226432 Chr.7: 127591260-127591260 on Build GRCh38
rs994803583 Chr.7: 127591473-127591473 on Build GRCh38
rs1013798069 Chr.7: 127591548-127591548 on Build GRCh38
rs113896519 Chr.7: 127591507-127591507 on Build GRCh38
rs531878415 Chr.7: 127591207-127591207 on Build GRCh38
rs771721984 Chr.7: 127591484-127591484 on Build GRCh38
rs933178464 Chr.7: 127591427-127591427 on Build GRCh38
rs976401278 Chr.7: 127591581-127591582 on Build GRCh38
rs956797054 Chr.7: 127591380-127591380 on Build GRCh38
rs1036919496 Chr.7: 127591366-127591366 on Build GRCh38
rs543667839 Chr.7: 127591582-127591582 on Build GRCh38
rs192153447 Chr.7: 127591374-127591374 on Build GRCh38
rs371181234 Chr.7: 127591303-127591303 on Build GRCh38
rs774035808 Chr.7: 127591282-127591282 on Build GRCh38
rs923082419 Chr.7: 127591408-127591408 on Build GRCh38
rs780948404 Chr.7: 127591229-127591229 on Build GRCh38
rs762893981 Chr.7: 127591342-127591342 on Build GRCh38
rs772150653 Chr.7: 127591293-127591293 on Build GRCh38
rs758633454 Chr.7: 127591190-127591190 on Build GRCh38
rs773554378 Chr.7: 127591294-127591294 on Build GRCh38
rs896645333 Chr.7: 127591514-127591514 on Build GRCh38
rs944602969 Chr.7: 127591288-127591288 on Build GRCh38
rs142827438 Chr.7: 127591281-127591281 on Build GRCh38
rs751185960 Chr.7: 127591211-127591211 on Build GRCh38
rs934046392 Chr.7: 127591142-127591142 on Build GRCh38
rs571706545 Chr.7: 127591416-127591416 on Build GRCh38
rs2301566 Chr.7: 127591334-127591334 on Build GRCh38
rs572794948 Chr.7: 127591495-127591495 on Build GRCh38
rs760826467 Chr.7: 127591295-127591295 on Build GRCh38
rs1038743214 Chr.7: 127591458-127591458 on Build GRCh38
rs988085733 Chr.7: 127591390-127591390 on Build GRCh38
rs3190668 Chr.7: 127591470-127591470 on Build GRCh38
rs768453188 Chr.7: 127591348-127591348 on Build GRCh38
rs554144811 Chr.7: 127591476-127591476 on Build GRCh38
rs966022521 Chr.7: 127591578-127591578 on Build GRCh38
rs988682868 Chr.7: 127591499-127591499 on Build GRCh38
rs368139645 Chr.7: 127591349-127591349 on Build GRCh38

View Full Product Details