Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128754437-128754933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALU
Entrez Gene ID
813
Gene Name
calumenin
Gene Aliases
-
UniGene
Hs.743262
Gene Chromosome Location
Chr.7: 128739292-128773423 on Build GRCh38
Transcripts
NM_001130674.2 NM_001199671.1 NM_001199672.1 NM_001199673.1 NM_001219.4 NR_074086.1 XM_017012659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
203544 c.408C>T p.Y136Y Chr.7: 128754448-128754448 on Build GRCh38
2769023 c.360A>G p.Q120Q Chr.7: 128754643-128754643 on Build GRCh38
3831829 c.397A>C p.T133P Chr.7: 128754437-128754437 on Build GRCh38
6211864 c.290C>T p.T97M Chr.7: 128754573-128754573 on Build GRCh38
2769018 c.402C>T p.Y134Y Chr.7: 128754442-128754442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546973440 Chr.7: 128754774-128754774 on Build GRCh38
rs1001895624 Chr.7: 128754866-128754866 on Build GRCh38
rs1037845924 Chr.7: 128754867-128754867 on Build GRCh38
rs746265584 Chr.7: 128754572-128754572 on Build GRCh38
rs1003977596 Chr.7: 128754565-128754565 on Build GRCh38
rs1034673453 Chr.7: 128754676-128754676 on Build GRCh38
rs751020603 Chr.7: 128754682-128754682 on Build GRCh38
rs768063197 Chr.7: 128754796-128754796 on Build GRCh38
rs546403121 Chr.7: 128754705-128754705 on Build GRCh38
rs377059578 Chr.7: 128754573-128754573 on Build GRCh38
rs755183703 Chr.7: 128754520-128754520 on Build GRCh38
rs142351036 Chr.7: 128754509-128754509 on Build GRCh38
rs886953247 Chr.7: 128754821-128754821 on Build GRCh38
rs986431842 Chr.7: 128754817-128754817 on Build GRCh38
rs993921435 Chr.7: 128754580-128754580 on Build GRCh38
rs558268751 Chr.7: 128754536-128754536 on Build GRCh38
rs368808641 Chr.7: 128754550-128754550 on Build GRCh38
rs371458036 Chr.7: 128754605-128754605 on Build GRCh38
rs376511847 Chr.7: 128754815-128754815 on Build GRCh38
rs919493924 Chr.7: 128754661-128754661 on Build GRCh38
rs750416117 Chr.7: 128754500-128754500 on Build GRCh38
rs767336502 Chr.7: 128754489-128754489 on Build GRCh38
rs751599561 Chr.7: 128754755-128754755 on Build GRCh38
rs922920739 Chr.7: 128754779-128754779 on Build GRCh38
rs374762041 Chr.7: 128754477-128754477 on Build GRCh38
rs2307040 Chr.7: 128754552-128754552 on Build GRCh38
rs774040896 Chr.7: 128754574-128754574 on Build GRCh38
rs1039665850 Chr.7: 128754789-128754789 on Build GRCh38
rs931247339 Chr.7: 128754801-128754801 on Build GRCh38
rs933551601 Chr.7: 128754541-128754541 on Build GRCh38
rs761336524 Chr.7: 128754484-128754484 on Build GRCh38
rs772306972 Chr.7: 128754577-128754577 on Build GRCh38
rs753004112 Chr.7: 128754714-128754714 on Build GRCh38
rs140316977 Chr.7: 128754701-128754701 on Build GRCh38
rs750929906 Chr.7: 128754839-128754839 on Build GRCh38
rs771359612 Chr.7: 128754557-128754557 on Build GRCh38
rs776917035 Chr.7: 128754560-128754560 on Build GRCh38
rs762538757 Chr.7: 128754472-128754472 on Build GRCh38
rs990379085 Chr.7: 128754647-128754647 on Build GRCh38
rs1024762776 Chr.7: 128754864-128754864 on Build GRCh38
rs376487937 Chr.7: 128754466-128754466 on Build GRCh38
rs564643793 Chr.7: 128754734-128754734 on Build GRCh38
rs563892606 Chr.7: 128754670-128754670 on Build GRCh38
rs760602920 Chr.7: 128754678-128754678 on Build GRCh38
rs753697698 Chr.7: 128754512-128754512 on Build GRCh38
rs117820033 Chr.7: 128754630-128754630 on Build GRCh38
rs773009689 Chr.7: 128754659-128754659 on Build GRCh38
rs1033365858 Chr.7: 128754871-128754871 on Build GRCh38
rs768757548 Chr.7: 128754518-128754522 on Build GRCh38
rs151188778 Chr.7: 128754553-128754553 on Build GRCh38
rs759625847 Chr.7: 128754707-128754708 on Build GRCh38
rs910793055 Chr.7: 128754861-128754861 on Build GRCh38
rs765532004 Chr.7: 128754709-128754709 on Build GRCh38
rs339044 Chr.7: 128754865-128754865 on Build GRCh38
rs41274227 Chr.7: 128754754-128754754 on Build GRCh38
rs763607153 Chr.7: 128754664-128754664 on Build GRCh38
rs755857759 Chr.7: 128754508-128754508 on Build GRCh38
rs763744180 Chr.7: 128754473-128754473 on Build GRCh38
rs919351123 Chr.7: 128754906-128754906 on Build GRCh38
rs748187816 Chr.7: 128754540-128754540 on Build GRCh38
rs1046361375 Chr.7: 128754619-128754619 on Build GRCh38
rs1034632098 Chr.7: 128754596-128754596 on Build GRCh38

View Full Product Details