Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130346252-130346749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA5
Entrez Gene ID
93979
Gene Name
carboxypeptidase A5
Gene Aliases
-
UniGene
Hs.567642
Gene Chromosome Location
Chr.7: 130344790-130368730 on Build GRCh38
Transcripts
NM_001127441.1 NM_001127442.1 NM_001318223.1 NM_080385.4 XM_005250710.1 XM_005250712.1 XM_011516698.1 XM_011516701.1 XM_011516702.1 XM_011516703.1 XM_011516704.1 XM_011516705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6176148 c.41C>G p.S14C Chr.7: 130346526-130346526 on Build GRCh38
3878083 c.27G>A p.T9T Chr.7: 130346512-130346512 on Build GRCh38
4459302 c.111C>T p.F37F Chr.7: 130346596-130346596 on Build GRCh38
4674285 c.22G>A p.G8R Chr.7: 130346507-130346507 on Build GRCh38
3878084 c.98G>A p.G33D Chr.7: 130346583-130346583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782418427 Chr.7: 130346478-130346478 on Build GRCh38
rs558364717 Chr.7: 130346449-130346449 on Build GRCh38
rs782300825 Chr.7: 130346471-130346471 on Build GRCh38
rs550865855 Chr.7: 130346398-130346398 on Build GRCh38
rs1002520552 Chr.7: 130346316-130346316 on Build GRCh38
rs1002551419 Chr.7: 130346526-130346526 on Build GRCh38
rs111827298 Chr.7: 130346441-130346441 on Build GRCh38
rs371314635 Chr.7: 130346628-130346628 on Build GRCh38
rs782206306 Chr.7: 130346509-130346509 on Build GRCh38
rs782277762 Chr.7: 130346653-130346653 on Build GRCh38
rs200717171 Chr.7: 130346507-130346507 on Build GRCh38
rs782246205 Chr.7: 130346514-130346514 on Build GRCh38
rs147176992 Chr.7: 130346592-130346592 on Build GRCh38
rs1028241581 Chr.7: 130346677-130346677 on Build GRCh38
rs180976386 Chr.7: 130346372-130346372 on Build GRCh38
rs143506536 Chr.7: 130346522-130346522 on Build GRCh38
rs976740550 Chr.7: 130346649-130346649 on Build GRCh38
rs781973473 Chr.7: 130346530-130346530 on Build GRCh38
rs782467316 Chr.7: 130346494-130346494 on Build GRCh38
rs781856522 Chr.7: 130346500-130346500 on Build GRCh38
rs781784996 Chr.7: 130346574-130346574 on Build GRCh38
rs782287845 Chr.7: 130346587-130346587 on Build GRCh38
rs782573202 Chr.7: 130346288-130346288 on Build GRCh38
rs909113317 Chr.7: 130346685-130346685 on Build GRCh38
rs545232007 Chr.7: 130346313-130346313 on Build GRCh38
rs938370894 Chr.7: 130346726-130346726 on Build GRCh38
rs144865240 Chr.7: 130346506-130346506 on Build GRCh38
rs965405452 Chr.7: 130346630-130346630 on Build GRCh38
rs11761867 Chr.7: 130346315-130346315 on Build GRCh38
rs186172044 Chr.7: 130346407-130346407 on Build GRCh38
rs782255534 Chr.7: 130346442-130346442 on Build GRCh38
rs986491612 Chr.7: 130346681-130346681 on Build GRCh38
rs6973851 Chr.7: 130346497-130346497 on Build GRCh38
rs919017093 Chr.7: 130346723-130346723 on Build GRCh38
rs782478769 Chr.7: 130346552-130346552 on Build GRCh38
rs373843590 Chr.7: 130346536-130346536 on Build GRCh38
rs782812500 Chr.7: 130346482-130346482 on Build GRCh38
rs6955991 Chr.7: 130346385-130346385 on Build GRCh38
rs376798873 Chr.7: 130346633-130346633 on Build GRCh38
rs781853455 Chr.7: 130346485-130346485 on Build GRCh38
rs367730990 Chr.7: 130346555-130346555 on Build GRCh38
rs782081863 Chr.7: 130346487-130346487 on Build GRCh38
rs191040511 Chr.7: 130346540-130346540 on Build GRCh38
rs781944248 Chr.7: 130346459-130346459 on Build GRCh38
rs958914098 Chr.7: 130346337-130346337 on Build GRCh38
rs782164665 Chr.7: 130346541-130346541 on Build GRCh38
rs782504506 Chr.7: 130346588-130346588 on Build GRCh38
rs6467306 Chr.7: 130346680-130346680 on Build GRCh38
rs566930313 Chr.7: 130346472-130346472 on Build GRCh38
rs782410158 Chr.7: 130346437-130346437 on Build GRCh38
rs971891259 Chr.7: 130346700-130346700 on Build GRCh38
rs781880842 Chr.7: 130346550-130346550 on Build GRCh38
rs533162578 Chr.7: 130346305-130346305 on Build GRCh38
rs35675773 Chr.7: 130346514-130346514 on Build GRCh38
rs782093643 Chr.7: 130346460-130346460 on Build GRCh38
rs35724550 Chr.7: 130346439-130346439 on Build GRCh38
rs539010159 Chr.7: 130346320-130346320 on Build GRCh38
rs782654344 Chr.7: 130346586-130346586 on Build GRCh38
rs782761130 Chr.7: 130346493-130346493 on Build GRCh38
rs201679339 Chr.7: 130346531-130346531 on Build GRCh38
rs781933897 Chr.7: 130346475-130346475 on Build GRCh38
rs782233615 Chr.7: 130346620-130346620 on Build GRCh38
rs117427621 Chr.7: 130346699-130346699 on Build GRCh38
rs781834339 Chr.7: 130346309-130346309 on Build GRCh38
rs782769742 Chr.7: 130346549-130346549 on Build GRCh38
rs781940951 Chr.7: 130346635-130346635 on Build GRCh38
rs782462133 Chr.7: 130346580-130346580 on Build GRCh38
rs782186925 Chr.7: 130346512-130346512 on Build GRCh38
rs6955995 Chr.7: 130346395-130346395 on Build GRCh38
rs1022169696 Chr.7: 130346393-130346393 on Build GRCh38

View Full Product Details