Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130394073-130394583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP41
Entrez Gene ID
95681
Gene Name
centrosomal protein 41
Gene Aliases
JBTS15, TSGA14
UniGene
Hs.368315
Gene Chromosome Location
Chr.7: 130393771-130441210 on Build GRCh38
Transcripts
NM_001257158.1 NM_001257159.1 NM_018718.2 NR_046443.1

SNP Detail

SNP ID Chromosome Location
rs904672851 Chr.7: 130394534-130394534 on Build GRCh38
rs782045222 Chr.7: 130394309-130394309 on Build GRCh38
rs781972597 Chr.7: 130394166-130394166 on Build GRCh38
rs535299613 Chr.7: 130394526-130394526 on Build GRCh38
rs374866275 Chr.7: 130394183-130394183 on Build GRCh38
rs565965512 Chr.7: 130394430-130394430 on Build GRCh38
rs948930004 Chr.7: 130394493-130394493 on Build GRCh38
rs986339569 Chr.7: 130394208-130394208 on Build GRCh38
rs782200325 Chr.7: 130394131-130394131 on Build GRCh38
rs916134541 Chr.7: 130394488-130394488 on Build GRCh38
rs781794703 Chr.7: 130394491-130394491 on Build GRCh38
rs140706708 Chr.7: 130394315-130394315 on Build GRCh38
rs145921907 Chr.7: 130394423-130394423 on Build GRCh38
rs117814753 Chr.7: 130394296-130394296 on Build GRCh38
rs886061987 Chr.7: 130394427-130394427 on Build GRCh38
rs781826261 Chr.7: 130394187-130394187 on Build GRCh38
rs1032113771 Chr.7: 130394121-130394121 on Build GRCh38
rs753024852 Chr.7: 130394292-130394292 on Build GRCh38
rs781813144 Chr.7: 130394379-130394379 on Build GRCh38
rs151309255 Chr.7: 130394297-130394297 on Build GRCh38
rs782085608 Chr.7: 130394177-130394177 on Build GRCh38
rs782310305 Chr.7: 130394153-130394153 on Build GRCh38
rs886061988 Chr.7: 130394474-130394474 on Build GRCh38
rs532146907 Chr.7: 130394327-130394327 on Build GRCh38
rs782450438 Chr.7: 130394500-130394500 on Build GRCh38
rs531784517 Chr.7: 130394140-130394140 on Build GRCh38
rs566536736 Chr.7: 130394419-130394419 on Build GRCh38
rs782536937 Chr.7: 130394316-130394316 on Build GRCh38
rs976006398 Chr.7: 130394159-130394159 on Build GRCh38
rs376157224 Chr.7: 130394453-130394453 on Build GRCh38
rs556578226 Chr.7: 130394506-130394506 on Build GRCh38
rs782512901 Chr.7: 130394452-130394452 on Build GRCh38
rs782105530 Chr.7: 130394248-130394248 on Build GRCh38
rs886061986 Chr.7: 130394232-130394232 on Build GRCh38
rs965337048 Chr.7: 130394122-130394122 on Build GRCh38
rs571059603 Chr.7: 130394343-130394343 on Build GRCh38
rs536112774 Chr.7: 130394440-130394440 on Build GRCh38
rs781982705 Chr.7: 130394276-130394276 on Build GRCh38
rs758729376 Chr.7: 130394547-130394547 on Build GRCh38
rs782687708 Chr.7: 130394411-130394411 on Build GRCh38

View Full Product Details