Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130395362-130395865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP41
Entrez Gene ID
95681
Gene Name
centrosomal protein 41
Gene Aliases
JBTS15, TSGA14
UniGene
Hs.368315
Gene Chromosome Location
Chr.7: 130393771-130441210 on Build GRCh38
Transcripts
NM_001257158.1 NM_001257159.1 NM_018718.2 NR_046443.1 XM_011516709.2 XM_011516710.2

SNP Detail

SNP ID Chromosome Location
rs782621177 Chr.7: 130395679-130395679 on Build GRCh38
rs1047672773 Chr.7: 130395797-130395797 on Build GRCh38
rs183669606 Chr.7: 130395447-130395447 on Build GRCh38
rs782715548 Chr.7: 130395523-130395523 on Build GRCh38
rs781787941 Chr.7: 130395549-130395549 on Build GRCh38
rs782597072 Chr.7: 130395745-130395745 on Build GRCh38
rs782254477 Chr.7: 130395814-130395814 on Build GRCh38
rs897131455 Chr.7: 130395792-130395792 on Build GRCh38
rs1023143971 Chr.7: 130395831-130395831 on Build GRCh38
rs886061990 Chr.7: 130395822-130395822 on Build GRCh38
rs144534675 Chr.7: 130395440-130395440 on Build GRCh38
rs765675123 Chr.7: 130395825-130395825 on Build GRCh38
rs956340061 Chr.7: 130395536-130395536 on Build GRCh38
rs547608590 Chr.7: 130395697-130395697 on Build GRCh38
rs112007936 Chr.7: 130395617-130395617 on Build GRCh38
rs552490309 Chr.7: 130395505-130395505 on Build GRCh38
rs1038351020 Chr.7: 130395727-130395727 on Build GRCh38
rs530785805 Chr.7: 130395494-130395494 on Build GRCh38
rs782100341 Chr.7: 130395707-130395707 on Build GRCh38
rs534794569 Chr.7: 130395515-130395515 on Build GRCh38
rs922806466 Chr.7: 130395566-130395566 on Build GRCh38
rs781902268 Chr.7: 130395500-130395500 on Build GRCh38
rs929238589 Chr.7: 130395794-130395794 on Build GRCh38
rs941369237 Chr.7: 130395695-130395695 on Build GRCh38
rs989008301 Chr.7: 130395542-130395542 on Build GRCh38
rs934156244 Chr.7: 130395611-130395611 on Build GRCh38
rs139710833 Chr.7: 130395445-130395445 on Build GRCh38
rs886061989 Chr.7: 130395698-130395698 on Build GRCh38
rs570602311 Chr.7: 130395506-130395506 on Build GRCh38
rs1014058037 Chr.7: 130395820-130395820 on Build GRCh38

View Full Product Details