Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130396159-130396667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP41
Entrez Gene ID
95681
Gene Name
centrosomal protein 41
Gene Aliases
JBTS15, TSGA14
UniGene
Hs.368315
Gene Chromosome Location
Chr.7: 130393771-130441210 on Build GRCh38
Transcripts
NM_001257158.1 NM_001257159.1 NM_018718.2 NR_046443.1 XM_011516709.2 XM_011516710.2

SNP Detail

SNP ID Chromosome Location
rs955596511 Chr.7: 130396291-130396291 on Build GRCh38
rs905679390 Chr.7: 130396222-130396222 on Build GRCh38
rs891463361 Chr.7: 130396247-130396247 on Build GRCh38
rs782252260 Chr.7: 130396498-130396498 on Build GRCh38
rs73152867 Chr.7: 130396502-130396502 on Build GRCh38
rs781941870 Chr.7: 130396589-130396589 on Build GRCh38
rs782289999 Chr.7: 130396294-130396294 on Build GRCh38
rs782667875 Chr.7: 130396520-130396520 on Build GRCh38
rs1044260434 Chr.7: 130396197-130396197 on Build GRCh38
rs781845370 Chr.7: 130396402-130396402 on Build GRCh38
rs917190260 Chr.7: 130396442-130396442 on Build GRCh38
rs374647228 Chr.7: 130396256-130396256 on Build GRCh38
rs34866043 Chr.7: 130396259-130396260 on Build GRCh38
rs189091601 Chr.7: 130396409-130396409 on Build GRCh38
rs886061992 Chr.7: 130396286-130396286 on Build GRCh38
rs763362166 Chr.7: 130396449-130396449 on Build GRCh38
rs782478229 Chr.7: 130396450-130396450 on Build GRCh38
rs781982629 Chr.7: 130396487-130396487 on Build GRCh38
rs775997340 Chr.7: 130396269-130396269 on Build GRCh38
rs963197810 Chr.7: 130396377-130396377 on Build GRCh38
rs546744390 Chr.7: 130396447-130396447 on Build GRCh38
rs782755677 Chr.7: 130396229-130396229 on Build GRCh38
rs182084979 Chr.7: 130396477-130396477 on Build GRCh38
rs782743235 Chr.7: 130396475-130396475 on Build GRCh38
rs886061993 Chr.7: 130396505-130396505 on Build GRCh38
rs548265556 Chr.7: 130396527-130396527 on Build GRCh38
rs782081483 Chr.7: 130396470-130396470 on Build GRCh38
rs781820219 Chr.7: 130396191-130396191 on Build GRCh38
rs1029880426 Chr.7: 130396286-130396286 on Build GRCh38
rs1054609748 Chr.7: 130396236-130396236 on Build GRCh38
rs782653622 Chr.7: 130396451-130396451 on Build GRCh38
rs1007177790 Chr.7: 130396607-130396607 on Build GRCh38
rs1050743948 Chr.7: 130396563-130396563 on Build GRCh38
rs890781189 Chr.7: 130396580-130396580 on Build GRCh38
rs1018476557 Chr.7: 130396371-130396371 on Build GRCh38
rs567737975 Chr.7: 130396225-130396225 on Build GRCh38
rs918636193 Chr.7: 130396394-130396394 on Build GRCh38
rs528289176 Chr.7: 130396410-130396410 on Build GRCh38
rs782516165 Chr.7: 130396204-130396204 on Build GRCh38
rs1009822009 Chr.7: 130396273-130396273 on Build GRCh38
rs569758927 Chr.7: 130396552-130396552 on Build GRCh38
rs782310225 Chr.7: 130396391-130396391 on Build GRCh38
rs927187455 Chr.7: 130396481-130396481 on Build GRCh38
rs782471496 Chr.7: 130396270-130396270 on Build GRCh38

View Full Product Details