Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130652773-130653243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG2
Entrez Gene ID
26958
Gene Name
coatomer protein complex subunit gamma 2
Gene Aliases
2-COP, gamma-2-COP
UniGene
Hs.6421
Gene Chromosome Location
Chr.7: 130506238-130668758 on Build GRCh38
Transcripts
NM_001290033.1 NM_012133.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410978 c.277A>G p.K93E Chr.7: 130652915-130652915 on Build GRCh38
6048321 c.262_263delTG p.C88fs*12 Chr.7: 130652929-130652930 on Build GRCh38
2770150 c.280G>T p.E94* Chr.7: 130652912-130652912 on Build GRCh38
3995353 c.287C>G p.A96G Chr.7: 130652905-130652905 on Build GRCh38
307689 c.269_273delTTACC p.L90fs*9 Chr.7: 130652919-130652923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782649668 Chr.7: 130652885-130652885 on Build GRCh38
rs782267873 Chr.7: 130652952-130652952 on Build GRCh38
rs374495041 Chr.7: 130652816-130652816 on Build GRCh38
rs782153267 Chr.7: 130652832-130652832 on Build GRCh38
rs158681 Chr.7: 130653075-130653075 on Build GRCh38
rs781883835 Chr.7: 130652886-130652886 on Build GRCh38
rs562911799 Chr.7: 130652853-130652853 on Build GRCh38
rs781791308 Chr.7: 130652841-130652841 on Build GRCh38
rs545732589 Chr.7: 130653032-130653032 on Build GRCh38
rs577913401 Chr.7: 130653022-130653022 on Build GRCh38
rs918627194 Chr.7: 130653072-130653072 on Build GRCh38
rs375839317 Chr.7: 130653075-130653075 on Build GRCh38
rs782807627 Chr.7: 130653034-130653034 on Build GRCh38
rs368960937 Chr.7: 130653062-130653062 on Build GRCh38
rs372799475 Chr.7: 130652914-130652914 on Build GRCh38
rs782411523 Chr.7: 130652955-130652955 on Build GRCh38
rs374487044 Chr.7: 130652918-130652918 on Build GRCh38
rs997309839 Chr.7: 130652804-130652804 on Build GRCh38
rs782039285 Chr.7: 130652981-130652981 on Build GRCh38
rs158680 Chr.7: 130653025-130653025 on Build GRCh38
rs781836981 Chr.7: 130652866-130652866 on Build GRCh38
rs782501578 Chr.7: 130652951-130652952 on Build GRCh38
rs1056576163 Chr.7: 130653086-130653086 on Build GRCh38
rs782663066 Chr.7: 130652959-130652959 on Build GRCh38
rs930038122 Chr.7: 130653073-130653073 on Build GRCh38
rs373002185 Chr.7: 130653144-130653144 on Build GRCh38
rs544345465 Chr.7: 130652818-130652818 on Build GRCh38
rs745483535 Chr.7: 130653149-130653149 on Build GRCh38
rs782062782 Chr.7: 130652857-130652857 on Build GRCh38
rs781797504 Chr.7: 130652875-130652875 on Build GRCh38
rs782502377 Chr.7: 130652877-130652877 on Build GRCh38
rs782745546 Chr.7: 130652840-130652840 on Build GRCh38
rs769368420 Chr.7: 130653152-130653152 on Build GRCh38
rs782553957 Chr.7: 130652900-130652900 on Build GRCh38
rs782187967 Chr.7: 130652902-130652902 on Build GRCh38
rs782326544 Chr.7: 130652986-130652986 on Build GRCh38
rs782048097 Chr.7: 130652985-130652985 on Build GRCh38
rs782731996 Chr.7: 130652860-130652860 on Build GRCh38
rs367576648 Chr.7: 130652887-130652887 on Build GRCh38
rs974474604 Chr.7: 130653035-130653035 on Build GRCh38
rs541278466 Chr.7: 130653065-130653065 on Build GRCh38
rs781886655 Chr.7: 130653145-130653145 on Build GRCh38

View Full Product Details