Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 133374548-133375048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC4
Entrez Gene ID
60412
Gene Name
exocyst complex component 4
Gene Aliases
SEC8, SEC8L1, Sec8p
UniGene
Hs.321273
Gene Chromosome Location
Chr.7: 133253067-134067137 on Build GRCh38
Transcripts
NM_001037126.1 NM_021807.3 XM_005250523.4 XM_017012495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442833 c.1126G>A p.D376N Chr.7: 133374946-133374946 on Build GRCh38
3633528 c.1131C>T p.I377I Chr.7: 133374951-133374951 on Build GRCh38
1085716 c.1164G>A p.K388K Chr.7: 133374984-133374984 on Build GRCh38
744474 c.1091A>T p.Q364L Chr.7: 133374911-133374911 on Build GRCh38
3633529 c.1131C>T p.I377I Chr.7: 133374951-133374951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748766690 Chr.7: 133374810-133374810 on Build GRCh38
rs34646219 Chr.7: 133374644-133374644 on Build GRCh38
rs1029366185 Chr.7: 133374879-133374879 on Build GRCh38
rs989297313 Chr.7: 133374631-133374631 on Build GRCh38
rs766525916 Chr.7: 133374750-133374750 on Build GRCh38
rs750166788 Chr.7: 133374916-133374916 on Build GRCh38
rs777013493 Chr.7: 133374887-133374887 on Build GRCh38
rs910939345 Chr.7: 133374712-133374712 on Build GRCh38
rs747354706 Chr.7: 133374971-133374971 on Build GRCh38
rs146469408 Chr.7: 133374880-133374880 on Build GRCh38
rs959461467 Chr.7: 133374625-133374625 on Build GRCh38
rs527959228 Chr.7: 133374982-133374982 on Build GRCh38
rs756029623 Chr.7: 133374791-133374791 on Build GRCh38
rs920754917 Chr.7: 133374949-133374949 on Build GRCh38
rs777906349 Chr.7: 133374787-133374790 on Build GRCh38
rs920787816 Chr.7: 133374658-133374658 on Build GRCh38
rs754549529 Chr.7: 133374812-133374812 on Build GRCh38
rs778560013 Chr.7: 133374943-133374943 on Build GRCh38
rs972748042 Chr.7: 133374993-133374993 on Build GRCh38
rs1022638198 Chr.7: 133374645-133374645 on Build GRCh38
rs930698403 Chr.7: 133374668-133374668 on Build GRCh38
rs746371837 Chr.7: 133375003-133375003 on Build GRCh38
rs775492018 Chr.7: 133375017-133375017 on Build GRCh38
rs933765730 Chr.7: 133374686-133374686 on Build GRCh38
rs574607690 Chr.7: 133374591-133374591 on Build GRCh38
rs1010602343 Chr.7: 133374929-133374929 on Build GRCh38
rs374531055 Chr.7: 133374822-133374822 on Build GRCh38
rs763026595 Chr.7: 133375022-133375022 on Build GRCh38
rs139805919 Chr.7: 133374939-133374939 on Build GRCh38
rs950041317 Chr.7: 133374732-133374732 on Build GRCh38
rs201442061 Chr.7: 133375007-133375007 on Build GRCh38
rs368348992 Chr.7: 133374785-133374785 on Build GRCh38
rs117417400 Chr.7: 133374609-133374609 on Build GRCh38
rs765294870 Chr.7: 133374890-133374890 on Build GRCh38
rs372570816 Chr.7: 133374790-133374790 on Build GRCh38
rs1037849676 Chr.7: 133374724-133374724 on Build GRCh38
rs766289665 Chr.7: 133374931-133374931 on Build GRCh38
rs747715190 Chr.7: 133374950-133374950 on Build GRCh38
rs1052262669 Chr.7: 133374866-133374866 on Build GRCh38
rs760682146 Chr.7: 133374683-133374683 on Build GRCh38
rs943535097 Chr.7: 133374715-133374715 on Build GRCh38
rs369040208 Chr.7: 133374985-133374985 on Build GRCh38
rs554684390 Chr.7: 133374587-133374587 on Build GRCh38
rs146672350 Chr.7: 133374963-133374963 on Build GRCh38
rs985198855 Chr.7: 133374705-133374705 on Build GRCh38
rs779887266 Chr.7: 133374795-133374795 on Build GRCh38
rs906657570 Chr.7: 133374580-133374580 on Build GRCh38
rs571542402 Chr.7: 133374784-133374784 on Build GRCh38
rs906468539 Chr.7: 133374801-133374801 on Build GRCh38
rs552053113 Chr.7: 133374607-133374607 on Build GRCh38
rs1003619619 Chr.7: 133374775-133374775 on Build GRCh38
rs149904766 Chr.7: 133374874-133374874 on Build GRCh38
rs563306574 Chr.7: 133374853-133374853 on Build GRCh38
rs775584338 Chr.7: 133374897-133374897 on Build GRCh38
rs906541597 Chr.7: 133374762-133374762 on Build GRCh38
rs977780224 Chr.7: 133374676-133374676 on Build GRCh38
rs760091384 Chr.7: 133374888-133374888 on Build GRCh38
rs1047959899 Chr.7: 133374959-133374959 on Build GRCh38
rs966631141 Chr.7: 133374666-133374666 on Build GRCh38
rs755892680 Chr.7: 133374918-133374918 on Build GRCh38
rs770345352 Chr.7: 133375016-133375016 on Build GRCh38
rs546077707 Chr.7: 133374898-133374898 on Build GRCh38
rs577020229 Chr.7: 133374886-133374886 on Build GRCh38
rs758062881 Chr.7: 133374957-133374957 on Build GRCh38
rs764321303 Chr.7: 133374913-133374913 on Build GRCh38
rs915908876 Chr.7: 133374707-133374707 on Build GRCh38
rs143199058 Chr.7: 133374940-133374940 on Build GRCh38
rs763137832 Chr.7: 133374903-133374903 on Build GRCh38
rs1027824010 Chr.7: 133374999-133374999 on Build GRCh38
rs35502244 Chr.7: 133374667-133374667 on Build GRCh38

View Full Product Details