Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134661708-134662207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPGM
Entrez Gene ID
669
Gene Name
bisphosphoglycerate mutase
Gene Aliases
DPGM
UniGene
Hs.198365
Gene Chromosome Location
Chr.7: 134646779-134679816 on Build GRCh38
Transcripts
NM_001293085.1 NM_001724.4 NM_199186.2 XM_011516527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2858753 c.219G>T p.E73D Chr.7: 134661726-134661726 on Build GRCh38
4911588 c.321T>C p.H107H Chr.7: 134661828-134661828 on Build GRCh38
4665952 c.525C>T p.P175P Chr.7: 134662032-134662032 on Build GRCh38
1755094 c.586C>T p.L196L Chr.7: 134662093-134662093 on Build GRCh38
150394 c.456G>C p.L152L Chr.7: 134661963-134661963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026042730 Chr.7: 134661886-134661886 on Build GRCh38
rs755284418 Chr.7: 134661748-134661748 on Build GRCh38
rs557253408 Chr.7: 134662112-134662112 on Build GRCh38
rs769984158 Chr.7: 134662053-134662053 on Build GRCh38
rs925069034 Chr.7: 134662109-134662109 on Build GRCh38
rs144528605 Chr.7: 134661876-134661876 on Build GRCh38
rs200416137 Chr.7: 134662033-134662033 on Build GRCh38
rs775128216 Chr.7: 134661795-134661795 on Build GRCh38
rs772421290 Chr.7: 134661943-134661943 on Build GRCh38
rs901476075 Chr.7: 134661857-134661857 on Build GRCh38
rs759610083 Chr.7: 134661971-134661971 on Build GRCh38
rs770878186 Chr.7: 134661765-134661765 on Build GRCh38
rs371151871 Chr.7: 134662019-134662019 on Build GRCh38
rs777431866 Chr.7: 134662141-134662141 on Build GRCh38
rs139392890 Chr.7: 134662101-134662101 on Build GRCh38
rs141221644 Chr.7: 134661925-134661925 on Build GRCh38
rs374333584 Chr.7: 134661955-134661955 on Build GRCh38
rs752242999 Chr.7: 134662136-134662136 on Build GRCh38
rs754851528 Chr.7: 134662141-134662142 on Build GRCh38
rs775765508 Chr.7: 134662063-134662063 on Build GRCh38
rs745809683 Chr.7: 134661788-134661788 on Build GRCh38
rs150724691 Chr.7: 134662042-134662042 on Build GRCh38
rs536526376 Chr.7: 134662032-134662032 on Build GRCh38
rs773624452 Chr.7: 134661945-134661945 on Build GRCh38
rs369978968 Chr.7: 134662148-134662148 on Build GRCh38
rs1048704773 Chr.7: 134662116-134662116 on Build GRCh38
rs752985789 Chr.7: 134661908-134661908 on Build GRCh38
rs887470094 Chr.7: 134662132-134662132 on Build GRCh38
rs757933635 Chr.7: 134662140-134662140 on Build GRCh38
rs149329328 Chr.7: 134661851-134661851 on Build GRCh38
rs768179498 Chr.7: 134661941-134661941 on Build GRCh38
rs943191904 Chr.7: 134662123-134662123 on Build GRCh38
rs761063342 Chr.7: 134661946-134661946 on Build GRCh38
rs764374755 Chr.7: 134662091-134662091 on Build GRCh38
rs756090558 Chr.7: 134661733-134661733 on Build GRCh38
rs566467108 Chr.7: 134662088-134662088 on Build GRCh38
rs201783021 Chr.7: 134661875-134661875 on Build GRCh38
rs546208510 Chr.7: 134661750-134661750 on Build GRCh38
rs773642912 Chr.7: 134661845-134661845 on Build GRCh38
rs376051484 Chr.7: 134662134-134662134 on Build GRCh38
rs543243292 Chr.7: 134661921-134661921 on Build GRCh38
rs139806212 Chr.7: 134661995-134661995 on Build GRCh38
rs142455208 Chr.7: 134661991-134661991 on Build GRCh38
rs548312727 Chr.7: 134661801-134661801 on Build GRCh38
rs121964925 Chr.7: 134661775-134661775 on Build GRCh38
rs146970055 Chr.7: 134661942-134661942 on Build GRCh38
rs760462261 Chr.7: 134662095-134662095 on Build GRCh38
rs372950660 Chr.7: 134661763-134661763 on Build GRCh38
rs756128410 Chr.7: 134661926-134661926 on Build GRCh38
rs754966146 Chr.7: 134662133-134662133 on Build GRCh38
rs142076414 Chr.7: 134662020-134662020 on Build GRCh38
rs190681045 Chr.7: 134661894-134661894 on Build GRCh38
rs750541830 Chr.7: 134662108-134662108 on Build GRCh38
rs768397590 Chr.7: 134661797-134661797 on Build GRCh38
rs749493519 Chr.7: 134662011-134662011 on Build GRCh38
rs762136951 Chr.7: 134661764-134661765 on Build GRCh38
rs747355115 Chr.7: 134662038-134662038 on Build GRCh38
rs1038973044 Chr.7: 134661835-134661835 on Build GRCh38
rs746836937 Chr.7: 134661764-134661764 on Build GRCh38
rs1007481067 Chr.7: 134661828-134661828 on Build GRCh38
rs201505421 Chr.7: 134662043-134662043 on Build GRCh38
rs532282608 Chr.7: 134661934-134661934 on Build GRCh38
rs767426687 Chr.7: 134661886-134661888 on Build GRCh38
rs901777714 Chr.7: 134662167-134662167 on Build GRCh38
rs778462618 Chr.7: 134662022-134662022 on Build GRCh38
rs903716285 Chr.7: 134661822-134661822 on Build GRCh38
rs1038941701 Chr.7: 134662161-134662161 on Build GRCh38
rs751753233 Chr.7: 134662093-134662093 on Build GRCh38
rs754001223 Chr.7: 134661740-134661740 on Build GRCh38
rs755896639 Chr.7: 134662000-134662000 on Build GRCh38
rs1043559831 Chr.7: 134661766-134661766 on Build GRCh38
rs553377218 Chr.7: 134661791-134661791 on Build GRCh38
rs370703499 Chr.7: 134661936-134661936 on Build GRCh38
rs374008784 Chr.7: 134661874-134661874 on Build GRCh38
rs781203617 Chr.7: 134661922-134661922 on Build GRCh38
rs368692179 Chr.7: 134661960-134661960 on Build GRCh38
rs765416722 Chr.7: 134661972-134661972 on Build GRCh38
rs573391134 Chr.7: 134661830-134661830 on Build GRCh38
rs781222092 Chr.7: 134661776-134661776 on Build GRCh38
rs780186103 Chr.7: 134661738-134661738 on Build GRCh38
rs774024611 Chr.7: 134661804-134661804 on Build GRCh38
rs762117892 Chr.7: 134661837-134661837 on Build GRCh38
rs755254463 Chr.7: 134662125-134662125 on Build GRCh38
rs765513810 Chr.7: 134661895-134661895 on Build GRCh38
rs376210775 Chr.7: 134661843-134661843 on Build GRCh38
rs145982146 Chr.7: 134661993-134661993 on Build GRCh38
rs370270863 Chr.7: 134661860-134661860 on Build GRCh38
rs747785534 Chr.7: 134662026-134662026 on Build GRCh38
rs1043873892 Chr.7: 134662105-134662105 on Build GRCh38

View Full Product Details