Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135730124-135730629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM180A
Entrez Gene ID
389558
Gene Name
family with sequence similarity 180 member A
Gene Aliases
UNQ1940
UniGene
Hs.55200
Gene Chromosome Location
Chr.7: 135729598-135748869 on Build GRCh38
Transcripts
NM_205855.3 XM_006715983.3

SNP Detail

SNP ID Chromosome Location
rs963677974 Chr.7: 135730298-135730298 on Build GRCh38
rs895080647 Chr.7: 135730439-135730439 on Build GRCh38
rs548045774 Chr.7: 135730437-135730437 on Build GRCh38
rs981520729 Chr.7: 135730275-135730275 on Build GRCh38
rs140768719 Chr.7: 135730181-135730181 on Build GRCh38
rs1047376777 Chr.7: 135730572-135730572 on Build GRCh38
rs766261650 Chr.7: 135730403-135730403 on Build GRCh38
rs550352317 Chr.7: 135730497-135730497 on Build GRCh38
rs1002253000 Chr.7: 135730242-135730242 on Build GRCh38
rs922930915 Chr.7: 135730560-135730560 on Build GRCh38
rs956104997 Chr.7: 135730512-135730512 on Build GRCh38
rs182941381 Chr.7: 135730593-135730593 on Build GRCh38
rs901480953 Chr.7: 135730348-135730348 on Build GRCh38
rs190565920 Chr.7: 135730345-135730345 on Build GRCh38
rs1009714491 Chr.7: 135730269-135730269 on Build GRCh38
rs991939684 Chr.7: 135730201-135730201 on Build GRCh38
rs1056372673 Chr.7: 135730545-135730545 on Build GRCh38
rs567935452 Chr.7: 135730458-135730458 on Build GRCh38
rs530652689 Chr.7: 135730459-135730459 on Build GRCh38
rs914729539 Chr.7: 135730525-135730528 on Build GRCh38
rs1035040744 Chr.7: 135730243-135730243 on Build GRCh38
rs774292451 Chr.7: 135730563-135730563 on Build GRCh38
rs1033096114 Chr.7: 135730286-135730286 on Build GRCh38
rs990339809 Chr.7: 135730523-135730523 on Build GRCh38
rs187997254 Chr.7: 135730191-135730191 on Build GRCh38
rs941277389 Chr.7: 135730426-135730426 on Build GRCh38
rs1047428726 Chr.7: 135730400-135730400 on Build GRCh38
rs758859641 Chr.7: 135730453-135730453 on Build GRCh38
rs771327510 Chr.7: 135730300-135730300 on Build GRCh38
rs570500376 Chr.7: 135730535-135730535 on Build GRCh38
rs778335292 Chr.7: 135730511-135730511 on Build GRCh38
rs528385158 Chr.7: 135730417-135730417 on Build GRCh38
rs1021112271 Chr.7: 135730271-135730271 on Build GRCh38
rs941279707 Chr.7: 135730153-135730153 on Build GRCh38
rs983055234 Chr.7: 135730412-135730412 on Build GRCh38
rs1016099223 Chr.7: 135730154-135730154 on Build GRCh38
rs3112371 Chr.7: 135730280-135730280 on Build GRCh38
rs755456524 Chr.7: 135730206-135730206 on Build GRCh38
rs560450729 Chr.7: 135730366-135730366 on Build GRCh38
rs769387139 Chr.7: 135730279-135730279 on Build GRCh38
rs937989681 Chr.7: 135730484-135730484 on Build GRCh38
rs62489529 Chr.7: 135730423-135730423 on Build GRCh38
rs975177389 Chr.7: 135730304-135730304 on Build GRCh38
rs974996707 Chr.7: 135730536-135730536 on Build GRCh38
rs981527015 Chr.7: 135730491-135730491 on Build GRCh38
rs762657514 Chr.7: 135730367-135730367 on Build GRCh38
rs933081355 Chr.7: 135730387-135730387 on Build GRCh38
rs776325945 Chr.7: 135730200-135730200 on Build GRCh38
rs971919509 Chr.7: 135730226-135730226 on Build GRCh38
rs896677430 Chr.7: 135730266-135730266 on Build GRCh38
rs370668805 Chr.7: 135730463-135730463 on Build GRCh38
rs971400220 Chr.7: 135730476-135730476 on Build GRCh38
rs187530004 Chr.7: 135730601-135730601 on Build GRCh38
rs867802385 Chr.7: 135730227-135730227 on Build GRCh38
rs1009648219 Chr.7: 135730591-135730591 on Build GRCh38
rs943614905 Chr.7: 135730342-135730342 on Build GRCh38
rs896605517 Chr.7: 135730554-135730554 on Build GRCh38
rs1022949520 Chr.7: 135730207-135730207 on Build GRCh38
rs932965633 Chr.7: 135730571-135730571 on Build GRCh38
rs941767688 Chr.7: 135730600-135730600 on Build GRCh38
rs1006096098 Chr.7: 135730150-135730150 on Build GRCh38

View Full Product Details