Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 136868996-136869496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM2
Entrez Gene ID
1129
Gene Name
cholinergic receptor muscarinic 2
Gene Aliases
HM2
UniGene
Hs.535891
Gene Chromosome Location
Chr.7: 136868652-137020255 on Build GRCh38
Transcripts
NM_000739.2 NM_001006626.1 NM_001006627.1 NM_001006628.1 NM_001006629.1 NM_001006630.1 NM_001006631.1 NM_001006632.1

SNP Detail

SNP ID Chromosome Location
rs377562880 Chr.7: 136869091-136869091 on Build GRCh38
rs139124053 Chr.7: 136869276-136869276 on Build GRCh38
rs546346157 Chr.7: 136869116-136869116 on Build GRCh38
rs970279254 Chr.7: 136869463-136869463 on Build GRCh38
rs1009862783 Chr.7: 136869465-136869465 on Build GRCh38
rs535258435 Chr.7: 136869187-136869187 on Build GRCh38
rs529937245 Chr.7: 136869041-136869041 on Build GRCh38
rs1010765407 Chr.7: 136869110-136869110 on Build GRCh38
rs557331424 Chr.7: 136869462-136869462 on Build GRCh38
rs984900645 Chr.7: 136869050-136869050 on Build GRCh38
rs1006467481 Chr.7: 136869446-136869446 on Build GRCh38
rs1001718245 Chr.7: 136869426-136869426 on Build GRCh38
rs751987395 Chr.7: 136869441-136869441 on Build GRCh38
rs926864145 Chr.7: 136869031-136869031 on Build GRCh38
rs946583977 Chr.7: 136869371-136869371 on Build GRCh38
rs952268352 Chr.7: 136869239-136869239 on Build GRCh38
rs934638853 Chr.7: 136869105-136869105 on Build GRCh38
rs1007799155 Chr.7: 136869387-136869387 on Build GRCh38
rs566459725 Chr.7: 136869177-136869177 on Build GRCh38
rs776379815 Chr.7: 136869086-136869086 on Build GRCh38
rs879010812 Chr.7: 136869125-136869125 on Build GRCh38
rs867276560 Chr.7: 136869108-136869108 on Build GRCh38
rs981997179 Chr.7: 136869469-136869469 on Build GRCh38
rs897030506 Chr.7: 136869124-136869124 on Build GRCh38
rs1040153501 Chr.7: 136869397-136869397 on Build GRCh38
rs915993591 Chr.7: 136869076-136869076 on Build GRCh38
rs180828453 Chr.7: 136869107-136869107 on Build GRCh38
rs531990501 Chr.7: 136869308-136869308 on Build GRCh38
rs1007272471 Chr.7: 136869176-136869176 on Build GRCh38
rs928690362 Chr.7: 136869090-136869090 on Build GRCh38
rs993111330 Chr.7: 136869190-136869190 on Build GRCh38
rs928756804 Chr.7: 136869474-136869474 on Build GRCh38
rs546478333 Chr.7: 136869138-136869138 on Build GRCh38
rs928696207 Chr.7: 136869291-136869291 on Build GRCh38
rs1043831801 Chr.7: 136869070-136869076 on Build GRCh38
rs888872110 Chr.7: 136869129-136869129 on Build GRCh38
rs992319375 Chr.7: 136869042-136869042 on Build GRCh38
rs984931593 Chr.7: 136869248-136869248 on Build GRCh38
rs555161488 Chr.7: 136869227-136869227 on Build GRCh38
rs995341323 Chr.7: 136869141-136869141 on Build GRCh38
rs879186892 Chr.7: 136869184-136869184 on Build GRCh38
rs945510378 Chr.7: 136869254-136869254 on Build GRCh38
rs577160316 Chr.7: 136869384-136869384 on Build GRCh38
rs1053087821 Chr.7: 136869321-136869321 on Build GRCh38
rs763147808 Chr.7: 136869376-136869376 on Build GRCh38
rs77128879 Chr.7: 136869413-136869413 on Build GRCh38
rs1046148275 Chr.7: 136869088-136869088 on Build GRCh38
rs1025901487 Chr.7: 136869435-136869435 on Build GRCh38
rs550519660 Chr.7: 136869085-136869085 on Build GRCh38
rs932747763 Chr.7: 136869332-136869332 on Build GRCh38
rs945458080 Chr.7: 136869056-136869056 on Build GRCh38
rs141428785 Chr.7: 136869281-136869281 on Build GRCh38
rs925343303 Chr.7: 136869296-136869296 on Build GRCh38
rs993137370 Chr.7: 136869424-136869424 on Build GRCh38
rs916040648 Chr.7: 136869260-136869260 on Build GRCh38
rs557282930 Chr.7: 136869352-136869352 on Build GRCh38
rs981581400 Chr.7: 136869285-136869285 on Build GRCh38
rs960972701 Chr.7: 136869224-136869224 on Build GRCh38
rs952299470 Chr.7: 136869443-136869443 on Build GRCh38
rs1015424179 Chr.7: 136869119-136869119 on Build GRCh38
rs1021850691 Chr.7: 136869181-136869189 on Build GRCh38
rs1048275094 Chr.7: 136869410-136869410 on Build GRCh38
rs1019634106 Chr.7: 136869193-136869193 on Build GRCh38

View Full Product Details