Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137875243-137875772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs954061841 Chr.7: 137875429-137875429 on Build GRCh38
rs935095960 Chr.7: 137875483-137875483 on Build GRCh38
rs557179614 Chr.7: 137875565-137875565 on Build GRCh38
rs896221455 Chr.7: 137875583-137875583 on Build GRCh38
rs201014952 Chr.7: 137875709-137875709 on Build GRCh38
rs182725243 Chr.7: 137875501-137875501 on Build GRCh38
rs146438304 Chr.7: 137875482-137875482 on Build GRCh38
rs1027004191 Chr.7: 137875322-137875322 on Build GRCh38
rs9584 Chr.7: 137875633-137875633 on Build GRCh38
rs56313230 Chr.7: 137875319-137875319 on Build GRCh38
rs151120352 Chr.7: 137875679-137875679 on Build GRCh38
rs904053187 Chr.7: 137875632-137875632 on Build GRCh38
rs765755617 Chr.7: 137875554-137875554 on Build GRCh38
rs200321006 Chr.7: 137875706-137875706 on Build GRCh38
rs398086211 Chr.7: 137875708-137875708 on Build GRCh38
rs1024017333 Chr.7: 137875620-137875620 on Build GRCh38
rs764357067 Chr.7: 137875370-137875370 on Build GRCh38
rs1028261297 Chr.7: 137875347-137875347 on Build GRCh38
rs972302823 Chr.7: 137875471-137875471 on Build GRCh38
rs944623396 Chr.7: 137875744-137875744 on Build GRCh38
rs931680979 Chr.7: 137875680-137875680 on Build GRCh38
rs552566417 Chr.7: 137875362-137875362 on Build GRCh38
rs1042936942 Chr.7: 137875617-137875617 on Build GRCh38
rs75871069 Chr.7: 137875340-137875340 on Build GRCh38
rs1019751477 Chr.7: 137875278-137875278 on Build GRCh38
rs201338205 Chr.7: 137875707-137875707 on Build GRCh38
rs761913433 Chr.7: 137875486-137875486 on Build GRCh38
rs879219562 Chr.7: 137875648-137875648 on Build GRCh38
rs952857488 Chr.7: 137875589-137875589 on Build GRCh38
rs759197545 Chr.7: 137875441-137875441 on Build GRCh38
rs911517158 Chr.7: 137875690-137875690 on Build GRCh38
rs566379719 Chr.7: 137875440-137875440 on Build GRCh38
rs994274334 Chr.7: 137875578-137875578 on Build GRCh38
rs1008726038 Chr.7: 137875268-137875268 on Build GRCh38
rs34643720 Chr.7: 137875707-137875707 on Build GRCh38
rs796917409 Chr.7: 137875709-137875709 on Build GRCh38
rs1019649220 Chr.7: 137875700-137875700 on Build GRCh38
rs561392112 Chr.7: 137875653-137875653 on Build GRCh38
rs897636458 Chr.7: 137875577-137875577 on Build GRCh38
rs537586531 Chr.7: 137875526-137875526 on Build GRCh38
rs748767732 Chr.7: 137875659-137875659 on Build GRCh38
rs61280276 Chr.7: 137875584-137875584 on Build GRCh38
rs986817513 Chr.7: 137875433-137875433 on Build GRCh38
rs1053446131 Chr.7: 137875504-137875504 on Build GRCh38
rs552956117 Chr.7: 137875630-137875630 on Build GRCh38
rs374979405 Chr.7: 137875634-137875634 on Build GRCh38
rs971011298 Chr.7: 137875622-137875622 on Build GRCh38
rs972868153 Chr.7: 137875713-137875713 on Build GRCh38
rs931655848 Chr.7: 137875540-137875540 on Build GRCh38
rs532820509 Chr.7: 137875349-137875349 on Build GRCh38
rs1012246401 Chr.7: 137875590-137875590 on Build GRCh38
rs144036961 Chr.7: 137875333-137875333 on Build GRCh38
rs893487044 Chr.7: 137875532-137875532 on Build GRCh38
rs890316586 Chr.7: 137875267-137875267 on Build GRCh38
rs751897292 Chr.7: 137875624-137875624 on Build GRCh38
rs750874099 Chr.7: 137875557-137875557 on Build GRCh38
rs191403055 Chr.7: 137875286-137875286 on Build GRCh38
rs141081819 Chr.7: 137875682-137875682 on Build GRCh38
rs186012421 Chr.7: 137875502-137875502 on Build GRCh38
rs778995750 Chr.7: 137875688-137875688 on Build GRCh38
rs368947030 Chr.7: 137875567-137875567 on Build GRCh38
rs57979055 Chr.7: 137875574-137875574 on Build GRCh38

View Full Product Details