Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137876078-137876580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs757519564 Chr.7: 137876369-137876370 on Build GRCh38
rs570971463 Chr.7: 137876107-137876107 on Build GRCh38
rs3832470 Chr.7: 137876263-137876264 on Build GRCh38
rs755878361 Chr.7: 137876342-137876342 on Build GRCh38
rs959253249 Chr.7: 137876452-137876452 on Build GRCh38
rs188144941 Chr.7: 137876500-137876500 on Build GRCh38
rs892724616 Chr.7: 137876126-137876126 on Build GRCh38
rs959538173 Chr.7: 137876114-137876114 on Build GRCh38
rs906626660 Chr.7: 137876463-137876463 on Build GRCh38
rs1026003141 Chr.7: 137876540-137876540 on Build GRCh38
rs1019084320 Chr.7: 137876526-137876526 on Build GRCh38
rs915770808 Chr.7: 137876364-137876364 on Build GRCh38
rs542326810 Chr.7: 137876517-137876517 on Build GRCh38
rs112402602 Chr.7: 137876332-137876337 on Build GRCh38
rs1033493638 Chr.7: 137876445-137876445 on Build GRCh38
rs1024899534 Chr.7: 137876469-137876469 on Build GRCh38
rs141698455 Chr.7: 137876244-137876244 on Build GRCh38
rs764213143 Chr.7: 137876334-137876335 on Build GRCh38
rs930559361 Chr.7: 137876280-137876280 on Build GRCh38
rs867662533 Chr.7: 137876512-137876512 on Build GRCh38
rs971777445 Chr.7: 137876528-137876528 on Build GRCh38
rs192177250 Chr.7: 137876470-137876470 on Build GRCh38
rs763499347 Chr.7: 137876252-137876252 on Build GRCh38
rs192886390 Chr.7: 137876525-137876525 on Build GRCh38
rs994185204 Chr.7: 137876351-137876351 on Build GRCh38
rs1001648536 Chr.7: 137876390-137876390 on Build GRCh38
rs917549473 Chr.7: 137876142-137876142 on Build GRCh38
rs10954590 Chr.7: 137876527-137876527 on Build GRCh38
rs1045186855 Chr.7: 137876459-137876459 on Build GRCh38
rs398006521 Chr.7: 137876368-137876369 on Build GRCh38
rs539216261 Chr.7: 137876150-137876150 on Build GRCh38
rs879150776 Chr.7: 137876529-137876529 on Build GRCh38
rs948547499 Chr.7: 137876454-137876454 on Build GRCh38
rs918891253 Chr.7: 137876251-137876251 on Build GRCh38
rs992112320 Chr.7: 137876141-137876141 on Build GRCh38
rs1022478024 Chr.7: 137876172-137876172 on Build GRCh38
rs944229002 Chr.7: 137876546-137876546 on Build GRCh38
rs765236027 Chr.7: 137876200-137876200 on Build GRCh38
rs761813501 Chr.7: 137876197-137876197 on Build GRCh38
rs966415137 Chr.7: 137876159-137876159 on Build GRCh38
rs1008135975 Chr.7: 137876523-137876523 on Build GRCh38
rs563799113 Chr.7: 137876519-137876519 on Build GRCh38
rs747762663 Chr.7: 137876262-137876263 on Build GRCh38
rs778077195 Chr.7: 137876332-137876335 on Build GRCh38
rs548405273 Chr.7: 137876127-137876127 on Build GRCh38
rs397890419 Chr.7: 137876367-137876368 on Build GRCh38
rs141715174 Chr.7: 137876361-137876370 on Build GRCh38
rs936461881 Chr.7: 137876190-137876190 on Build GRCh38
rs183038030 Chr.7: 137876494-137876494 on Build GRCh38
rs200249353 Chr.7: 137876332-137876339 on Build GRCh38
rs1048466132 Chr.7: 137876381-137876381 on Build GRCh38
rs1011178686 Chr.7: 137876510-137876510 on Build GRCh38
rs796364158 Chr.7: 137876369-137876370 on Build GRCh38
rs750121072 Chr.7: 137876524-137876527 on Build GRCh38
rs79501917 Chr.7: 137876189-137876189 on Build GRCh38
rs10534110 Chr.7: 137876332-137876333 on Build GRCh38
rs888524421 Chr.7: 137876386-137876386 on Build GRCh38
rs762288372 Chr.7: 137876297-137876297 on Build GRCh38
rs1052395526 Chr.7: 137876479-137876479 on Build GRCh38
rs766838434 Chr.7: 137876380-137876380 on Build GRCh38
rs1011081438 Chr.7: 137876167-137876167 on Build GRCh38
rs951675126 Chr.7: 137876551-137876551 on Build GRCh38
rs114295063 Chr.7: 137876248-137876248 on Build GRCh38

View Full Product Details