Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137879504-137880012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

SNP Detail

SNP ID Chromosome Location
rs766105431 Chr.7: 137879811-137879811 on Build GRCh38
rs551936723 Chr.7: 137879620-137879620 on Build GRCh38
rs189720289 Chr.7: 137879778-137879778 on Build GRCh38
rs149220295 Chr.7: 137879826-137879826 on Build GRCh38
rs997014350 Chr.7: 137879791-137879791 on Build GRCh38
rs778934798 Chr.7: 137879661-137879661 on Build GRCh38
rs796593504 Chr.7: 137879940-137879940 on Build GRCh38
rs890886804 Chr.7: 137879814-137879814 on Build GRCh38
rs1015225600 Chr.7: 137879593-137879593 on Build GRCh38
rs75756974 Chr.7: 137879880-137879880 on Build GRCh38
rs980837750 Chr.7: 137879711-137879711 on Build GRCh38
rs904202164 Chr.7: 137879607-137879607 on Build GRCh38
rs181690919 Chr.7: 137879634-137879634 on Build GRCh38
rs995859062 Chr.7: 137879860-137879860 on Build GRCh38
rs991569429 Chr.7: 137879645-137879645 on Build GRCh38
rs1054699286 Chr.7: 137879743-137879743 on Build GRCh38
rs969802675 Chr.7: 137879706-137879706 on Build GRCh38
rs973994520 Chr.7: 137879619-137879619 on Build GRCh38
rs541243333 Chr.7: 137879849-137879849 on Build GRCh38
rs977512677 Chr.7: 137879932-137879932 on Build GRCh38
rs7783145 Chr.7: 137879755-137879755 on Build GRCh38
rs372335517 Chr.7: 137879946-137879947 on Build GRCh38
rs554012454 Chr.7: 137879660-137879660 on Build GRCh38
rs534427946 Chr.7: 137879642-137879642 on Build GRCh38
rs181204007 Chr.7: 137879877-137879877 on Build GRCh38
rs924608545 Chr.7: 137879705-137879705 on Build GRCh38
rs1015694020 Chr.7: 137879841-137879841 on Build GRCh38
rs960756163 Chr.7: 137879985-137879985 on Build GRCh38
rs995909158 Chr.7: 137879612-137879612 on Build GRCh38
rs567631639 Chr.7: 137879682-137879682 on Build GRCh38
rs920577587 Chr.7: 137879697-137879697 on Build GRCh38
rs371553639 Chr.7: 137879608-137879608 on Build GRCh38
rs572119409 Chr.7: 137879825-137879825 on Build GRCh38
rs1015075895 Chr.7: 137879904-137879904 on Build GRCh38
rs971470264 Chr.7: 137879926-137879926 on Build GRCh38
rs144914680 Chr.7: 137879955-137879955 on Build GRCh38
rs939789400 Chr.7: 137879979-137879979 on Build GRCh38
rs556183109 Chr.7: 137879740-137879740 on Build GRCh38
rs367700655 Chr.7: 137879777-137879777 on Build GRCh38
rs936272574 Chr.7: 137879729-137879729 on Build GRCh38
rs1010134665 Chr.7: 137879576-137879576 on Build GRCh38
rs751567078 Chr.7: 137879746-137879746 on Build GRCh38
rs959239579 Chr.7: 137879879-137879879 on Build GRCh38
rs891705407 Chr.7: 137879550-137879550 on Build GRCh38
rs921517284 Chr.7: 137879949-137879949 on Build GRCh38
rs111289860 Chr.7: 137879983-137879983 on Build GRCh38
rs962388806 Chr.7: 137879845-137879845 on Build GRCh38
rs141904248 Chr.7: 137879934-137879935 on Build GRCh38
rs924633328 Chr.7: 137879939-137879939 on Build GRCh38
rs1051278803 Chr.7: 137879958-137879958 on Build GRCh38
rs558694226 Chr.7: 137879820-137879820 on Build GRCh38
rs928034212 Chr.7: 137879721-137879721 on Build GRCh38
rs1004288753 Chr.7: 137879543-137879543 on Build GRCh38
rs950100935 Chr.7: 137879694-137879694 on Build GRCh38
rs1024732080 Chr.7: 137879901-137879901 on Build GRCh38
rs763932060 Chr.7: 137879944-137879944 on Build GRCh38
rs35521387 Chr.7: 137879977-137879978 on Build GRCh38
rs545204679 Chr.7: 137879760-137879760 on Build GRCh38
rs753605407 Chr.7: 137879978-137879978 on Build GRCh38
rs186926380 Chr.7: 137879695-137879695 on Build GRCh38
rs543432868 Chr.7: 137879952-137879952 on Build GRCh38

View Full Product Details