Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137882274-137882810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416976 c.1487T>A p.L496Q Chr.7: 137882412-137882412 on Build GRCh38
2859690 c.1331C>T p.P444L Chr.7: 137882568-137882568 on Build GRCh38
2859688 c.1406C>T p.P469L Chr.7: 137882493-137882493 on Build GRCh38
5793459 c.1396G>T p.E466* Chr.7: 137882503-137882503 on Build GRCh38
1085972 c.1329C>T p.S443S Chr.7: 137882570-137882570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781089222 Chr.7: 137882380-137882380 on Build GRCh38
rs775090315 Chr.7: 137882573-137882573 on Build GRCh38
rs1007457881 Chr.7: 137882433-137882433 on Build GRCh38
rs55868765 Chr.7: 137882340-137882340 on Build GRCh38
rs775318022 Chr.7: 137882528-137882528 on Build GRCh38
rs566427092 Chr.7: 137882561-137882561 on Build GRCh38
rs1032508973 Chr.7: 137882412-137882412 on Build GRCh38
rs780077410 Chr.7: 137882548-137882548 on Build GRCh38
rs946782789 Chr.7: 137882444-137882444 on Build GRCh38
rs186583183 Chr.7: 137882714-137882714 on Build GRCh38
rs375939444 Chr.7: 137882493-137882493 on Build GRCh38
rs916432914 Chr.7: 137882510-137882510 on Build GRCh38
rs959016053 Chr.7: 137882769-137882769 on Build GRCh38
rs776625124 Chr.7: 137882515-137882515 on Build GRCh38
rs745786624 Chr.7: 137882571-137882571 on Build GRCh38
rs951694675 Chr.7: 137882738-137882738 on Build GRCh38
rs766471490 Chr.7: 137882670-137882670 on Build GRCh38
rs779966108 Chr.7: 137882786-137882786 on Build GRCh38
rs145559600 Chr.7: 137882567-137882567 on Build GRCh38
rs868685052 Chr.7: 137882590-137882590 on Build GRCh38
rs376688758 Chr.7: 137882587-137882587 on Build GRCh38
rs750933727 Chr.7: 137882545-137882545 on Build GRCh38
rs779420003 Chr.7: 137882552-137882553 on Build GRCh38
rs151002001 Chr.7: 137882462-137882462 on Build GRCh38
rs114314625 Chr.7: 137882384-137882384 on Build GRCh38
rs770705547 Chr.7: 137882504-137882504 on Build GRCh38
rs750406588 Chr.7: 137882481-137882481 on Build GRCh38
rs752139663 Chr.7: 137882475-137882475 on Build GRCh38
rs755146141 Chr.7: 137882549-137882549 on Build GRCh38
rs764519676 Chr.7: 137882470-137882470 on Build GRCh38
rs941854584 Chr.7: 137882332-137882332 on Build GRCh38
rs780317387 Chr.7: 137882490-137882490 on Build GRCh38
rs910050415 Chr.7: 137882343-137882343 on Build GRCh38
rs762651648 Chr.7: 137882530-137882530 on Build GRCh38
rs988505664 Chr.7: 137882730-137882730 on Build GRCh38
rs137882853 Chr.7: 137882602-137882602 on Build GRCh38
rs750048206 Chr.7: 137882417-137882418 on Build GRCh38
rs143590560 Chr.7: 137882411-137882411 on Build GRCh38
rs980922013 Chr.7: 137882694-137882694 on Build GRCh38
rs758879821 Chr.7: 137882468-137882468 on Build GRCh38
rs769321233 Chr.7: 137882526-137882526 on Build GRCh38
rs371784147 Chr.7: 137882381-137882381 on Build GRCh38
rs774203532 Chr.7: 137882404-137882404 on Build GRCh38
rs141128498 Chr.7: 137882465-137882465 on Build GRCh38
rs764573030 Chr.7: 137882537-137882537 on Build GRCh38
rs768091777 Chr.7: 137882712-137882712 on Build GRCh38
rs140287420 Chr.7: 137882562-137882562 on Build GRCh38
rs373493579 Chr.7: 137882477-137882477 on Build GRCh38
rs376504453 Chr.7: 137882478-137882478 on Build GRCh38
rs753042980 Chr.7: 137882370-137882370 on Build GRCh38
rs779511217 Chr.7: 137882512-137882513 on Build GRCh38
rs913897541 Chr.7: 137882739-137882739 on Build GRCh38
rs965596624 Chr.7: 137882446-137882446 on Build GRCh38
rs761208117 Chr.7: 137882632-137882632 on Build GRCh38
rs145388881 Chr.7: 137882568-137882568 on Build GRCh38
rs748608400 Chr.7: 137882653-137882654 on Build GRCh38
rs762389207 Chr.7: 137882544-137882544 on Build GRCh38
rs558679935 Chr.7: 137882492-137882492 on Build GRCh38
rs769140806 Chr.7: 137882538-137882539 on Build GRCh38
rs202115793 Chr.7: 137882441-137882441 on Build GRCh38
rs79226062 Chr.7: 137882375-137882375 on Build GRCh38
rs145304675 Chr.7: 137882702-137882702 on Build GRCh38
rs984514456 Chr.7: 137882747-137882747 on Build GRCh38
rs1016957290 Chr.7: 137882755-137882755 on Build GRCh38
rs368005647 Chr.7: 137882584-137882584 on Build GRCh38
rs909009376 Chr.7: 137882318-137882318 on Build GRCh38
rs751916803 Chr.7: 137882539-137882539 on Build GRCh38
rs199561057 Chr.7: 137882595-137882595 on Build GRCh38
rs946730089 Chr.7: 137882759-137882759 on Build GRCh38
rs1043870539 Chr.7: 137882457-137882457 on Build GRCh38
rs368855415 Chr.7: 137882379-137882379 on Build GRCh38
rs753845813 Chr.7: 137882672-137882672 on Build GRCh38
rs756198124 Chr.7: 137882546-137882546 on Build GRCh38
rs768358209 Chr.7: 137882586-137882586 on Build GRCh38
rs765646277 Chr.7: 137882364-137882364 on Build GRCh38
rs149450359 Chr.7: 137882603-137882603 on Build GRCh38
rs750937382 Chr.7: 137882622-137882622 on Build GRCh38
rs1043815599 Chr.7: 137882772-137882772 on Build GRCh38
rs748865684 Chr.7: 137882536-137882536 on Build GRCh38
rs369406871 Chr.7: 137882429-137882429 on Build GRCh38
rs748717116 Chr.7: 137882475-137882475 on Build GRCh38
rs762706707 Chr.7: 137882580-137882580 on Build GRCh38
rs144732810 Chr.7: 137882486-137882486 on Build GRCh38
rs746315965 Chr.7: 137882688-137882688 on Build GRCh38
rs758830032 Chr.7: 137882374-137882374 on Build GRCh38
rs370472182 Chr.7: 137882519-137882519 on Build GRCh38
rs140163626 Chr.7: 137882491-137882491 on Build GRCh38
rs759230081 Chr.7: 137882463-137882463 on Build GRCh38

View Full Product Details