Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138732489-138733028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A4
Entrez Gene ID
50617
Gene Name
ATPase H+ transporting V0 subunit a4
Gene Aliases
A4, ATP6N1B, ATP6N2, RDRTA2, RTA1C, RTADR, STV1, VPH1, VPP2
UniGene
Hs.98967
Gene Chromosome Location
Chr.7: 138706294-138799839 on Build GRCh38
Transcripts
NM_020632.2 NM_130840.2 NM_130841.2 XM_005250393.1 XM_005250394.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6270938 c.1810C>A p.H604N Chr.7: 138732975-138732975 on Build GRCh38
7111919 c.1851C>G p.I617M Chr.7: 138732934-138732934 on Build GRCh38
484905 c.1886A>G p.N629S Chr.7: 138732899-138732899 on Build GRCh38
1086030 c.1758G>A p.L586L Chr.7: 138733027-138733027 on Build GRCh38
5674593 c.1908G>C p.Q636H Chr.7: 138732877-138732877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777449116 Chr.7: 138732846-138732846 on Build GRCh38
rs10273800 Chr.7: 138732655-138732655 on Build GRCh38
rs773128017 Chr.7: 138732952-138732952 on Build GRCh38
rs373030847 Chr.7: 138732663-138732663 on Build GRCh38
rs780506721 Chr.7: 138732905-138732905 on Build GRCh38
rs368166747 Chr.7: 138732867-138732867 on Build GRCh38
rs57692175 Chr.7: 138732852-138732852 on Build GRCh38
rs141568521 Chr.7: 138732979-138732979 on Build GRCh38
rs1042099841 Chr.7: 138732690-138732690 on Build GRCh38
rs371249920 Chr.7: 138732717-138732717 on Build GRCh38
rs73730479 Chr.7: 138732897-138732897 on Build GRCh38
rs966147198 Chr.7: 138732625-138732625 on Build GRCh38
rs972587713 Chr.7: 138732632-138732632 on Build GRCh38
rs3778693 Chr.7: 138732682-138732682 on Build GRCh38
rs914547246 Chr.7: 138732519-138732519 on Build GRCh38
rs538208659 Chr.7: 138732660-138732660 on Build GRCh38
rs143883618 Chr.7: 138732937-138732937 on Build GRCh38
rs775648355 Chr.7: 138732876-138732876 on Build GRCh38
rs889708886 Chr.7: 138732734-138732734 on Build GRCh38
rs1049554810 Chr.7: 138732719-138732719 on Build GRCh38
rs903627584 Chr.7: 138732692-138732692 on Build GRCh38
rs557868826 Chr.7: 138732662-138732662 on Build GRCh38
rs749701746 Chr.7: 138732936-138732936 on Build GRCh38
rs536077763 Chr.7: 138732644-138732644 on Build GRCh38
rs558988577 Chr.7: 138732868-138732868 on Build GRCh38
rs1004521517 Chr.7: 138732959-138732959 on Build GRCh38
rs758371610 Chr.7: 138732834-138732834 on Build GRCh38
rs903722497 Chr.7: 138732563-138732563 on Build GRCh38
rs748730087 Chr.7: 138732668-138732668 on Build GRCh38
rs556261104 Chr.7: 138732751-138732751 on Build GRCh38
rs141741258 Chr.7: 138732608-138732608 on Build GRCh38
rs764823481 Chr.7: 138732866-138732868 on Build GRCh38
rs774903056 Chr.7: 138732942-138732942 on Build GRCh38
rs992847393 Chr.7: 138732941-138732941 on Build GRCh38
rs779720368 Chr.7: 138733004-138733004 on Build GRCh38
rs796363484 Chr.7: 138732864-138732864 on Build GRCh38
rs549185406 Chr.7: 138732961-138732961 on Build GRCh38
rs776966607 Chr.7: 138732972-138732972 on Build GRCh38
rs774817040 Chr.7: 138732890-138732890 on Build GRCh38
rs145939630 Chr.7: 138732946-138732946 on Build GRCh38
rs1018933628 Chr.7: 138732743-138732743 on Build GRCh38
rs143558330 Chr.7: 138732892-138732892 on Build GRCh38
rs374330946 Chr.7: 138732978-138732978 on Build GRCh38
rs1020776812 Chr.7: 138732553-138732553 on Build GRCh38
rs780967135 Chr.7: 138732862-138732862 on Build GRCh38
rs965796895 Chr.7: 138732688-138732688 on Build GRCh38
rs398111944 Chr.7: 138732864-138732865 on Build GRCh38
rs755678100 Chr.7: 138732929-138732929 on Build GRCh38
rs751558951 Chr.7: 138732974-138732974 on Build GRCh38
rs745537081 Chr.7: 138732924-138732924 on Build GRCh38
rs769099810 Chr.7: 138732886-138732886 on Build GRCh38
rs148944306 Chr.7: 138732851-138732852 on Build GRCh38
rs368553213 Chr.7: 138732993-138732993 on Build GRCh38
rs990467932 Chr.7: 138732750-138732750 on Build GRCh38
rs966464299 Chr.7: 138732769-138732769 on Build GRCh38
rs3778694 Chr.7: 138732592-138732592 on Build GRCh38
rs745874243 Chr.7: 138732840-138732841 on Build GRCh38
rs771998055 Chr.7: 138732898-138732898 on Build GRCh38
rs201399022 Chr.7: 138732909-138732909 on Build GRCh38
rs751542587 Chr.7: 138732901-138732901 on Build GRCh38
rs879005610 Chr.7: 138732852-138732853 on Build GRCh38
rs1000675093 Chr.7: 138732570-138732570 on Build GRCh38
rs144864467 Chr.7: 138732545-138732545 on Build GRCh38
rs748638764 Chr.7: 138732945-138732945 on Build GRCh38
rs563280125 Chr.7: 138732853-138732854 on Build GRCh38
rs370658535 Chr.7: 138732893-138732893 on Build GRCh38
rs753542791 Chr.7: 138732854-138732855 on Build GRCh38
rs760974203 Chr.7: 138732960-138732960 on Build GRCh38
rs564775348 Chr.7: 138732554-138732554 on Build GRCh38
rs576079818 Chr.7: 138732823-138732823 on Build GRCh38
rs57971062 Chr.7: 138732806-138732807 on Build GRCh38
rs554005711 Chr.7: 138732667-138732667 on Build GRCh38
rs905462071 Chr.7: 138732514-138732514 on Build GRCh38
rs997328514 Chr.7: 138732709-138732709 on Build GRCh38
rs752686315 Chr.7: 138732832-138732832 on Build GRCh38
rs543393663 Chr.7: 138732853-138732853 on Build GRCh38
rs751052544 Chr.7: 138732982-138732982 on Build GRCh38
rs10247054 Chr.7: 138732735-138732735 on Build GRCh38
rs533881229 Chr.7: 138732664-138732664 on Build GRCh38
rs774118172 Chr.7: 138732850-138732851 on Build GRCh38
rs540574853 Chr.7: 138732931-138732931 on Build GRCh38
rs756800454 Chr.7: 138732852-138732852 on Build GRCh38
rs543791385 Chr.7: 138732708-138732708 on Build GRCh38
rs896196338 Chr.7: 138732529-138732529 on Build GRCh38
rs3807154 Chr.7: 138732973-138732973 on Build GRCh38
rs763125336 Chr.7: 138732904-138732904 on Build GRCh38
rs1008127519 Chr.7: 138732995-138732995 on Build GRCh38
rs1019403216 Chr.7: 138732616-138732616 on Build GRCh38
rs186686461 Chr.7: 138732520-138732520 on Build GRCh38
rs574484944 Chr.7: 138732852-138732853 on Build GRCh38
rs746570967 Chr.7: 138732849-138732849 on Build GRCh38
rs554812164 Chr.7: 138732646-138732646 on Build GRCh38
rs769919220 Chr.7: 138732841-138732845 on Build GRCh38
rs749821651 Chr.7: 138732878-138732878 on Build GRCh38
rs538346979 Chr.7: 138732522-138732522 on Build GRCh38
rs767489604 Chr.7: 138732891-138732891 on Build GRCh38
rs138711284 Chr.7: 138732581-138732581 on Build GRCh38
rs953269044 Chr.7: 138732736-138732736 on Build GRCh38

View Full Product Details