Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139544521-139545029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC2L
Entrez Gene ID
154790
Gene Name
C-type lectin domain family 2 member L
Gene Aliases
-
UniGene
Hs.57806
Gene Chromosome Location
Chr.7: 139523928-139544985 on Build GRCh38
Transcripts
NM_001080511.2 XM_017011770.1

SNP Detail

SNP ID Chromosome Location
rs996045087 Chr.7: 139544956-139544956 on Build GRCh38
rs914469089 Chr.7: 139544860-139544860 on Build GRCh38
rs530581517 Chr.7: 139544684-139544684 on Build GRCh38
rs1027188368 Chr.7: 139544984-139544984 on Build GRCh38
rs73477452 Chr.7: 139544889-139544889 on Build GRCh38
rs1055310526 Chr.7: 139544965-139544965 on Build GRCh38
rs773801945 Chr.7: 139544606-139544606 on Build GRCh38
rs538576965 Chr.7: 139544820-139544820 on Build GRCh38
rs35805331 Chr.7: 139544588-139544588 on Build GRCh38
rs546469865 Chr.7: 139544785-139544785 on Build GRCh38
rs777010045 Chr.7: 139544941-139544942 on Build GRCh38
rs79893044 Chr.7: 139544874-139544874 on Build GRCh38
rs148542431 Chr.7: 139544688-139544688 on Build GRCh38
rs906898276 Chr.7: 139544888-139544888 on Build GRCh38
rs929705205 Chr.7: 139544886-139544886 on Build GRCh38
rs1041441079 Chr.7: 139544625-139544625 on Build GRCh38
rs771450212 Chr.7: 139544765-139544765 on Build GRCh38
rs965413603 Chr.7: 139544812-139544812 on Build GRCh38
rs887981052 Chr.7: 139544767-139544767 on Build GRCh38
rs985312119 Chr.7: 139544839-139544839 on Build GRCh38
rs1034502887 Chr.7: 139544675-139544675 on Build GRCh38
rs1034308962 Chr.7: 139544777-139544777 on Build GRCh38
rs929736536 Chr.7: 139544741-139544741 on Build GRCh38
rs1025002693 Chr.7: 139544788-139544788 on Build GRCh38
rs888200389 Chr.7: 139544890-139544890 on Build GRCh38
rs551323608 Chr.7: 139544615-139544615 on Build GRCh38
rs769989628 Chr.7: 139544566-139544566 on Build GRCh38
rs536741453 Chr.7: 139544940-139544940 on Build GRCh38
rs911103471 Chr.7: 139544850-139544850 on Build GRCh38
rs11979954 Chr.7: 139544668-139544668 on Build GRCh38
rs931769098 Chr.7: 139544824-139544824 on Build GRCh38
rs966778793 Chr.7: 139544594-139544595 on Build GRCh38
rs975440650 Chr.7: 139544813-139544813 on Build GRCh38
rs969665315 Chr.7: 139544841-139544841 on Build GRCh38
rs1002649519 Chr.7: 139544669-139544669 on Build GRCh38
rs139977219 Chr.7: 139544995-139544995 on Build GRCh38
rs921791426 Chr.7: 139544559-139544559 on Build GRCh38
rs912600675 Chr.7: 139544798-139544798 on Build GRCh38
rs142911271 Chr.7: 139544692-139544692 on Build GRCh38
rs957830652 Chr.7: 139544836-139544836 on Build GRCh38
rs1006402486 Chr.7: 139544773-139544773 on Build GRCh38
rs571171886 Chr.7: 139544809-139544809 on Build GRCh38
rs367916571 Chr.7: 139544595-139544596 on Build GRCh38
rs982737631 Chr.7: 139544744-139544744 on Build GRCh38
rs921377627 Chr.7: 139544814-139544814 on Build GRCh38
rs893056058 Chr.7: 139544619-139544619 on Build GRCh38
rs895417889 Chr.7: 139544778-139544778 on Build GRCh38
rs896885938 Chr.7: 139544698-139544698 on Build GRCh38
rs1055926059 Chr.7: 139544923-139544923 on Build GRCh38
rs959601346 Chr.7: 139544560-139544560 on Build GRCh38
rs1009875280 Chr.7: 139544620-139544620 on Build GRCh38
rs917970034 Chr.7: 139544575-139544575 on Build GRCh38
rs1054303777 Chr.7: 139544852-139544852 on Build GRCh38
rs995706335 Chr.7: 139544691-139544691 on Build GRCh38
rs985813296 Chr.7: 139544682-139544682 on Build GRCh38
rs945988754 Chr.7: 139544869-139544869 on Build GRCh38
rs559190572 Chr.7: 139544690-139544690 on Build GRCh38
rs951669208 Chr.7: 139544697-139544697 on Build GRCh38
rs532939957 Chr.7: 139544578-139544578 on Build GRCh38
rs943999985 Chr.7: 139544866-139544866 on Build GRCh38

View Full Product Details