Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140776775-140777186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1448605 c.1525C>T p.R509* Chr.7: 140777081-140777081 on Build GRCh38
5872003 c.1691T>C p.M564T Chr.7: 140776915-140776915 on Build GRCh38
6349404 c.1605C>A p.S535S Chr.7: 140777001-140777001 on Build GRCh38
6919719 c.1526G>A p.R509Q Chr.7: 140777080-140777080 on Build GRCh38
5406679 c.1587C>T p.T529T Chr.7: 140777019-140777019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397507479 Chr.7: 140777011-140777011 on Build GRCh38
rs772768251 Chr.7: 140776995-140776995 on Build GRCh38
rs368578780 Chr.7: 140776899-140776899 on Build GRCh38
rs1057517930 Chr.7: 140776910-140776910 on Build GRCh38
rs761334141 Chr.7: 140776866-140776866 on Build GRCh38
rs779055779 Chr.7: 140776891-140776891 on Build GRCh38
rs763895273 Chr.7: 140776971-140776971 on Build GRCh38
rs777991153 Chr.7: 140777115-140777115 on Build GRCh38
rs869025340 Chr.7: 140777032-140777032 on Build GRCh38
rs184144181 Chr.7: 140776898-140776898 on Build GRCh38
rs750933612 Chr.7: 140776867-140776867 on Build GRCh38
rs759843625 Chr.7: 140777037-140777037 on Build GRCh38
rs1007345025 Chr.7: 140777161-140777161 on Build GRCh38
rs180177041 Chr.7: 140777006-140777006 on Build GRCh38
rs940384157 Chr.7: 140777093-140777093 on Build GRCh38
rs756413322 Chr.7: 140776882-140776882 on Build GRCh38
rs62487918 Chr.7: 140777136-140777136 on Build GRCh38
rs1050253104 Chr.7: 140777131-140777131 on Build GRCh38
rs758498850 Chr.7: 140777097-140777097 on Build GRCh38
rs775147056 Chr.7: 140776852-140776852 on Build GRCh38
rs761240860 Chr.7: 140777004-140777004 on Build GRCh38
rs779875213 Chr.7: 140777125-140777125 on Build GRCh38
rs371497168 Chr.7: 140776989-140776989 on Build GRCh38
rs551111837 Chr.7: 140776979-140776979 on Build GRCh38
rs749083550 Chr.7: 140776949-140776949 on Build GRCh38
rs745783052 Chr.7: 140776916-140776916 on Build GRCh38
rs982876525 Chr.7: 140777001-140777001 on Build GRCh38
rs774022437 Chr.7: 140776968-140776968 on Build GRCh38
rs199745091 Chr.7: 140777095-140777095 on Build GRCh38
rs960852908 Chr.7: 140776859-140776859 on Build GRCh38
rs768303459 Chr.7: 140776964-140776964 on Build GRCh38
rs369261757 Chr.7: 140777111-140777111 on Build GRCh38
rs766753485 Chr.7: 140776885-140776885 on Build GRCh38
rs969439754 Chr.7: 140776860-140776860 on Build GRCh38
rs552471935 Chr.7: 140777034-140777034 on Build GRCh38
rs749069319 Chr.7: 140777132-140777132 on Build GRCh38
rs367578527 Chr.7: 140777116-140777116 on Build GRCh38
rs606231228 Chr.7: 140777013-140777013 on Build GRCh38
rs781213584 Chr.7: 140776909-140776909 on Build GRCh38
rs766837571 Chr.7: 140777033-140777033 on Build GRCh38
rs978315054 Chr.7: 140776832-140776832 on Build GRCh38
rs760524720 Chr.7: 140776939-140776939 on Build GRCh38
rs397507478 Chr.7: 140777014-140777014 on Build GRCh38
rs756706958 Chr.7: 140776967-140776967 on Build GRCh38
rs547857147 Chr.7: 140776886-140776886 on Build GRCh38
rs149702741 Chr.7: 140776974-140776974 on Build GRCh38
rs878854675 Chr.7: 140776945-140776945 on Build GRCh38
rs768338880 Chr.7: 140777134-140777134 on Build GRCh38
rs924201178 Chr.7: 140776836-140776836 on Build GRCh38
rs765463149 Chr.7: 140777085-140777085 on Build GRCh38
rs912739724 Chr.7: 140777098-140777098 on Build GRCh38
rs769683898 Chr.7: 140776938-140776938 on Build GRCh38
rs201647306 Chr.7: 140777124-140777124 on Build GRCh38

View Full Product Details