Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142940608-142941112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf34
Entrez Gene ID
135927
Gene Name
chromosome 7 open reading frame 34
Gene Aliases
ctm-1
UniGene
Hs.239676
Gene Chromosome Location
Chr.7: 142939506-142940870 on Build GRCh38
Transcripts
NM_178829.4

SNP Detail

SNP ID Chromosome Location
rs1002936225 Chr.7: 142940894-142940894 on Build GRCh38
rs8176050 Chr.7: 142940884-142940884 on Build GRCh38
rs112178188 Chr.7: 142940702-142940702 on Build GRCh38
rs183990249 Chr.7: 142940723-142940723 on Build GRCh38
rs780297748 Chr.7: 142940670-142940670 on Build GRCh38
rs565859772 Chr.7: 142940680-142940680 on Build GRCh38
rs757198325 Chr.7: 142940653-142940653 on Build GRCh38
rs866334809 Chr.7: 142940695-142940695 on Build GRCh38
rs1020262569 Chr.7: 142940885-142940885 on Build GRCh38
rs556777585 Chr.7: 142940810-142940810 on Build GRCh38
rs897495418 Chr.7: 142940804-142940804 on Build GRCh38
rs572655861 Chr.7: 142940920-142940920 on Build GRCh38
rs78529453 Chr.7: 142940696-142940696 on Build GRCh38
rs187619642 Chr.7: 142940871-142940871 on Build GRCh38
rs545771549 Chr.7: 142940952-142940952 on Build GRCh38
rs370859435 Chr.7: 142941067-142941067 on Build GRCh38
rs1007454734 Chr.7: 142940738-142940738 on Build GRCh38
rs910274756 Chr.7: 142941086-142941086 on Build GRCh38
rs909172682 Chr.7: 142940993-142940993 on Build GRCh38
rs960139634 Chr.7: 142940647-142940647 on Build GRCh38
rs755886137 Chr.7: 142940672-142940672 on Build GRCh38
rs1036014216 Chr.7: 142940776-142940776 on Build GRCh38
rs958595867 Chr.7: 142940908-142940908 on Build GRCh38
rs968980331 Chr.7: 142940960-142940960 on Build GRCh38
rs1035986110 Chr.7: 142940668-142940669 on Build GRCh38
rs759289921 Chr.7: 142940633-142940633 on Build GRCh38
rs999543178 Chr.7: 142940968-142940968 on Build GRCh38
rs755284252 Chr.7: 142940761-142940761 on Build GRCh38
rs577260587 Chr.7: 142940734-142940734 on Build GRCh38
rs971704412 Chr.7: 142941018-142941018 on Build GRCh38
rs111344092 Chr.7: 142940735-142940735 on Build GRCh38
rs940394165 Chr.7: 142940660-142940660 on Build GRCh38
rs769662660 Chr.7: 142940636-142940636 on Build GRCh38
rs74471393 Chr.7: 142941049-142941049 on Build GRCh38
rs1034611344 Chr.7: 142940901-142940901 on Build GRCh38
rs933082106 Chr.7: 142941056-142941056 on Build GRCh38
rs1044971550 Chr.7: 142940727-142940727 on Build GRCh38
rs8176049 Chr.7: 142940891-142940891 on Build GRCh38
rs1051462405 Chr.7: 142940849-142940849 on Build GRCh38
rs776608687 Chr.7: 142940630-142940630 on Build GRCh38
rs924631600 Chr.7: 142940634-142940634 on Build GRCh38
rs893375862 Chr.7: 142940876-142940876 on Build GRCh38
rs960048189 Chr.7: 142941005-142941005 on Build GRCh38
rs534153663 Chr.7: 142940716-142940716 on Build GRCh38
rs553617889 Chr.7: 142940676-142940676 on Build GRCh38
rs1031645185 Chr.7: 142940998-142940998 on Build GRCh38

View Full Product Details