Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143321626-143322046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN1
Entrez Gene ID
1180
Gene Name
chloride voltage-gated channel 1
Gene Aliases
CLC1
UniGene
Hs.121483
Gene Chromosome Location
Chr.7: 143316126-143352008 on Build GRCh38
Transcripts
NM_000083.2 NR_046453.1 XM_017011739.1 XM_017011740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3635180 c.687G>A p.V229V Chr.7: 143321839-143321839 on Build GRCh38
4556523 c.694G>A p.E232K Chr.7: 143321846-143321846 on Build GRCh38
3635177 c.582G>A p.M194I Chr.7: 143321734-143321734 on Build GRCh38
1086740 c.622A>T p.T208S Chr.7: 143321774-143321774 on Build GRCh38
6312044 c.677G>T p.G226V Chr.7: 143321829-143321829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374154132 Chr.7: 143321782-143321782 on Build GRCh38
rs201113768 Chr.7: 143321787-143321787 on Build GRCh38
rs765064204 Chr.7: 143321696-143321696 on Build GRCh38
rs765869090 Chr.7: 143321858-143321858 on Build GRCh38
rs759508927 Chr.7: 143321751-143321751 on Build GRCh38
rs775413124 Chr.7: 143321764-143321764 on Build GRCh38
rs923380712 Chr.7: 143321848-143321848 on Build GRCh38
rs121912810 Chr.7: 143321718-143321718 on Build GRCh38
rs774268769 Chr.7: 143321836-143321836 on Build GRCh38
rs763640926 Chr.7: 143321707-143321707 on Build GRCh38
rs913819667 Chr.7: 143321973-143321973 on Build GRCh38
rs267601360 Chr.7: 143321765-143321765 on Build GRCh38
rs754300978 Chr.7: 143321798-143321798 on Build GRCh38
rs756694573 Chr.7: 143321921-143321921 on Build GRCh38
rs756755417 Chr.7: 143321789-143321789 on Build GRCh38
rs749614246 Chr.7: 143321726-143321726 on Build GRCh38
rs781408809 Chr.7: 143321925-143321925 on Build GRCh38
rs749577673 Chr.7: 143321681-143321681 on Build GRCh38
rs775295032 Chr.7: 143321821-143321821 on Build GRCh38
rs567732602 Chr.7: 143321888-143321888 on Build GRCh38
rs542831318 Chr.7: 143321661-143321661 on Build GRCh38
rs746401512 Chr.7: 143321721-143321721 on Build GRCh38
rs986558615 Chr.7: 143321900-143321900 on Build GRCh38
rs189963844 Chr.7: 143321804-143321804 on Build GRCh38
rs763883834 Chr.7: 143321783-143321783 on Build GRCh38
rs1052093517 Chr.7: 143321930-143321930 on Build GRCh38
rs137932654 Chr.7: 143321952-143321952 on Build GRCh38
rs770986156 Chr.7: 143321818-143321818 on Build GRCh38
rs1050437727 Chr.7: 143321742-143321742 on Build GRCh38
rs185444578 Chr.7: 143321688-143321688 on Build GRCh38
rs80356685 Chr.7: 143321744-143321744 on Build GRCh38
rs748326003 Chr.7: 143321733-143321733 on Build GRCh38
rs768360914 Chr.7: 143321834-143321834 on Build GRCh38
rs369773321 Chr.7: 143321720-143321720 on Build GRCh38
rs35058443 Chr.7: 143321996-143321997 on Build GRCh38
rs145502082 Chr.7: 143321737-143321737 on Build GRCh38
rs756669568 Chr.7: 143321724-143321724 on Build GRCh38
rs1046200989 Chr.7: 143321998-143321998 on Build GRCh38
rs892148550 Chr.7: 143321948-143321948 on Build GRCh38
rs945238759 Chr.7: 143321992-143321992 on Build GRCh38
rs138246784 Chr.7: 143321785-143321785 on Build GRCh38
rs780434904 Chr.7: 143321794-143321794 on Build GRCh38
rs769612798 Chr.7: 143321753-143321753 on Build GRCh38
rs780511123 Chr.7: 143321725-143321725 on Build GRCh38
rs868186189 Chr.7: 143321892-143321892 on Build GRCh38
rs144109732 Chr.7: 143321803-143321803 on Build GRCh38
rs200579591 Chr.7: 143321694-143321694 on Build GRCh38
rs147317366 Chr.7: 143321815-143321815 on Build GRCh38
rs566080451 Chr.7: 143321728-143321728 on Build GRCh38
rs762908937 Chr.7: 143321822-143321822 on Build GRCh38
rs797045032 Chr.7: 143321720-143321721 on Build GRCh38
rs933342820 Chr.7: 143321735-143321735 on Build GRCh38
rs555518498 Chr.7: 143321881-143321881 on Build GRCh38
rs760103429 Chr.7: 143321854-143321854 on Build GRCh38
rs761601545 Chr.7: 143321837-143321837 on Build GRCh38
rs201404573 Chr.7: 143321706-143321706 on Build GRCh38
rs754320081 Chr.7: 143321869-143321869 on Build GRCh38
rs143506735 Chr.7: 143321747-143321747 on Build GRCh38
rs2272258 Chr.7: 143321657-143321657 on Build GRCh38
rs958957213 Chr.7: 143321663-143321663 on Build GRCh38
rs80356700 Chr.7: 143321841-143321841 on Build GRCh38
rs778411246 Chr.7: 143321669-143321670 on Build GRCh38
rs747078264 Chr.7: 143321816-143321816 on Build GRCh38
rs868784999 Chr.7: 143321846-143321846 on Build GRCh38
rs755485162 Chr.7: 143321874-143321874 on Build GRCh38
rs41276048 Chr.7: 143321692-143321692 on Build GRCh38
rs963470037 Chr.7: 143321768-143321768 on Build GRCh38
rs761511512 Chr.7: 143321784-143321784 on Build GRCh38
rs751167296 Chr.7: 143321717-143321717 on Build GRCh38
rs752139031 Chr.7: 143321874-143321875 on Build GRCh38
rs748415883 Chr.7: 143321814-143321814 on Build GRCh38
rs865899737 Chr.7: 143321853-143321853 on Build GRCh38
rs768733291 Chr.7: 143321684-143321684 on Build GRCh38
rs80356686 Chr.7: 143321729-143321729 on Build GRCh38

View Full Product Details