Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143359645-143360150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131B
Entrez Gene ID
9715
Gene Name
family with sequence similarity 131 member B
Gene Aliases
-
UniGene
Hs.648908
Gene Chromosome Location
Chr.7: 143353400-143382304 on Build GRCh38
Transcripts
NM_001031690.2 NM_001278297.1 NM_014690.4 XM_005250073.3 XM_006716186.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406837 c.36G>A p.E12E Chr.7: 143360142-143360142 on Build GRCh38
5530812 c.57G>C p.K19N Chr.7: 143360121-143360121 on Build GRCh38
5510266 c.124C>T p.R42W Chr.7: 143360054-143360054 on Build GRCh38
3922974 c.40A>T p.I14F Chr.7: 143360138-143360138 on Build GRCh38
3258174 c.108C>T p.H36H Chr.7: 143360070-143360070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916834829 Chr.7: 143359913-143359913 on Build GRCh38
rs751568716 Chr.7: 143360057-143360057 on Build GRCh38
rs751542540 Chr.7: 143359704-143359704 on Build GRCh38
rs565735952 Chr.7: 143359757-143359757 on Build GRCh38
rs752893250 Chr.7: 143360046-143360046 on Build GRCh38
rs778972276 Chr.7: 143360068-143360068 on Build GRCh38
rs895385654 Chr.7: 143359674-143359676 on Build GRCh38
rs866267391 Chr.7: 143359768-143359768 on Build GRCh38
rs757532524 Chr.7: 143359899-143359899 on Build GRCh38
rs747792970 Chr.7: 143360093-143360093 on Build GRCh38
rs1047537441 Chr.7: 143360051-143360051 on Build GRCh38
rs374926130 Chr.7: 143359711-143359711 on Build GRCh38
rs898449443 Chr.7: 143360043-143360043 on Build GRCh38
rs375053726 Chr.7: 143360100-143360100 on Build GRCh38
rs771365837 Chr.7: 143360110-143360110 on Build GRCh38
rs759903895 Chr.7: 143360026-143360026 on Build GRCh38
rs555949032 Chr.7: 143359922-143359922 on Build GRCh38
rs776874595 Chr.7: 143360024-143360024 on Build GRCh38
rs756000946 Chr.7: 143359745-143359745 on Build GRCh38
rs755162793 Chr.7: 143359696-143359696 on Build GRCh38
rs928826302 Chr.7: 143359901-143359901 on Build GRCh38
rs368299887 Chr.7: 143359686-143359686 on Build GRCh38
rs535163992 Chr.7: 143359876-143359876 on Build GRCh38
rs994185118 Chr.7: 143359733-143359733 on Build GRCh38
rs967132465 Chr.7: 143359871-143359871 on Build GRCh38
rs761440967 Chr.7: 143359693-143359693 on Build GRCh38
rs913493190 Chr.7: 143359890-143359890 on Build GRCh38
rs117550503 Chr.7: 143360053-143360053 on Build GRCh38
rs749052738 Chr.7: 143359788-143359788 on Build GRCh38
rs766640553 Chr.7: 143359693-143359693 on Build GRCh38
rs371590026 Chr.7: 143360049-143360049 on Build GRCh38
rs1055894551 Chr.7: 143359999-143359999 on Build GRCh38
rs891143725 Chr.7: 143359808-143359808 on Build GRCh38
rs768212135 Chr.7: 143359793-143359793 on Build GRCh38
rs550666460 Chr.7: 143360054-143360054 on Build GRCh38
rs199503162 Chr.7: 143359771-143359771 on Build GRCh38
rs937399376 Chr.7: 143359985-143359985 on Build GRCh38
rs766843624 Chr.7: 143360023-143360023 on Build GRCh38
rs1045930660 Chr.7: 143359896-143359896 on Build GRCh38
rs948255639 Chr.7: 143359680-143359680 on Build GRCh38
rs946325867 Chr.7: 143359891-143359891 on Build GRCh38
rs948275072 Chr.7: 143359981-143359981 on Build GRCh38
rs41277419 Chr.7: 143359809-143359809 on Build GRCh38
rs938374683 Chr.7: 143359900-143359900 on Build GRCh38
rs201214299 Chr.7: 143360061-143360061 on Build GRCh38
rs200470899 Chr.7: 143359695-143359695 on Build GRCh38
rs372264099 Chr.7: 143359699-143359699 on Build GRCh38
rs759866104 Chr.7: 143360118-143360118 on Build GRCh38
rs766911168 Chr.7: 143359727-143359739 on Build GRCh38
rs188003538 Chr.7: 143359856-143359856 on Build GRCh38
rs749070585 Chr.7: 143360067-143360067 on Build GRCh38
rs771689749 Chr.7: 143359804-143359804 on Build GRCh38
rs756015911 Chr.7: 143360058-143360058 on Build GRCh38
rs759483131 Chr.7: 143359740-143359740 on Build GRCh38
rs367894660 Chr.7: 143359715-143359715 on Build GRCh38
rs74625193 Chr.7: 143359811-143359811 on Build GRCh38
rs774108976 Chr.7: 143359795-143359795 on Build GRCh38
rs200492257 Chr.7: 143360105-143360105 on Build GRCh38
rs141754890 Chr.7: 143359738-143359738 on Build GRCh38
rs990768893 Chr.7: 143359857-143359857 on Build GRCh38
rs746471383 Chr.7: 143360106-143360106 on Build GRCh38
rs765417027 Chr.7: 143360030-143360030 on Build GRCh38
rs977551105 Chr.7: 143359888-143359888 on Build GRCh38
rs150742474 Chr.7: 143359741-143359741 on Build GRCh38
rs139118461 Chr.7: 143360069-143360069 on Build GRCh38
rs577541783 Chr.7: 143360117-143360117 on Build GRCh38
rs1021434026 Chr.7: 143359869-143359869 on Build GRCh38
rs778760642 Chr.7: 143360084-143360084 on Build GRCh38
rs886341286 Chr.7: 143360070-143360070 on Build GRCh38

View Full Product Details