Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38510977-38511478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf86
Entrez Gene ID
389649
Gene Name
chromosome 8 open reading frame 86
Gene Aliases
-
UniGene
Hs.546586
Gene Chromosome Location
Chr.8: 38510834-38528662 on Build GRCh38
Transcripts
NM_001303531.1 NM_207412.2

SNP Detail

SNP ID Chromosome Location
rs898429103 Chr.8: 38511083-38511083 on Build GRCh38
rs1035594889 Chr.8: 38511184-38511184 on Build GRCh38
rs1024021921 Chr.8: 38511381-38511381 on Build GRCh38
rs1005002015 Chr.8: 38511140-38511140 on Build GRCh38
rs1039560536 Chr.8: 38511068-38511068 on Build GRCh38
rs187003967 Chr.8: 38511049-38511049 on Build GRCh38
rs886480090 Chr.8: 38511392-38511392 on Build GRCh38
rs891002275 Chr.8: 38511064-38511064 on Build GRCh38
rs971059098 Chr.8: 38511180-38511180 on Build GRCh38
rs945540922 Chr.8: 38511402-38511402 on Build GRCh38
rs1006679879 Chr.8: 38511065-38511065 on Build GRCh38
rs1009646758 Chr.8: 38511037-38511037 on Build GRCh38
rs926977034 Chr.8: 38511395-38511395 on Build GRCh38
rs190038679 Chr.8: 38511371-38511371 on Build GRCh38
rs968313593 Chr.8: 38511236-38511236 on Build GRCh38
rs328298 Chr.8: 38511188-38511188 on Build GRCh38
rs1027678534 Chr.8: 38511097-38511097 on Build GRCh38
rs1012402389 Chr.8: 38511448-38511448 on Build GRCh38
rs549966403 Chr.8: 38511235-38511235 on Build GRCh38
rs527516526 Chr.8: 38511000-38511000 on Build GRCh38
rs377019575 Chr.8: 38511096-38511096 on Build GRCh38
rs755149380 Chr.8: 38511366-38511366 on Build GRCh38
rs577034203 Chr.8: 38511085-38511085 on Build GRCh38
rs193222709 Chr.8: 38511398-38511398 on Build GRCh38
rs779155160 Chr.8: 38511376-38511376 on Build GRCh38
rs1042543503 Chr.8: 38511434-38511434 on Build GRCh38
rs1042440091 Chr.8: 38511040-38511040 on Build GRCh38
rs935074980 Chr.8: 38511293-38511293 on Build GRCh38
rs80038579 Chr.8: 38511234-38511234 on Build GRCh38
rs1050826095 Chr.8: 38511312-38511312 on Build GRCh38
rs564923822 Chr.8: 38511001-38511001 on Build GRCh38
rs138312973 Chr.8: 38511116-38511116 on Build GRCh38
rs911980456 Chr.8: 38511444-38511444 on Build GRCh38
rs1042513525 Chr.8: 38511281-38511281 on Build GRCh38
rs935126565 Chr.8: 38511059-38511059 on Build GRCh38
rs960799691 Chr.8: 38511431-38511431 on Build GRCh38
rs915472274 Chr.8: 38511399-38511399 on Build GRCh38
rs769259345 Chr.8: 38511024-38511024 on Build GRCh38
rs184274702 Chr.8: 38511178-38511178 on Build GRCh38
rs774943995 Chr.8: 38511063-38511063 on Build GRCh38
rs538764843 Chr.8: 38511372-38511372 on Build GRCh38
rs58674983 Chr.8: 38511215-38511216 on Build GRCh38
rs900456445 Chr.8: 38511316-38511316 on Build GRCh38
rs865903467 Chr.8: 38511069-38511069 on Build GRCh38
rs759091955 Chr.8: 38511004-38511004 on Build GRCh38
rs754091374 Chr.8: 38511240-38511240 on Build GRCh38
rs566096876 Chr.8: 38511441-38511441 on Build GRCh38
rs768013041 Chr.8: 38511086-38511086 on Build GRCh38
rs34503346 Chr.8: 38511211-38511211 on Build GRCh38
rs1049523781 Chr.8: 38511391-38511391 on Build GRCh38
rs191791526 Chr.8: 38511159-38511159 on Build GRCh38
rs926877291 Chr.8: 38511192-38511192 on Build GRCh38
rs923667042 Chr.8: 38511245-38511245 on Build GRCh38
rs912307630 Chr.8: 38511358-38511358 on Build GRCh38
rs1009319239 Chr.8: 38511311-38511311 on Build GRCh38
rs1035083211 Chr.8: 38511405-38511405 on Build GRCh38
rs890515465 Chr.8: 38511307-38511307 on Build GRCh38

View Full Product Details