Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42541838-42542370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC20A2
Entrez Gene ID
6575
Gene Name
solute carrier family 20 member 2
Gene Aliases
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
UniGene
Hs.653173
Gene Chromosome Location
Chr.8: 42416462-42542213 on Build GRCh38
Transcripts
NM_006749.4 XM_017013748.1 XM_017013752.1 NM_001135674.1 NM_001135675.1 NM_001135676.1 NM_138436.3 XM_005273398.4

Gene Symbol
SMIM19
Entrez Gene ID
114926
Gene Name
small integral membrane protein 19
Gene Aliases
C8orf40
UniGene
Hs.655320
Gene Chromosome Location
Chr.8: 42541155-42552997 on Build GRCh38
Transcripts
NM_006749.4 XM_017013748.1 XM_017013752.1 NM_001135674.1 NM_001135675.1 NM_001135676.1 NM_138436.3 XM_005273398.4

SNP Detail

SNP ID Chromosome Location
rs955133108 Chr.8: 42542159-42542159 on Build GRCh38
rs899706753 Chr.8: 42541910-42541910 on Build GRCh38
rs1024210172 Chr.8: 42542234-42542234 on Build GRCh38
rs959262734 Chr.8: 42541866-42541866 on Build GRCh38
rs1013462864 Chr.8: 42542086-42542086 on Build GRCh38
rs574357175 Chr.8: 42541945-42541945 on Build GRCh38
rs956826050 Chr.8: 42542025-42542025 on Build GRCh38
rs775060562 Chr.8: 42542207-42542207 on Build GRCh38
rs1052242649 Chr.8: 42541871-42541871 on Build GRCh38
rs987710938 Chr.8: 42542176-42542176 on Build GRCh38
rs1035017891 Chr.8: 42542257-42542257 on Build GRCh38
rs573343900 Chr.8: 42541937-42541937 on Build GRCh38
rs1023842038 Chr.8: 42541984-42541984 on Build GRCh38
rs879904962 Chr.8: 42541971-42541971 on Build GRCh38
rs531329456 Chr.8: 42542103-42542103 on Build GRCh38
rs1033627842 Chr.8: 42542034-42542034 on Build GRCh38
rs978114569 Chr.8: 42541925-42541925 on Build GRCh38
rs946571967 Chr.8: 42542093-42542093 on Build GRCh38
rs368027645 Chr.8: 42541970-42541970 on Build GRCh38
rs927950282 Chr.8: 42542023-42542023 on Build GRCh38
rs145711927 Chr.8: 42542148-42542148 on Build GRCh38
rs939777877 Chr.8: 42541969-42541969 on Build GRCh38
rs967198190 Chr.8: 42541914-42541914 on Build GRCh38
rs908422046 Chr.8: 42542347-42542347 on Build GRCh38
rs1033577131 Chr.8: 42541862-42541862 on Build GRCh38
rs895051186 Chr.8: 42542085-42542085 on Build GRCh38
rs1054796499 Chr.8: 42542256-42542257 on Build GRCh38
rs922175106 Chr.8: 42541946-42541946 on Build GRCh38
rs184224334 Chr.8: 42542278-42542278 on Build GRCh38
rs1050789806 Chr.8: 42542116-42542116 on Build GRCh38
rs982107185 Chr.8: 42542002-42542002 on Build GRCh38
rs886882710 Chr.8: 42541933-42541933 on Build GRCh38
rs969584653 Chr.8: 42541992-42541992 on Build GRCh38
rs527578924 Chr.8: 42542244-42542244 on Build GRCh38
rs1039631224 Chr.8: 42541909-42541909 on Build GRCh38
rs113509094 Chr.8: 42542104-42542104 on Build GRCh38
rs898133432 Chr.8: 42541857-42541857 on Build GRCh38
rs1015543325 Chr.8: 42541962-42541962 on Build GRCh38
rs995155883 Chr.8: 42542026-42542026 on Build GRCh38
rs2923444 Chr.8: 42541861-42541861 on Build GRCh38
rs886768505 Chr.8: 42542123-42542123 on Build GRCh38
rs922248202 Chr.8: 42542149-42542149 on Build GRCh38
rs956712067 Chr.8: 42542277-42542277 on Build GRCh38
rs974992473 Chr.8: 42541941-42541941 on Build GRCh38
rs1036758945 Chr.8: 42541990-42541990 on Build GRCh38
rs946658000 Chr.8: 42541895-42541895 on Build GRCh38
rs35794418 Chr.8: 42542292-42542292 on Build GRCh38
rs995376633 Chr.8: 42541972-42541972 on Build GRCh38
rs34658275 Chr.8: 42542266-42542267 on Build GRCh38
rs933539478 Chr.8: 42541957-42541957 on Build GRCh38
rs933932369 Chr.8: 42542314-42542314 on Build GRCh38
rs1011611886 Chr.8: 42541980-42541980 on Build GRCh38
rs1003849503 Chr.8: 42542124-42542124 on Build GRCh38
rs1003747467 Chr.8: 42542253-42542253 on Build GRCh38
rs915076906 Chr.8: 42541892-42541892 on Build GRCh38
rs986724862 Chr.8: 42542325-42542325 on Build GRCh38
rs940820621 Chr.8: 42542203-42542203 on Build GRCh38
rs367803643 Chr.8: 42542196-42542196 on Build GRCh38
rs988507266 Chr.8: 42542281-42542281 on Build GRCh38
rs933880530 Chr.8: 42542107-42542107 on Build GRCh38
rs915277072 Chr.8: 42542092-42542092 on Build GRCh38
rs886574926 Chr.8: 42541967-42541967 on Build GRCh38
rs1032373564 Chr.8: 42542114-42542114 on Build GRCh38
rs968506702 Chr.8: 42542285-42542285 on Build GRCh38
rs553351952 Chr.8: 42541931-42541931 on Build GRCh38
rs776238505 Chr.8: 42542310-42542311 on Build GRCh38
rs995030700 Chr.8: 42542215-42542215 on Build GRCh38
rs931003265 Chr.8: 42542249-42542249 on Build GRCh38
rs111499617 Chr.8: 42542284-42542284 on Build GRCh38
rs865827733 Chr.8: 42541997-42541997 on Build GRCh38
rs1023909742 Chr.8: 42541973-42541973 on Build GRCh38
rs975300014 Chr.8: 42542288-42542288 on Build GRCh38
rs996179203 Chr.8: 42542343-42542343 on Build GRCh38
rs1036811319 Chr.8: 42542218-42542218 on Build GRCh38
rs997866933 Chr.8: 42541928-42541928 on Build GRCh38
rs113755761 Chr.8: 42542090-42542090 on Build GRCh38

View Full Product Details