Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42752926-42753419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA6
Entrez Gene ID
8973
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
CHNRA6
UniGene
Hs.103128
Gene Chromosome Location
Chr.8: 42752620-42768786 on Build GRCh38
Transcripts
NM_001199279.1 NM_004198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2961999 c.1374C>A p.Y458* Chr.8: 42753290-42753290 on Build GRCh38
6563355 c.1392C>G p.D464E Chr.8: 42753272-42753272 on Build GRCh38
5905747 c.1368G>A p.W456* Chr.8: 42753296-42753296 on Build GRCh38
3649361 c.1455G>A p.Q485Q Chr.8: 42753209-42753209 on Build GRCh38
750313 c.1431T>C p.F477F Chr.8: 42753233-42753233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770116227 Chr.8: 42753134-42753134 on Build GRCh38
rs202141415 Chr.8: 42753228-42753228 on Build GRCh38
rs200262118 Chr.8: 42752968-42752968 on Build GRCh38
rs933635513 Chr.8: 42753055-42753055 on Build GRCh38
rs867320926 Chr.8: 42753242-42753242 on Build GRCh38
rs776544169 Chr.8: 42753267-42753267 on Build GRCh38
rs747732293 Chr.8: 42753290-42753290 on Build GRCh38
rs201264351 Chr.8: 42752975-42752975 on Build GRCh38
rs201581010 Chr.8: 42753097-42753097 on Build GRCh38
rs200897609 Chr.8: 42753052-42753052 on Build GRCh38
rs770998077 Chr.8: 42753152-42753152 on Build GRCh38
rs199638155 Chr.8: 42753319-42753319 on Build GRCh38
rs913056438 Chr.8: 42753089-42753089 on Build GRCh38
rs988625157 Chr.8: 42753392-42753392 on Build GRCh38
rs773493524 Chr.8: 42753141-42753141 on Build GRCh38
rs986862581 Chr.8: 42753348-42753348 on Build GRCh38
rs201461030 Chr.8: 42753119-42753119 on Build GRCh38
rs994984867 Chr.8: 42753161-42753161 on Build GRCh38
rs987971727 Chr.8: 42752988-42752988 on Build GRCh38
rs746006983 Chr.8: 42753248-42753248 on Build GRCh38
rs764113730 Chr.8: 42753184-42753184 on Build GRCh38
rs1045528030 Chr.8: 42753162-42753162 on Build GRCh38
rs1016980984 Chr.8: 42753007-42753007 on Build GRCh38
rs748652876 Chr.8: 42753220-42753220 on Build GRCh38
rs956633437 Chr.8: 42752981-42752981 on Build GRCh38
rs975298749 Chr.8: 42753305-42753305 on Build GRCh38
rs150660043 Chr.8: 42753211-42753211 on Build GRCh38
rs771212852 Chr.8: 42753245-42753245 on Build GRCh38
rs752140998 Chr.8: 42753337-42753337 on Build GRCh38
rs201269857 Chr.8: 42753156-42753156 on Build GRCh38
rs199543280 Chr.8: 42752995-42752995 on Build GRCh38
rs921043825 Chr.8: 42753026-42753026 on Build GRCh38
rs771945637 Chr.8: 42753328-42753329 on Build GRCh38
rs770198289 Chr.8: 42753229-42753229 on Build GRCh38
rs969362345 Chr.8: 42753013-42753013 on Build GRCh38
rs199674043 Chr.8: 42752958-42752958 on Build GRCh38
rs202223533 Chr.8: 42753173-42753173 on Build GRCh38
rs980873393 Chr.8: 42753033-42753033 on Build GRCh38
rs998336865 Chr.8: 42752984-42752984 on Build GRCh38
rs912141934 Chr.8: 42753082-42753082 on Build GRCh38
rs765118748 Chr.8: 42753194-42753194 on Build GRCh38
rs1036579204 Chr.8: 42753102-42753102 on Build GRCh38
rs371024226 Chr.8: 42753358-42753358 on Build GRCh38
rs74432466 Chr.8: 42753096-42753096 on Build GRCh38
rs747966071 Chr.8: 42753259-42753262 on Build GRCh38
rs1035335767 Chr.8: 42752956-42752956 on Build GRCh38
rs1003560319 Chr.8: 42752947-42752947 on Build GRCh38
rs778121297 Chr.8: 42753234-42753234 on Build GRCh38
rs753804344 Chr.8: 42753185-42753185 on Build GRCh38
rs761856913 Chr.8: 42753293-42753293 on Build GRCh38
rs887327443 Chr.8: 42753001-42753001 on Build GRCh38
rs774385978 Chr.8: 42753246-42753246 on Build GRCh38
rs750186865 Chr.8: 42753196-42753196 on Build GRCh38
rs1022227052 Chr.8: 42753276-42753276 on Build GRCh38
rs200851744 Chr.8: 42753186-42753186 on Build GRCh38
rs554471161 Chr.8: 42752987-42752987 on Build GRCh38
rs866205267 Chr.8: 42753197-42753197 on Build GRCh38
rs758076325 Chr.8: 42753203-42753203 on Build GRCh38
rs1035521297 Chr.8: 42753368-42753368 on Build GRCh38
rs759490347 Chr.8: 42753172-42753172 on Build GRCh38
rs778210923 Chr.8: 42753342-42753342 on Build GRCh38
rs779533211 Chr.8: 42753212-42753212 on Build GRCh38
rs556036982 Chr.8: 42753307-42753307 on Build GRCh38
rs199911712 Chr.8: 42753140-42753140 on Build GRCh38
rs149386752 Chr.8: 42753299-42753299 on Build GRCh38
rs754588393 Chr.8: 42753322-42753322 on Build GRCh38
rs570846573 Chr.8: 42753042-42753042 on Build GRCh38
rs200852305 Chr.8: 42753104-42753104 on Build GRCh38
rs200318794 Chr.8: 42753177-42753177 on Build GRCh38
rs939691447 Chr.8: 42753386-42753386 on Build GRCh38
rs868428586 Chr.8: 42753231-42753231 on Build GRCh38
rs199642142 Chr.8: 42753085-42753085 on Build GRCh38
rs967882674 Chr.8: 42753003-42753003 on Build GRCh38
rs200410758 Chr.8: 42753090-42753090 on Build GRCh38
rs2304297 Chr.8: 42753056-42753056 on Build GRCh38
rs1002180317 Chr.8: 42753338-42753338 on Build GRCh38
rs371523970 Chr.8: 42753199-42753199 on Build GRCh38
rs769162257 Chr.8: 42753367-42753367 on Build GRCh38
rs761738595 Chr.8: 42753193-42753193 on Build GRCh38
rs763145825 Chr.8: 42753148-42753148 on Build GRCh38
rs975531271 Chr.8: 42753019-42753019 on Build GRCh38
rs1035085122 Chr.8: 42753036-42753036 on Build GRCh38
rs377013442 Chr.8: 42753340-42753340 on Build GRCh38
rs55662044 Chr.8: 42753238-42753238 on Build GRCh38
rs201034972 Chr.8: 42753213-42753213 on Build GRCh38
rs781695653 Chr.8: 42753218-42753218 on Build GRCh38
rs1005803871 Chr.8: 42753002-42753002 on Build GRCh38
rs773174628 Chr.8: 42753303-42753303 on Build GRCh38
rs778650621 Chr.8: 42753207-42753207 on Build GRCh38
rs75874295 Chr.8: 42753143-42753143 on Build GRCh38
rs772013814 Chr.8: 42753284-42753284 on Build GRCh38
rs200044094 Chr.8: 42753098-42753098 on Build GRCh38
rs767004102 Chr.8: 42753324-42753324 on Build GRCh38

View Full Product Details