Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42756156-42756666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA6
Entrez Gene ID
8973
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
CHNRA6
UniGene
Hs.103128
Gene Chromosome Location
Chr.8: 42752620-42768786 on Build GRCh38
Transcripts
NM_001199279.1 NM_004198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367357 c.1033T>G p.F345V Chr.8: 42756166-42756166 on Build GRCh38
1197639 c.719C>A p.P240Q Chr.8: 42756480-42756480 on Build GRCh38
354645 c.549A>G p.K183K Chr.8: 42756650-42756650 on Build GRCh38
1457185 c.916T>C p.Y306H Chr.8: 42756283-42756283 on Build GRCh38
6628737 c.894G>T p.L298L Chr.8: 42756305-42756305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767534563 Chr.8: 42756642-42756642 on Build GRCh38
rs185610062 Chr.8: 42756224-42756224 on Build GRCh38
rs200829161 Chr.8: 42756268-42756268 on Build GRCh38
rs773917226 Chr.8: 42756271-42756271 on Build GRCh38
rs778753253 Chr.8: 42756204-42756204 on Build GRCh38
rs200072442 Chr.8: 42756313-42756313 on Build GRCh38
rs779838446 Chr.8: 42756217-42756217 on Build GRCh38
rs775630227 Chr.8: 42756412-42756412 on Build GRCh38
rs766345085 Chr.8: 42756435-42756435 on Build GRCh38
rs780776442 Chr.8: 42756354-42756354 on Build GRCh38
rs368839862 Chr.8: 42756517-42756517 on Build GRCh38
rs142629147 Chr.8: 42756300-42756300 on Build GRCh38
rs201796500 Chr.8: 42756406-42756406 on Build GRCh38
rs754789848 Chr.8: 42756462-42756462 on Build GRCh38
rs199955670 Chr.8: 42756627-42756627 on Build GRCh38
rs199671238 Chr.8: 42756471-42756471 on Build GRCh38
rs775696869 Chr.8: 42756287-42756287 on Build GRCh38
rs200533962 Chr.8: 42756335-42756335 on Build GRCh38
rs760963856 Chr.8: 42756420-42756420 on Build GRCh38
rs200987418 Chr.8: 42756451-42756451 on Build GRCh38
rs769038131 Chr.8: 42756366-42756373 on Build GRCh38
rs778578382 Chr.8: 42756584-42756584 on Build GRCh38
rs199675892 Chr.8: 42756215-42756215 on Build GRCh38
rs373608967 Chr.8: 42756336-42756336 on Build GRCh38
rs773376342 Chr.8: 42756629-42756629 on Build GRCh38
rs922882584 Chr.8: 42756209-42756209 on Build GRCh38
rs574697613 Chr.8: 42756516-42756516 on Build GRCh38
rs769758264 Chr.8: 42756264-42756266 on Build GRCh38
rs771583412 Chr.8: 42756385-42756385 on Build GRCh38
rs199892210 Chr.8: 42756253-42756253 on Build GRCh38
rs199769813 Chr.8: 42756350-42756350 on Build GRCh38
rs544312612 Chr.8: 42756334-42756334 on Build GRCh38
rs201833648 Chr.8: 42756245-42756245 on Build GRCh38
rs146921905 Chr.8: 42756467-42756467 on Build GRCh38
rs747826040 Chr.8: 42756369-42756369 on Build GRCh38
rs201295503 Chr.8: 42756402-42756402 on Build GRCh38
rs769343905 Chr.8: 42756250-42756250 on Build GRCh38
rs982008922 Chr.8: 42756210-42756210 on Build GRCh38
rs769005774 Chr.8: 42756614-42756614 on Build GRCh38
rs199627128 Chr.8: 42756585-42756585 on Build GRCh38
rs763481468 Chr.8: 42756186-42756186 on Build GRCh38
rs369966241 Chr.8: 42756501-42756501 on Build GRCh38
rs143385261 Chr.8: 42756591-42756591 on Build GRCh38
rs372469952 Chr.8: 42756604-42756604 on Build GRCh38
rs200667694 Chr.8: 42756547-42756547 on Build GRCh38
rs772665574 Chr.8: 42756595-42756595 on Build GRCh38
rs745799927 Chr.8: 42756211-42756211 on Build GRCh38
rs763061271 Chr.8: 42756349-42756352 on Build GRCh38
rs370091414 Chr.8: 42756479-42756479 on Build GRCh38
rs373928428 Chr.8: 42756424-42756424 on Build GRCh38
rs201856103 Chr.8: 42756618-42756618 on Build GRCh38
rs144350308 Chr.8: 42756381-42756381 on Build GRCh38
rs761949537 Chr.8: 42756512-42756512 on Build GRCh38
rs372414818 Chr.8: 42756275-42756275 on Build GRCh38
rs746856231 Chr.8: 42756323-42756323 on Build GRCh38
rs200513077 Chr.8: 42756198-42756198 on Build GRCh38
rs753856041 Chr.8: 42756312-42756312 on Build GRCh38
rs56315032 Chr.8: 42756580-42756580 on Build GRCh38
rs150991696 Chr.8: 42756365-42756365 on Build GRCh38
rs753533097 Chr.8: 42756573-42756573 on Build GRCh38
rs78464081 Chr.8: 42756401-42756401 on Build GRCh38
rs776803911 Chr.8: 42756511-42756511 on Build GRCh38
rs910018042 Chr.8: 42756576-42756576 on Build GRCh38
rs373761304 Chr.8: 42756507-42756507 on Build GRCh38
rs746371215 Chr.8: 42756491-42756491 on Build GRCh38
rs140821226 Chr.8: 42756380-42756380 on Build GRCh38
rs199750589 Chr.8: 42756434-42756434 on Build GRCh38
rs750671647 Chr.8: 42756305-42756305 on Build GRCh38
rs776324742 Chr.8: 42756232-42756232 on Build GRCh38
rs774973747 Chr.8: 42756400-42756400 on Build GRCh38
rs767322053 Chr.8: 42756551-42756551 on Build GRCh38
rs200497100 Chr.8: 42756480-42756480 on Build GRCh38
rs41265264 Chr.8: 42756423-42756423 on Build GRCh38
rs138339131 Chr.8: 42756468-42756468 on Build GRCh38
rs989145751 Chr.8: 42756263-42756263 on Build GRCh38
rs763093183 Chr.8: 42756315-42756315 on Build GRCh38
rs201530242 Chr.8: 42756202-42756202 on Build GRCh38
rs368168912 Chr.8: 42756560-42756560 on Build GRCh38
rs769939913 Chr.8: 42756623-42756623 on Build GRCh38
rs751495876 Chr.8: 42756450-42756450 on Build GRCh38
rs976951248 Chr.8: 42756207-42756207 on Build GRCh38
rs199921540 Chr.8: 42756288-42756288 on Build GRCh38
rs373535041 Chr.8: 42756191-42756191 on Build GRCh38
rs751500929 Chr.8: 42756633-42756635 on Build GRCh38
rs141661289 Chr.8: 42756611-42756611 on Build GRCh38
rs762037634 Chr.8: 42756622-42756622 on Build GRCh38
rs762896869 Chr.8: 42756632-42756632 on Build GRCh38
rs767679720 Chr.8: 42756293-42756293 on Build GRCh38
rs199987912 Chr.8: 42756522-42756522 on Build GRCh38
rs748772192 Chr.8: 42756383-42756383 on Build GRCh38
rs201866921 Chr.8: 42756494-42756494 on Build GRCh38
rs376849555 Chr.8: 42756220-42756220 on Build GRCh38
rs888527245 Chr.8: 42756246-42756246 on Build GRCh38
rs752563455 Chr.8: 42756557-42756557 on Build GRCh38
rs200331504 Chr.8: 42756244-42756244 on Build GRCh38
rs371233399 Chr.8: 42756309-42756309 on Build GRCh38
rs141129760 Chr.8: 42756425-42756425 on Build GRCh38
rs201852908 Chr.8: 42756280-42756280 on Build GRCh38
rs545769727 Chr.8: 42756621-42756621 on Build GRCh38
rs760634963 Chr.8: 42756289-42756289 on Build GRCh38
rs534708905 Chr.8: 42756492-42756492 on Build GRCh38
rs758246174 Chr.8: 42756213-42756213 on Build GRCh38
rs759926957 Chr.8: 42756281-42756281 on Build GRCh38
rs375238294 Chr.8: 42756302-42756302 on Build GRCh38
rs894446733 Chr.8: 42756327-42756327 on Build GRCh38
rs775611153 Chr.8: 42756197-42756197 on Build GRCh38
rs35812517 Chr.8: 42756258-42756258 on Build GRCh38
rs199691486 Chr.8: 42756193-42756193 on Build GRCh38
rs201044590 Chr.8: 42756299-42756299 on Build GRCh38
rs758336371 Chr.8: 42756320-42756320 on Build GRCh38
rs202140850 Chr.8: 42756308-42756308 on Build GRCh38

View Full Product Details