Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 668514-669007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH1
Entrez Gene ID
157697
Gene Name
glutamate rich 1
Gene Aliases
HSPC319
UniGene
Hs.655310
Gene Chromosome Location
Chr.8: 614737-731239 on Build GRCh38
Transcripts
NM_001303100.1 NM_207332.2 XM_006716234.3 XM_011534731.2 XM_011534735.2 XM_011534739.2 XM_017013114.1 XM_017013115.1 XM_017013116.1 XM_017013117.1 XM_017013118.1 XM_017013119.1 XM_017013120.1 XM_017013121.1 XM_017013122.1 XM_017013123.1 XM_017013124.1 XM_017013125.1 XM_017013126.1 XM_017013127.1 XM_017013128.1 XM_017013130.1 XM_017013131.1 XM_017013132.1 XM_017013133.1 XM_017013134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776611 c.1258+9C>G p.? Chr.8: 668589-668589 on Build GRCh38
1205638 c.1225G>A p.E409K Chr.8: 668631-668631 on Build GRCh38
7366575 c.1075G>A p.D359N Chr.8: 668781-668781 on Build GRCh38
1205639 c.1198G>A p.D400N Chr.8: 668658-668658 on Build GRCh38
6378608 c.1066G>T p.V356F Chr.8: 668790-668790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774405503 Chr.8: 668763-668763 on Build GRCh38
rs535431658 Chr.8: 668726-668726 on Build GRCh38
rs188645650 Chr.8: 668884-668884 on Build GRCh38
rs760113812 Chr.8: 668779-668779 on Build GRCh38
rs183010078 Chr.8: 668866-668866 on Build GRCh38
rs1013942665 Chr.8: 668898-668898 on Build GRCh38
rs759231041 Chr.8: 668841-668841 on Build GRCh38
rs938433073 Chr.8: 668588-668588 on Build GRCh38
rs200277044 Chr.8: 668675-668675 on Build GRCh38
rs778213357 Chr.8: 668743-668743 on Build GRCh38
rs770370565 Chr.8: 668834-668834 on Build GRCh38
rs181922694 Chr.8: 668541-668541 on Build GRCh38
rs369657391 Chr.8: 668585-668585 on Build GRCh38
rs569132318 Chr.8: 668943-668943 on Build GRCh38
rs758768926 Chr.8: 668740-668740 on Build GRCh38
rs1036973500 Chr.8: 668986-668986 on Build GRCh38
rs756526316 Chr.8: 668556-668556 on Build GRCh38
rs751053946 Chr.8: 668690-668690 on Build GRCh38
rs1024814841 Chr.8: 668552-668552 on Build GRCh38
rs767485567 Chr.8: 668803-668803 on Build GRCh38
rs925913567 Chr.8: 668604-668604 on Build GRCh38
rs762990627 Chr.8: 668682-668682 on Build GRCh38
rs1056005049 Chr.8: 668876-668876 on Build GRCh38
rs748136718 Chr.8: 668770-668770 on Build GRCh38
rs201135225 Chr.8: 668609-668609 on Build GRCh38
rs754349559 Chr.8: 668564-668564 on Build GRCh38
rs551238591 Chr.8: 668673-668673 on Build GRCh38
rs566874199 Chr.8: 668716-668716 on Build GRCh38
rs555321417 Chr.8: 668730-668730 on Build GRCh38
rs186569427 Chr.8: 668543-668543 on Build GRCh38
rs778917977 Chr.8: 668823-668823 on Build GRCh38
rs544295245 Chr.8: 668752-668752 on Build GRCh38
rs954857585 Chr.8: 668659-668659 on Build GRCh38
rs747892835 Chr.8: 668551-668551 on Build GRCh38
rs373196730 Chr.8: 668612-668612 on Build GRCh38
rs771703982 Chr.8: 668773-668773 on Build GRCh38
rs777252710 Chr.8: 668637-668638 on Build GRCh38
rs142624349 Chr.8: 668812-668812 on Build GRCh38
rs776178951 Chr.8: 668796-668797 on Build GRCh38
rs994428674 Chr.8: 668697-668697 on Build GRCh38
rs200683319 Chr.8: 668814-668814 on Build GRCh38
rs772011533 Chr.8: 668665-668665 on Build GRCh38
rs750012573 Chr.8: 668731-668731 on Build GRCh38
rs899312025 Chr.8: 668987-668987 on Build GRCh38
rs183178838 Chr.8: 668959-668959 on Build GRCh38
rs370214725 Chr.8: 668732-668732 on Build GRCh38
rs781002943 Chr.8: 668628-668628 on Build GRCh38
rs765504989 Chr.8: 668733-668733 on Build GRCh38
rs186877818 Chr.8: 668807-668807 on Build GRCh38
rs372275561 Chr.8: 668563-668563 on Build GRCh38
rs376556884 Chr.8: 668753-668753 on Build GRCh38
rs191290346 Chr.8: 668864-668864 on Build GRCh38
rs534063827 Chr.8: 668777-668777 on Build GRCh38
rs1005133210 Chr.8: 668979-668979 on Build GRCh38
rs752083676 Chr.8: 668744-668744 on Build GRCh38
rs770397148 Chr.8: 668793-668794 on Build GRCh38
rs766208164 Chr.8: 668537-668537 on Build GRCh38
rs780608053 Chr.8: 668560-668560 on Build GRCh38
rs755401025 Chr.8: 668644-668644 on Build GRCh38
rs749597075 Chr.8: 668637-668637 on Build GRCh38
rs373241506 Chr.8: 668738-668738 on Build GRCh38
rs746551282 Chr.8: 668742-668743 on Build GRCh38
rs112423539 Chr.8: 668907-668907 on Build GRCh38
rs148345133 Chr.8: 668674-668674 on Build GRCh38
rs775833966 Chr.8: 668561-668561 on Build GRCh38
rs568864553 Chr.8: 668982-668982 on Build GRCh38
rs73670365 Chr.8: 668932-668932 on Build GRCh38
rs144057545 Chr.8: 668626-668626 on Build GRCh38
rs754978347 Chr.8: 668542-668542 on Build GRCh38
rs900801691 Chr.8: 668630-668630 on Build GRCh38
rs1053296030 Chr.8: 668597-668597 on Build GRCh38
rs780141712 Chr.8: 668760-668760 on Build GRCh38
rs747221474 Chr.8: 668667-668667 on Build GRCh38
rs1028268974 Chr.8: 668983-668984 on Build GRCh38
rs563131911 Chr.8: 668546-668546 on Build GRCh38
rs891541222 Chr.8: 668735-668735 on Build GRCh38
rs755275460 Chr.8: 668698-668698 on Build GRCh38
rs776471080 Chr.8: 668589-668589 on Build GRCh38
rs757935034 Chr.8: 668545-668545 on Build GRCh38
rs192980727 Chr.8: 668888-668888 on Build GRCh38
rs763255357 Chr.8: 668796-668796 on Build GRCh38
rs765991975 Chr.8: 668781-668781 on Build GRCh38
rs528363820 Chr.8: 668839-668839 on Build GRCh38
rs752034726 Chr.8: 668799-668799 on Build GRCh38
rs146050477 Chr.8: 668776-668776 on Build GRCh38
rs890681048 Chr.8: 668811-668811 on Build GRCh38
rs890607678 Chr.8: 668662-668662 on Build GRCh38
rs763695667 Chr.8: 668689-668689 on Build GRCh38
rs532483269 Chr.8: 668549-668549 on Build GRCh38
rs748083763 Chr.8: 668826-668826 on Build GRCh38
rs375281433 Chr.8: 668584-668584 on Build GRCh38
rs555387008 Chr.8: 668980-668980 on Build GRCh38
rs957279271 Chr.8: 668754-668754 on Build GRCh38
rs771857193 Chr.8: 668624-668624 on Build GRCh38
rs139649568 Chr.8: 668751-668751 on Build GRCh38
rs147752635 Chr.8: 668701-668701 on Build GRCh38
rs201878687 Chr.8: 668568-668568 on Build GRCh38
rs996754712 Chr.8: 668971-668971 on Build GRCh38
rs750107900 Chr.8: 668694-668694 on Build GRCh38
rs377202241 Chr.8: 668668-668668 on Build GRCh38
rs778539169 Chr.8: 668913-668913 on Build GRCh38
rs749566717 Chr.8: 668816-668816 on Build GRCh38
rs187857176 Chr.8: 668960-668960 on Build GRCh38
rs144294718 Chr.8: 668756-668756 on Build GRCh38
rs760410283 Chr.8: 668536-668536 on Build GRCh38
rs989089242 Chr.8: 668889-668889 on Build GRCh38
rs775272180 Chr.8: 668680-668680 on Build GRCh38
rs746154675 Chr.8: 668719-668719 on Build GRCh38
rs772210714 Chr.8: 668573-668573 on Build GRCh38
rs763058845 Chr.8: 668590-668590 on Build GRCh38
rs776180035 Chr.8: 668793-668793 on Build GRCh38
rs768590634 Chr.8: 668761-668761 on Build GRCh38
rs779238507 Chr.8: 668646-668646 on Build GRCh38
rs1703879 Chr.8: 668647-668647 on Build GRCh38
rs982078087 Chr.8: 668869-668869 on Build GRCh38
rs371144455 Chr.8: 668827-668827 on Build GRCh38
rs2271263 Chr.8: 668728-668728 on Build GRCh38
rs752715054 Chr.8: 668615-668615 on Build GRCh38
rs890712250 Chr.8: 668976-668976 on Build GRCh38
rs1042795374 Chr.8: 668920-668920 on Build GRCh38
rs763709216 Chr.8: 668617-668617 on Build GRCh38
rs751227736 Chr.8: 668619-668619 on Build GRCh38
rs769448521 Chr.8: 668725-668725 on Build GRCh38
rs761409952 Chr.8: 668691-668691 on Build GRCh38
rs774925861 Chr.8: 668567-668567 on Build GRCh38
rs973498819 Chr.8: 668862-668862 on Build GRCh38
rs764492648 Chr.8: 668797-668797 on Build GRCh38
rs547294322 Chr.8: 668937-668937 on Build GRCh38
rs764305855 Chr.8: 668649-668649 on Build GRCh38
rs999057575 Chr.8: 668929-668929 on Build GRCh38
rs570956180 Chr.8: 668692-668692 on Build GRCh38
rs757831528 Chr.8: 668802-668802 on Build GRCh38
rs1036540814 Chr.8: 668840-668840 on Build GRCh38
rs760106102 Chr.8: 668825-668830 on Build GRCh38
rs879210864 Chr.8: 668799-668800 on Build GRCh38
rs757797804 Chr.8: 668616-668616 on Build GRCh38
rs141539093 Chr.8: 668700-668700 on Build GRCh38
rs773030046 Chr.8: 668829-668829 on Build GRCh38
rs368591089 Chr.8: 668822-668822 on Build GRCh38

View Full Product Details