Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 2938415-2938928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
UniGene
Hs.571466
Gene Chromosome Location
Chr.8: 2935353-4995035 on Build GRCh38
Transcripts
NM_033225.5 XM_011534752.2 XM_011534753.2 XM_011534754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2788368 c.9763G>A p.E3255K Chr.8: 2938704-2938704 on Build GRCh38
3899358 c.10682G>T p.C3561F Chr.8: 2938598-2938598 on Build GRCh38
5358920 c.9794A>G p.N3265S Chr.8: 2938673-2938673 on Build GRCh38
6044156 c.9803A>G p.Y3268C Chr.8: 2938664-2938664 on Build GRCh38
4804967 c.9782C>A p.A3261E Chr.8: 2938685-2938685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986423269 Chr.8: 2938454-2938454 on Build GRCh38
rs544247592 Chr.8: 2938719-2938719 on Build GRCh38
rs889709492 Chr.8: 2938482-2938482 on Build GRCh38
rs866552505 Chr.8: 2938660-2938660 on Build GRCh38
rs777923555 Chr.8: 2938722-2938722 on Build GRCh38
rs769537827 Chr.8: 2938744-2938744 on Build GRCh38
rs1057079455 Chr.8: 2938846-2938846 on Build GRCh38
rs532409447 Chr.8: 2938625-2938625 on Build GRCh38
rs557663092 Chr.8: 2938453-2938453 on Build GRCh38
rs585000 Chr.8: 2938755-2938755 on Build GRCh38
rs972853848 Chr.8: 2938768-2938768 on Build GRCh38
rs1023591927 Chr.8: 2938885-2938885 on Build GRCh38
rs748022675 Chr.8: 2938720-2938720 on Build GRCh38
rs546070791 Chr.8: 2938497-2938497 on Build GRCh38
rs776408855 Chr.8: 2938584-2938584 on Build GRCh38
rs1006384912 Chr.8: 2938876-2938876 on Build GRCh38
rs754738855 Chr.8: 2938636-2938636 on Build GRCh38
rs537835225 Chr.8: 2938872-2938872 on Build GRCh38
rs772083950 Chr.8: 2938653-2938653 on Build GRCh38
rs573209145 Chr.8: 2938664-2938664 on Build GRCh38
rs549080854 Chr.8: 2938828-2938828 on Build GRCh38
rs760229655 Chr.8: 2938573-2938573 on Build GRCh38
rs768439391 Chr.8: 2938537-2938537 on Build GRCh38
rs117539271 Chr.8: 2938690-2938690 on Build GRCh38
rs754708627 Chr.8: 2938735-2938735 on Build GRCh38
rs766473193 Chr.8: 2938783-2938783 on Build GRCh38
rs1044669383 Chr.8: 2938529-2938529 on Build GRCh38
rs183364287 Chr.8: 2938875-2938875 on Build GRCh38
rs758114301 Chr.8: 2938721-2938721 on Build GRCh38
rs780763350 Chr.8: 2938602-2938602 on Build GRCh38
rs774156708 Chr.8: 2938540-2938540 on Build GRCh38
rs756176284 Chr.8: 2938603-2938603 on Build GRCh38
rs748057437 Chr.8: 2938541-2938541 on Build GRCh38
rs764461319 Chr.8: 2938590-2938590 on Build GRCh38
rs1026377174 Chr.8: 2938611-2938611 on Build GRCh38
rs758828471 Chr.8: 2938442-2938442 on Build GRCh38
rs912271076 Chr.8: 2938463-2938463 on Build GRCh38
rs190750143 Chr.8: 2938822-2938822 on Build GRCh38
rs1003152225 Chr.8: 2938895-2938895 on Build GRCh38
rs542330902 Chr.8: 2938672-2938672 on Build GRCh38
rs766383996 Chr.8: 2938698-2938698 on Build GRCh38
rs917884530 Chr.8: 2938848-2938848 on Build GRCh38
rs746875853 Chr.8: 2938726-2938726 on Build GRCh38
rs372725413 Chr.8: 2938600-2938600 on Build GRCh38
rs766182595 Chr.8: 2938583-2938583 on Build GRCh38
rs530793092 Chr.8: 2938713-2938713 on Build GRCh38
rs748064544 Chr.8: 2938644-2938644 on Build GRCh38
rs1001379845 Chr.8: 2938899-2938899 on Build GRCh38
rs772712865 Chr.8: 2938492-2938492 on Build GRCh38
rs770588010 Chr.8: 2938741-2938741 on Build GRCh38
rs568999814 Chr.8: 2938857-2938857 on Build GRCh38
rs376991095 Chr.8: 2938476-2938476 on Build GRCh38
rs564031277 Chr.8: 2938742-2938742 on Build GRCh38
rs762064948 Chr.8: 2938689-2938689 on Build GRCh38
rs764398530 Chr.8: 2938682-2938682 on Build GRCh38
rs577656509 Chr.8: 2938459-2938459 on Build GRCh38
rs931161969 Chr.8: 2938525-2938525 on Build GRCh38
rs546434087 Chr.8: 2938793-2938793 on Build GRCh38
rs770561266 Chr.8: 2938668-2938668 on Build GRCh38
rs372134458 Chr.8: 2938750-2938750 on Build GRCh38
rs1040334731 Chr.8: 2938847-2938847 on Build GRCh38
rs778969760 Chr.8: 2938715-2938715 on Build GRCh38
rs865908333 Chr.8: 2938471-2938471 on Build GRCh38
rs1023180117 Chr.8: 2938647-2938647 on Build GRCh38
rs757743271 Chr.8: 2938592-2938592 on Build GRCh38
rs901923447 Chr.8: 2938513-2938513 on Build GRCh38
rs979943875 Chr.8: 2938806-2938806 on Build GRCh38
rs776437341 Chr.8: 2938669-2938669 on Build GRCh38
rs756194734 Chr.8: 2938693-2938693 on Build GRCh38
rs774821554 Chr.8: 2938685-2938685 on Build GRCh38
rs773259880 Chr.8: 2938663-2938663 on Build GRCh38
rs368826091 Chr.8: 2938587-2938587 on Build GRCh38
rs919728812 Chr.8: 2938504-2938504 on Build GRCh38
rs1021056682 Chr.8: 2938681-2938681 on Build GRCh38
rs993567016 Chr.8: 2938530-2938530 on Build GRCh38
rs1007520626 Chr.8: 2938483-2938483 on Build GRCh38
rs938573551 Chr.8: 2938826-2938826 on Build GRCh38
rs778135901 Chr.8: 2938898-2938898 on Build GRCh38
rs771502494 Chr.8: 2938557-2938557 on Build GRCh38
rs35310352 Chr.8: 2938633-2938633 on Build GRCh38
rs762355279 Chr.8: 2938553-2938553 on Build GRCh38
rs750929251 Chr.8: 2938692-2938692 on Build GRCh38
rs185597376 Chr.8: 2938797-2938797 on Build GRCh38
rs772735444 Chr.8: 2938559-2938559 on Build GRCh38
rs534392361 Chr.8: 2938833-2938839 on Build GRCh38
rs751225742 Chr.8: 2938699-2938699 on Build GRCh38
rs931086117 Chr.8: 2938802-2938802 on Build GRCh38
rs797021984 Chr.8: 2938870-2938870 on Build GRCh38
rs761117702 Chr.8: 2938767-2938767 on Build GRCh38
rs970325093 Chr.8: 2938888-2938888 on Build GRCh38
rs373703291 Chr.8: 2938860-2938860 on Build GRCh38
rs376675870 Chr.8: 2938631-2938631 on Build GRCh38
rs773726523 Chr.8: 2938764-2938764 on Build GRCh38
rs1049636720 Chr.8: 2938473-2938473 on Build GRCh38

View Full Product Details