Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 2942381-2942877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
UniGene
Hs.571466
Gene Chromosome Location
Chr.8: 2935353-4995035 on Build GRCh38
Transcripts
NM_033225.5 XM_011534752.2 XM_011534753.2 XM_011534754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2788395 c.9680C>T p.A3227V Chr.8: 2942514-2942514 on Build GRCh38
2788397 c.9654G>A p.A3218A Chr.8: 2942540-2942540 on Build GRCh38
3925114 c.9670C>T p.P3224S Chr.8: 2942524-2942524 on Build GRCh38
1098852 c.10488C>A p.F3496L Chr.8: 2942519-2942519 on Build GRCh38
6496037 c.10414C>T p.P3472S Chr.8: 2942593-2942593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026855782 Chr.8: 2942543-2942543 on Build GRCh38
rs763101468 Chr.8: 2942587-2942587 on Build GRCh38
rs377332641 Chr.8: 2942497-2942497 on Build GRCh38
rs1022711469 Chr.8: 2942684-2942684 on Build GRCh38
rs540013752 Chr.8: 2942748-2942748 on Build GRCh38
rs745443792 Chr.8: 2942449-2942449 on Build GRCh38
rs757302349 Chr.8: 2942442-2942442 on Build GRCh38
rs889429236 Chr.8: 2942735-2942735 on Build GRCh38
rs564251139 Chr.8: 2942593-2942593 on Build GRCh38
rs755659271 Chr.8: 2942541-2942541 on Build GRCh38
rs994126075 Chr.8: 2942617-2942617 on Build GRCh38
rs539673879 Chr.8: 2942456-2942456 on Build GRCh38
rs1005875445 Chr.8: 2942836-2942836 on Build GRCh38
rs866647445 Chr.8: 2942518-2942518 on Build GRCh38
rs759726370 Chr.8: 2942498-2942498 on Build GRCh38
rs888061312 Chr.8: 2942822-2942825 on Build GRCh38
rs765162750 Chr.8: 2942501-2942501 on Build GRCh38
rs938072289 Chr.8: 2942421-2942421 on Build GRCh38
rs778517811 Chr.8: 2942564-2942564 on Build GRCh38
rs11984691 Chr.8: 2942574-2942574 on Build GRCh38
rs372313030 Chr.8: 2942594-2942594 on Build GRCh38
rs769947864 Chr.8: 2942585-2942585 on Build GRCh38
rs980018190 Chr.8: 2942557-2942557 on Build GRCh38
rs546516177 Chr.8: 2942616-2942616 on Build GRCh38
rs755968988 Chr.8: 2942611-2942611 on Build GRCh38
rs753341631 Chr.8: 2942626-2942626 on Build GRCh38
rs892435078 Chr.8: 2942508-2942508 on Build GRCh38
rs78595521 Chr.8: 2942538-2942538 on Build GRCh38
rs6997981 Chr.8: 2942459-2942459 on Build GRCh38
rs566597823 Chr.8: 2942797-2942797 on Build GRCh38
rs917978023 Chr.8: 2942641-2942641 on Build GRCh38
rs758217214 Chr.8: 2942427-2942427 on Build GRCh38
rs560246209 Chr.8: 2942628-2942628 on Build GRCh38
rs1019795044 Chr.8: 2942533-2942533 on Build GRCh38
rs561767929 Chr.8: 2942527-2942527 on Build GRCh38
rs556741428 Chr.8: 2942779-2942779 on Build GRCh38
rs571104487 Chr.8: 2942707-2942707 on Build GRCh38
rs773522169 Chr.8: 2942480-2942480 on Build GRCh38
rs537479601 Chr.8: 2942698-2942698 on Build GRCh38
rs185027947 Chr.8: 2942563-2942563 on Build GRCh38
rs764381003 Chr.8: 2942516-2942516 on Build GRCh38
rs777662035 Chr.8: 2942436-2942436 on Build GRCh38
rs568888850 Chr.8: 2942688-2942688 on Build GRCh38
rs775472545 Chr.8: 2942586-2942586 on Build GRCh38
rs1026979551 Chr.8: 2942821-2942821 on Build GRCh38
rs978147442 Chr.8: 2942826-2942826 on Build GRCh38
rs189406580 Chr.8: 2942800-2942800 on Build GRCh38
rs781348126 Chr.8: 2942528-2942528 on Build GRCh38
rs370039183 Chr.8: 2942651-2942651 on Build GRCh38
rs928432661 Chr.8: 2942692-2942692 on Build GRCh38
rs889449578 Chr.8: 2942504-2942504 on Build GRCh38
rs181030709 Chr.8: 2942801-2942801 on Build GRCh38
rs926619920 Chr.8: 2942402-2942402 on Build GRCh38
rs1034670080 Chr.8: 2942792-2942792 on Build GRCh38
rs1019560333 Chr.8: 2942608-2942608 on Build GRCh38
rs575211339 Chr.8: 2942803-2942803 on Build GRCh38
rs765551657 Chr.8: 2942424-2942424 on Build GRCh38
rs1026781662 Chr.8: 2942776-2942776 on Build GRCh38
rs369998371 Chr.8: 2942505-2942505 on Build GRCh38
rs541781609 Chr.8: 2942510-2942510 on Build GRCh38
rs367589721 Chr.8: 2942538-2942539 on Build GRCh38
rs764419894 Chr.8: 2942777-2942777 on Build GRCh38
rs760993113 Chr.8: 2942486-2942486 on Build GRCh38
rs374886190 Chr.8: 2942422-2942422 on Build GRCh38
rs375524577 Chr.8: 2942602-2942602 on Build GRCh38
rs767156556 Chr.8: 2942601-2942601 on Build GRCh38
rs369815085 Chr.8: 2942457-2942457 on Build GRCh38
rs751579729 Chr.8: 2942438-2942438 on Build GRCh38
rs778605665 Chr.8: 2942635-2942635 on Build GRCh38
rs749005777 Chr.8: 2942549-2942549 on Build GRCh38
rs1019298913 Chr.8: 2942747-2942747 on Build GRCh38
rs951761982 Chr.8: 2942642-2942642 on Build GRCh38
rs914499596 Chr.8: 2942714-2942714 on Build GRCh38
rs764466822 Chr.8: 2942590-2942590 on Build GRCh38
rs760926404 Chr.8: 2942743-2942743 on Build GRCh38
rs34589551 Chr.8: 2942411-2942411 on Build GRCh38
rs762213315 Chr.8: 2942825-2942826 on Build GRCh38
rs936854583 Chr.8: 2942830-2942830 on Build GRCh38
rs772444063 Chr.8: 2942476-2942476 on Build GRCh38
rs1053062544 Chr.8: 2942472-2942472 on Build GRCh38
rs751450586 Chr.8: 2942522-2942522 on Build GRCh38
rs904036336 Chr.8: 2942685-2942685 on Build GRCh38
rs925429180 Chr.8: 2942827-2942827 on Build GRCh38
rs946716196 Chr.8: 2942744-2942744 on Build GRCh38
rs752857280 Chr.8: 2942425-2942425 on Build GRCh38
rs1042523660 Chr.8: 2942662-2942662 on Build GRCh38
rs746303001 Chr.8: 2942578-2942578 on Build GRCh38
rs889238555 Chr.8: 2942565-2942565 on Build GRCh38
rs551235452 Chr.8: 2942702-2942702 on Build GRCh38
rs529003948 Chr.8: 2942640-2942640 on Build GRCh38
rs966729243 Chr.8: 2942816-2942816 on Build GRCh38
rs750448282 Chr.8: 2942440-2942441 on Build GRCh38
rs775737022 Chr.8: 2942667-2942667 on Build GRCh38
rs553301331 Chr.8: 2942766-2942766 on Build GRCh38
rs993927160 Chr.8: 2942768-2942768 on Build GRCh38
rs917957553 Chr.8: 2942806-2942806 on Build GRCh38
rs548974330 Chr.8: 2942654-2942654 on Build GRCh38
rs1043740829 Chr.8: 2942752-2942752 on Build GRCh38
rs373366226 Chr.8: 2942540-2942540 on Build GRCh38

View Full Product Details