Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 2949201-2949743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
UniGene
Hs.571466
Gene Chromosome Location
Chr.8: 2935353-4995035 on Build GRCh38
Transcripts
NM_033225.5 XM_011534752.2 XM_011534753.2 XM_011534754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3647924 c.9559G>A p.D3187N Chr.8: 2949329-2949329 on Build GRCh38
4519957 c.10366G>A p.G3456R Chr.8: 2949335-2949335 on Build GRCh38
5874503 c.10350T>G p.F3450L Chr.8: 2949351-2949351 on Build GRCh38
3647922 c.9565G>A p.G3189R Chr.8: 2949323-2949323 on Build GRCh38
5733391 c.9566G>T p.G3189V Chr.8: 2949322-2949322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59743965 Chr.8: 2949679-2949679 on Build GRCh38
rs1027359126 Chr.8: 2949702-2949702 on Build GRCh38
rs369821761 Chr.8: 2949675-2949675 on Build GRCh38
rs772236278 Chr.8: 2949386-2949386 on Build GRCh38
rs368712154 Chr.8: 2949679-2949680 on Build GRCh38
rs558818722 Chr.8: 2949616-2949616 on Build GRCh38
rs994370101 Chr.8: 2949451-2949451 on Build GRCh38
rs766284756 Chr.8: 2949396-2949400 on Build GRCh38
rs568103775 Chr.8: 2949685-2949685 on Build GRCh38
rs199630799 Chr.8: 2949416-2949416 on Build GRCh38
rs754490630 Chr.8: 2949291-2949291 on Build GRCh38
rs764670757 Chr.8: 2949295-2949295 on Build GRCh38
rs752253287 Chr.8: 2949426-2949426 on Build GRCh38
rs938680944 Chr.8: 2949316-2949316 on Build GRCh38
rs201732061 Chr.8: 2949675-2949676 on Build GRCh38
rs752369840 Chr.8: 2949300-2949300 on Build GRCh38
rs890661021 Chr.8: 2949661-2949661 on Build GRCh38
rs774308054 Chr.8: 2949247-2949247 on Build GRCh38
rs1105081 Chr.8: 2949530-2949530 on Build GRCh38
rs1031099653 Chr.8: 2949665-2949665 on Build GRCh38
rs775636267 Chr.8: 2949544-2949545 on Build GRCh38
rs372864008 Chr.8: 2949370-2949370 on Build GRCh38
rs372798740 Chr.8: 2949676-2949676 on Build GRCh38
rs754463929 Chr.8: 2949408-2949409 on Build GRCh38
rs962075493 Chr.8: 2949499-2949499 on Build GRCh38
rs73657526 Chr.8: 2949398-2949398 on Build GRCh38
rs188927899 Chr.8: 2949359-2949359 on Build GRCh38
rs927863846 Chr.8: 2949575-2949575 on Build GRCh38
rs956460978 Chr.8: 2949669-2949669 on Build GRCh38
rs200732352 Chr.8: 2949684-2949684 on Build GRCh38
rs953227964 Chr.8: 2949708-2949708 on Build GRCh38
rs201340269 Chr.8: 2949424-2949424 on Build GRCh38
rs746211932 Chr.8: 2949254-2949254 on Build GRCh38
rs932129914 Chr.8: 2949649-2949649 on Build GRCh38
rs374032948 Chr.8: 2949264-2949264 on Build GRCh38
rs796862142 Chr.8: 2949476-2949476 on Build GRCh38
rs1050621921 Chr.8: 2949655-2949655 on Build GRCh38
rs980614701 Chr.8: 2949566-2949566 on Build GRCh38
rs767372626 Chr.8: 2949280-2949280 on Build GRCh38
rs749751514 Chr.8: 2949364-2949364 on Build GRCh38
rs369475423 Chr.8: 2949656-2949656 on Build GRCh38
rs58688576 Chr.8: 2949680-2949681 on Build GRCh38
rs779091673 Chr.8: 2949371-2949371 on Build GRCh38
rs766288404 Chr.8: 2949288-2949288 on Build GRCh38
rs897484351 Chr.8: 2949686-2949686 on Build GRCh38
rs1014592179 Chr.8: 2949496-2949496 on Build GRCh38
rs143912854 Chr.8: 2949676-2949677 on Build GRCh38
rs970363793 Chr.8: 2949650-2949650 on Build GRCh38
rs267601885 Chr.8: 2949367-2949367 on Build GRCh38
rs543462035 Chr.8: 2949523-2949523 on Build GRCh38
rs993199280 Chr.8: 2949600-2949600 on Build GRCh38
rs777459005 Chr.8: 2949338-2949338 on Build GRCh38
rs758007040 Chr.8: 2949324-2949324 on Build GRCh38
rs970373709 Chr.8: 2949519-2949519 on Build GRCh38
rs1050342572 Chr.8: 2949592-2949592 on Build GRCh38
rs139429901 Chr.8: 2949631-2949631 on Build GRCh38
rs376342606 Chr.8: 2949277-2949277 on Build GRCh38
rs926065409 Chr.8: 2949645-2949645 on Build GRCh38
rs994487646 Chr.8: 2949692-2949692 on Build GRCh38
rs5741961 Chr.8: 2949398-2949399 on Build GRCh38
rs60411696 Chr.8: 2949678-2949679 on Build GRCh38
rs761679519 Chr.8: 2949710-2949710 on Build GRCh38
rs897266214 Chr.8: 2949616-2949616 on Build GRCh38
rs559219001 Chr.8: 2949428-2949428 on Build GRCh38
rs1008282390 Chr.8: 2949440-2949440 on Build GRCh38
rs748212272 Chr.8: 2949379-2949379 on Build GRCh38
rs145997196 Chr.8: 2949411-2949412 on Build GRCh38
rs981827269 Chr.8: 2949270-2949270 on Build GRCh38
rs368841292 Chr.8: 2949402-2949403 on Build GRCh38
rs753773868 Chr.8: 2949290-2949290 on Build GRCh38
rs750088444 Chr.8: 2949407-2949408 on Build GRCh38
rs764758136 Chr.8: 2949425-2949425 on Build GRCh38
rs762511500 Chr.8: 2949427-2949427 on Build GRCh38
rs57461657 Chr.8: 2949415-2949416 on Build GRCh38
rs762953817 Chr.8: 2949373-2949373 on Build GRCh38
rs556809577 Chr.8: 2949249-2949249 on Build GRCh38
rs201743560 Chr.8: 2949548-2949548 on Build GRCh38
rs888614467 Chr.8: 2949629-2949629 on Build GRCh38
rs1041507419 Chr.8: 2949610-2949610 on Build GRCh38
rs1048531369 Chr.8: 2949623-2949623 on Build GRCh38
rs989302227 Chr.8: 2949547-2949547 on Build GRCh38
rs183683450 Chr.8: 2949714-2949714 on Build GRCh38
rs750809802 Chr.8: 2949342-2949342 on Build GRCh38
rs1003240404 Chr.8: 2949652-2949652 on Build GRCh38
rs376344419 Chr.8: 2949547-2949550 on Build GRCh38
rs539190756 Chr.8: 2949395-2949395 on Build GRCh38
rs931902319 Chr.8: 2949330-2949330 on Build GRCh38
rs759264211 Chr.8: 2949612-2949612 on Build GRCh38
rs374385872 Chr.8: 2949284-2949284 on Build GRCh38
rs72288499 Chr.8: 2949545-2949548 on Build GRCh38
rs777251815 Chr.8: 2949399-2949400 on Build GRCh38
rs528299886 Chr.8: 2949637-2949637 on Build GRCh38
rs144666650 Chr.8: 2949501-2949501 on Build GRCh38
rs756445765 Chr.8: 2949432-2949432 on Build GRCh38
rs768925991 Chr.8: 2949416-2949417 on Build GRCh38
rs751155197 Chr.8: 2949429-2949429 on Build GRCh38
rs140807526 Chr.8: 2949443-2949443 on Build GRCh38
rs1008943083 Chr.8: 2949480-2949480 on Build GRCh38
rs897314024 Chr.8: 2949447-2949447 on Build GRCh38
rs556305936 Chr.8: 2949278-2949278 on Build GRCh38
rs773937583 Chr.8: 2949590-2949590 on Build GRCh38
rs944060645 Chr.8: 2949605-2949605 on Build GRCh38
rs150819604 Chr.8: 2949578-2949578 on Build GRCh38
rs1057167365 Chr.8: 2949328-2949328 on Build GRCh38
rs773472772 Chr.8: 2949392-2949392 on Build GRCh38
rs986043612 Chr.8: 2949713-2949713 on Build GRCh38
rs961827178 Chr.8: 2949229-2949229 on Build GRCh38
rs749443879 Chr.8: 2949275-2949275 on Build GRCh38
rs778590309 Chr.8: 2949399-2949403 on Build GRCh38
rs746191318 Chr.8: 2949412-2949413 on Build GRCh38
rs1003822096 Chr.8: 2949522-2949522 on Build GRCh38
rs541490978 Chr.8: 2949477-2949477 on Build GRCh38
rs760689584 Chr.8: 2949395-2949396 on Build GRCh38
rs200448663 Chr.8: 2949417-2949417 on Build GRCh38
rs200675652 Chr.8: 2949423-2949423 on Build GRCh38
rs201123517 Chr.8: 2949677-2949678 on Build GRCh38
rs952362636 Chr.8: 2949238-2949238 on Build GRCh38
rs969177735 Chr.8: 2949532-2949532 on Build GRCh38
rs1023216412 Chr.8: 2949502-2949502 on Build GRCh38
rs368178082 Chr.8: 2949622-2949622 on Build GRCh38
rs368405203 Chr.8: 2949343-2949343 on Build GRCh38

View Full Product Details