Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 2950074-2950585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD1
Entrez Gene ID
64478
Gene Name
CUB and Sushi multiple domains 1
Gene Aliases
PPP1R24
UniGene
Hs.571466
Gene Chromosome Location
Chr.8: 2935353-4995035 on Build GRCh38
Transcripts
NM_033225.5 XM_011534752.2 XM_011534753.2 XM_011534754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834532 c.9411C>T p.A3137A Chr.8: 2950321-2950321 on Build GRCh38
5552086 c.9452A>T p.D3151V Chr.8: 2950280-2950280 on Build GRCh38
3647929 c.10241C>T p.A3414V Chr.8: 2950304-2950304 on Build GRCh38
3647928 c.9498T>A p.G3166G Chr.8: 2950234-2950234 on Build GRCh38
7141494 c.9471C>A p.F3157L Chr.8: 2950261-2950261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367746075 Chr.8: 2950209-2950209 on Build GRCh38
rs769781161 Chr.8: 2950237-2950237 on Build GRCh38
rs751709909 Chr.8: 2950386-2950386 on Build GRCh38
rs867308736 Chr.8: 2950312-2950312 on Build GRCh38
rs1019141070 Chr.8: 2950204-2950204 on Build GRCh38
rs966536646 Chr.8: 2950101-2950101 on Build GRCh38
rs1052684588 Chr.8: 2950158-2950158 on Build GRCh38
rs761482256 Chr.8: 2950263-2950263 on Build GRCh38
rs909905785 Chr.8: 2950118-2950118 on Build GRCh38
rs868547189 Chr.8: 2950295-2950295 on Build GRCh38
rs777861665 Chr.8: 2950307-2950307 on Build GRCh38
rs769510760 Chr.8: 2950330-2950330 on Build GRCh38
rs901829917 Chr.8: 2950478-2950478 on Build GRCh38
rs112827726 Chr.8: 2950456-2950456 on Build GRCh38
rs993272827 Chr.8: 2950477-2950477 on Build GRCh38
rs773947103 Chr.8: 2950354-2950354 on Build GRCh38
rs775049133 Chr.8: 2950440-2950442 on Build GRCh38
rs928063403 Chr.8: 2950304-2950304 on Build GRCh38
rs775400212 Chr.8: 2950249-2950249 on Build GRCh38
rs1019368743 Chr.8: 2950281-2950281 on Build GRCh38
rs567765454 Chr.8: 2950125-2950125 on Build GRCh38
rs568540519 Chr.8: 2950480-2950480 on Build GRCh38
rs917635346 Chr.8: 2950195-2950195 on Build GRCh38
rs371863339 Chr.8: 2950416-2950416 on Build GRCh38
rs984995698 Chr.8: 2950164-2950164 on Build GRCh38
rs961050684 Chr.8: 2950536-2950536 on Build GRCh38
rs959891094 Chr.8: 2950385-2950385 on Build GRCh38
rs993485059 Chr.8: 2950482-2950482 on Build GRCh38
rs1022876404 Chr.8: 2950507-2950507 on Build GRCh38
rs937683029 Chr.8: 2950170-2950170 on Build GRCh38
rs765641761 Chr.8: 2950365-2950365 on Build GRCh38
rs772998900 Chr.8: 2950360-2950360 on Build GRCh38
rs550651115 Chr.8: 2950540-2950540 on Build GRCh38
rs556344550 Chr.8: 2950250-2950250 on Build GRCh38
rs779544527 Chr.8: 2950189-2950189 on Build GRCh38
rs774510042 Chr.8: 2950365-2950366 on Build GRCh38
rs762884219 Chr.8: 2950378-2950378 on Build GRCh38
rs946911902 Chr.8: 2950373-2950373 on Build GRCh38
rs150154982 Chr.8: 2950436-2950436 on Build GRCh38
rs775418337 Chr.8: 2950334-2950334 on Build GRCh38
rs561750597 Chr.8: 2950182-2950182 on Build GRCh38
rs926915646 Chr.8: 2950167-2950167 on Build GRCh38
rs770656370 Chr.8: 2950213-2950213 on Build GRCh38
rs369432148 Chr.8: 2950305-2950305 on Build GRCh38
rs767243472 Chr.8: 2950266-2950266 on Build GRCh38
rs532914026 Chr.8: 2950361-2950361 on Build GRCh38
rs771495665 Chr.8: 2950253-2950253 on Build GRCh38
rs552808140 Chr.8: 2950098-2950098 on Build GRCh38
rs969342340 Chr.8: 2950516-2950516 on Build GRCh38
rs749196788 Chr.8: 2950335-2950335 on Build GRCh38
rs952033511 Chr.8: 2950216-2950216 on Build GRCh38
rs761202576 Chr.8: 2950355-2950355 on Build GRCh38
rs765914897 Chr.8: 2950289-2950289 on Build GRCh38
rs754424518 Chr.8: 2950185-2950185 on Build GRCh38
rs1049672322 Chr.8: 2950256-2950256 on Build GRCh38
rs772191931 Chr.8: 2950200-2950200 on Build GRCh38
rs1042140814 Chr.8: 2950410-2950410 on Build GRCh38
rs755384385 Chr.8: 2950287-2950287 on Build GRCh38
rs762371826 Chr.8: 2950254-2950254 on Build GRCh38
rs764477883 Chr.8: 2950379-2950379 on Build GRCh38
rs889671330 Chr.8: 2950433-2950433 on Build GRCh38
rs753183110 Chr.8: 2950290-2950290 on Build GRCh38
rs1002190567 Chr.8: 2950518-2950518 on Build GRCh38
rs767339734 Chr.8: 2950356-2950356 on Build GRCh38
rs537296322 Chr.8: 2950498-2950498 on Build GRCh38
rs945052939 Chr.8: 2950406-2950406 on Build GRCh38
rs901672364 Chr.8: 2950205-2950205 on Build GRCh38
rs980832061 Chr.8: 2950134-2950134 on Build GRCh38
rs1026347500 Chr.8: 2950486-2950486 on Build GRCh38
rs943053002 Chr.8: 2950420-2950420 on Build GRCh38
rs780707819 Chr.8: 2950451-2950451 on Build GRCh38
rs952136546 Chr.8: 2950489-2950489 on Build GRCh38
rs952269916 Chr.8: 2950157-2950157 on Build GRCh38
rs959621546 Chr.8: 2950519-2950519 on Build GRCh38
rs528796511 Chr.8: 2950427-2950427 on Build GRCh38
rs754307971 Chr.8: 2950274-2950274 on Build GRCh38
rs371486636 Chr.8: 2950260-2950260 on Build GRCh38
rs776372320 Chr.8: 2950218-2950218 on Build GRCh38
rs147971721 Chr.8: 2950201-2950201 on Build GRCh38
rs747226914 Chr.8: 2950309-2950309 on Build GRCh38
rs757540481 Chr.8: 2950321-2950321 on Build GRCh38
rs758899599 Chr.8: 2950376-2950376 on Build GRCh38
rs753168036 Chr.8: 2950371-2950371 on Build GRCh38
rs755579216 Chr.8: 2950187-2950187 on Build GRCh38
rs748300179 Chr.8: 2950190-2950190 on Build GRCh38
rs867822299 Chr.8: 2950292-2950292 on Build GRCh38
rs35043129 Chr.8: 2950261-2950261 on Build GRCh38
rs564208817 Chr.8: 2950329-2950329 on Build GRCh38
rs952196794 Chr.8: 2950366-2950366 on Build GRCh38
rs768080843 Chr.8: 2950353-2950353 on Build GRCh38
rs1049598189 Chr.8: 2950428-2950428 on Build GRCh38
rs984956340 Chr.8: 2950375-2950375 on Build GRCh38
rs991813033 Chr.8: 2950388-2950388 on Build GRCh38
rs759296417 Chr.8: 2950232-2950232 on Build GRCh38
rs781379644 Chr.8: 2950324-2950324 on Build GRCh38
rs1006089595 Chr.8: 2950502-2950502 on Build GRCh38
rs910211884 Chr.8: 2950397-2950397 on Build GRCh38
rs1034279641 Chr.8: 2950513-2950513 on Build GRCh38
rs78507879 Chr.8: 2950156-2950156 on Build GRCh38
rs11987402 Chr.8: 2950396-2950396 on Build GRCh38
rs901615706 Chr.8: 2950447-2950447 on Build GRCh38

View Full Product Details