Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6501898-6502410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT2
Entrez Gene ID
285
Gene Name
angiopoietin 2
Gene Aliases
AGPT2, ANG2
UniGene
Hs.583870
Gene Chromosome Location
Chr.8: 6499651-6563420 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Gene Symbol
MCPH1
Entrez Gene ID
79648
Gene Name
microcephalin 1
Gene Aliases
BRIT1, MCT
UniGene
Hs.593807
Gene Chromosome Location
Chr.8: 6406592-6648505 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

SNP Detail

SNP ID Chromosome Location
rs1000757241 Chr.8: 6502206-6502206 on Build GRCh38
rs958376173 Chr.8: 6502292-6502292 on Build GRCh38
rs2167071 Chr.8: 6501959-6501959 on Build GRCh38
rs969023166 Chr.8: 6502059-6502059 on Build GRCh38
rs921149290 Chr.8: 6502370-6502370 on Build GRCh38
rs1043933356 Chr.8: 6502201-6502201 on Build GRCh38
rs909740921 Chr.8: 6502353-6502354 on Build GRCh38
rs537465583 Chr.8: 6502354-6502354 on Build GRCh38
rs552027714 Chr.8: 6502170-6502170 on Build GRCh38
rs927527068 Chr.8: 6502078-6502078 on Build GRCh38
rs570518261 Chr.8: 6502174-6502174 on Build GRCh38
rs1026610636 Chr.8: 6502356-6502356 on Build GRCh38
rs398047058 Chr.8: 6501941-6501941 on Build GRCh38
rs892282331 Chr.8: 6502222-6502222 on Build GRCh38
rs985366345 Chr.8: 6502101-6502101 on Build GRCh38
rs867572704 Chr.8: 6502140-6502140 on Build GRCh38
rs1030796615 Chr.8: 6502026-6502026 on Build GRCh38
rs1040412925 Chr.8: 6502129-6502129 on Build GRCh38
rs903772618 Chr.8: 6502164-6502164 on Build GRCh38
rs762033126 Chr.8: 6502231-6502231 on Build GRCh38
rs1024714349 Chr.8: 6502255-6502255 on Build GRCh38
rs1029729830 Chr.8: 6501940-6501940 on Build GRCh38
rs796132394 Chr.8: 6502214-6502214 on Build GRCh38
rs892512447 Chr.8: 6502178-6502178 on Build GRCh38
rs192082001 Chr.8: 6502345-6502345 on Build GRCh38
rs565817663 Chr.8: 6501958-6501958 on Build GRCh38
rs958345171 Chr.8: 6502037-6502037 on Build GRCh38
rs931977106 Chr.8: 6502156-6502156 on Build GRCh38
rs187682777 Chr.8: 6502218-6502218 on Build GRCh38
rs775528288 Chr.8: 6502103-6502103 on Build GRCh38
rs952283533 Chr.8: 6502083-6502083 on Build GRCh38
rs951295107 Chr.8: 6502099-6502099 on Build GRCh38
rs1024667285 Chr.8: 6502043-6502043 on Build GRCh38
rs573074166 Chr.8: 6502153-6502153 on Build GRCh38
rs566280020 Chr.8: 6502109-6502109 on Build GRCh38
rs1002272912 Chr.8: 6502314-6502314 on Build GRCh38
rs529318587 Chr.8: 6502021-6502021 on Build GRCh38
rs535137399 Chr.8: 6502300-6502300 on Build GRCh38
rs980407241 Chr.8: 6502061-6502061 on Build GRCh38
rs982927688 Chr.8: 6502353-6502353 on Build GRCh38
rs940792343 Chr.8: 6502106-6502106 on Build GRCh38
rs1010062730 Chr.8: 6502189-6502189 on Build GRCh38
rs1013837911 Chr.8: 6502297-6502297 on Build GRCh38
rs7835591 Chr.8: 6502038-6502038 on Build GRCh38
rs903719937 Chr.8: 6502019-6502019 on Build GRCh38
rs1024391507 Chr.8: 6502305-6502306 on Build GRCh38
rs956453354 Chr.8: 6502032-6502032 on Build GRCh38
rs569610397 Chr.8: 6502274-6502274 on Build GRCh38
rs765074784 Chr.8: 6502262-6502262 on Build GRCh38
rs1022972011 Chr.8: 6501968-6501968 on Build GRCh38
rs952404473 Chr.8: 6502363-6502363 on Build GRCh38
rs1029312332 Chr.8: 6502007-6502007 on Build GRCh38
rs904004608 Chr.8: 6502213-6502213 on Build GRCh38
rs539528897 Chr.8: 6502194-6502194 on Build GRCh38
rs33945826 Chr.8: 6501930-6501930 on Build GRCh38
rs764432251 Chr.8: 6502149-6502149 on Build GRCh38
rs933981446 Chr.8: 6502375-6502375 on Build GRCh38
rs375340605 Chr.8: 6502366-6502366 on Build GRCh38
rs1050406760 Chr.8: 6502159-6502159 on Build GRCh38
rs941317363 Chr.8: 6502121-6502121 on Build GRCh38

View Full Product Details