Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6924603-6925113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFA6
Entrez Gene ID
1671
Gene Name
defensin alpha 6
Gene Aliases
DEF6, HD-6
UniGene
Hs.711
Gene Chromosome Location
Chr.8: 6924694-6926076 on Build GRCh38
Transcripts
NM_001926.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3650341 c.258G>A p.G86G Chr.8: 6924863-6924863 on Build GRCh38
3650342 c.244G>A p.E82K Chr.8: 6924877-6924877 on Build GRCh38
6642177 c.264C>T p.C88C Chr.8: 6924857-6924857 on Build GRCh38
3650343 c.239C>T p.S80L Chr.8: 6924882-6924882 on Build GRCh38
7081909 c.288A>G p.R96R Chr.8: 6924833-6924833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948269246 Chr.8: 6924851-6924851 on Build GRCh38
rs765671754 Chr.8: 6924756-6924756 on Build GRCh38
rs777825102 Chr.8: 6924918-6924918 on Build GRCh38
rs766664569 Chr.8: 6924957-6924957 on Build GRCh38
rs926843621 Chr.8: 6924969-6924969 on Build GRCh38
rs146000057 Chr.8: 6924679-6924679 on Build GRCh38
rs1016520554 Chr.8: 6924704-6924704 on Build GRCh38
rs55637039 Chr.8: 6925079-6925079 on Build GRCh38
rs769278174 Chr.8: 6924866-6924866 on Build GRCh38
rs760006114 Chr.8: 6924965-6924965 on Build GRCh38
rs761957877 Chr.8: 6924936-6924937 on Build GRCh38
rs778873847 Chr.8: 6924832-6924832 on Build GRCh38
rs56109247 Chr.8: 6924978-6924978 on Build GRCh38
rs368976343 Chr.8: 6924719-6924719 on Build GRCh38
rs867653453 Chr.8: 6924805-6924805 on Build GRCh38
rs556446021 Chr.8: 6924709-6924709 on Build GRCh38
rs377371973 Chr.8: 6925020-6925020 on Build GRCh38
rs748714753 Chr.8: 6924939-6924939 on Build GRCh38
rs370145720 Chr.8: 6924953-6924953 on Build GRCh38
rs370918510 Chr.8: 6924837-6924837 on Build GRCh38
rs760986660 Chr.8: 6924888-6924888 on Build GRCh38
rs1049634199 Chr.8: 6925006-6925006 on Build GRCh38
rs767558757 Chr.8: 6924647-6924647 on Build GRCh38
rs141214756 Chr.8: 6924929-6924929 on Build GRCh38
rs769315660 Chr.8: 6924932-6924932 on Build GRCh38
rs779545096 Chr.8: 6924997-6924997 on Build GRCh38
rs1002623522 Chr.8: 6924911-6924911 on Build GRCh38
rs533722369 Chr.8: 6924672-6924672 on Build GRCh38
rs570620186 Chr.8: 6924998-6924998 on Build GRCh38
rs762579292 Chr.8: 6924881-6924881 on Build GRCh38
rs770892638 Chr.8: 6924857-6924857 on Build GRCh38
rs866258632 Chr.8: 6924766-6924766 on Build GRCh38
rs993585753 Chr.8: 6925001-6925001 on Build GRCh38
rs750769948 Chr.8: 6924767-6924767 on Build GRCh38
rs895282746 Chr.8: 6924640-6924640 on Build GRCh38
rs933394460 Chr.8: 6924731-6924731 on Build GRCh38
rs866733910 Chr.8: 6924848-6924848 on Build GRCh38
rs1025990165 Chr.8: 6924921-6924921 on Build GRCh38
rs116236599 Chr.8: 6924682-6924682 on Build GRCh38
rs988329650 Chr.8: 6924950-6924950 on Build GRCh38
rs1006157231 Chr.8: 6924928-6924928 on Build GRCh38
rs576432352 Chr.8: 6925091-6925091 on Build GRCh38
rs375822764 Chr.8: 6924936-6924936 on Build GRCh38
rs542330679 Chr.8: 6924696-6924696 on Build GRCh38
rs1003470629 Chr.8: 6924782-6924782 on Build GRCh38
rs561743853 Chr.8: 6924754-6924754 on Build GRCh38
rs754002131 Chr.8: 6924902-6924902 on Build GRCh38
rs372519339 Chr.8: 6924974-6924974 on Build GRCh38
rs758557142 Chr.8: 6924917-6924917 on Build GRCh38
rs899711383 Chr.8: 6925031-6925031 on Build GRCh38
rs1054011348 Chr.8: 6924819-6924819 on Build GRCh38
rs745532765 Chr.8: 6924865-6924865 on Build GRCh38
rs546079495 Chr.8: 6924687-6924687 on Build GRCh38
rs1045751726 Chr.8: 6924774-6924774 on Build GRCh38
rs369318357 Chr.8: 6924867-6924867 on Build GRCh38
rs910873892 Chr.8: 6924629-6924629 on Build GRCh38
rs1018582560 Chr.8: 6924926-6924926 on Build GRCh38
rs906663803 Chr.8: 6925043-6925043 on Build GRCh38
rs768183878 Chr.8: 6924942-6924942 on Build GRCh38
rs201094510 Chr.8: 6924824-6924824 on Build GRCh38
rs186037043 Chr.8: 6924977-6924977 on Build GRCh38
rs746781619 Chr.8: 6924853-6924853 on Build GRCh38
rs866387127 Chr.8: 6924847-6924847 on Build GRCh38
rs45618544 Chr.8: 6925014-6925014 on Build GRCh38
rs948294752 Chr.8: 6925007-6925007 on Build GRCh38
rs761183084 Chr.8: 6924954-6924954 on Build GRCh38
rs746977942 Chr.8: 6924924-6924924 on Build GRCh38
rs771901375 Chr.8: 6924840-6924840 on Build GRCh38
rs61745836 Chr.8: 6924863-6924863 on Build GRCh38
rs764296888 Chr.8: 6924973-6924973 on Build GRCh38
rs1022691068 Chr.8: 6924648-6924648 on Build GRCh38
rs886073376 Chr.8: 6924927-6924927 on Build GRCh38
rs975252270 Chr.8: 6924718-6924718 on Build GRCh38
rs181510765 Chr.8: 6924908-6924908 on Build GRCh38
rs905462622 Chr.8: 6924778-6924778 on Build GRCh38
rs778684635 Chr.8: 6924810-6924810 on Build GRCh38
rs895321398 Chr.8: 6924828-6924828 on Build GRCh38
rs1005910425 Chr.8: 6925078-6925078 on Build GRCh38
rs755160781 Chr.8: 6924904-6924904 on Build GRCh38
rs773660947 Chr.8: 6924951-6924951 on Build GRCh38
rs191201019 Chr.8: 6925067-6925067 on Build GRCh38
rs938421637 Chr.8: 6925054-6925054 on Build GRCh38
rs565866929 Chr.8: 6924654-6924654 on Build GRCh38
rs756517829 Chr.8: 6924801-6924801 on Build GRCh38
rs56239909 Chr.8: 6924757-6924757 on Build GRCh38
rs967832610 Chr.8: 6925088-6925088 on Build GRCh38
rs770285253 Chr.8: 6925028-6925028 on Build GRCh38
rs868528284 Chr.8: 6924870-6924870 on Build GRCh38
rs1016330735 Chr.8: 6925080-6925080 on Build GRCh38
rs777917250 Chr.8: 6924852-6924852 on Build GRCh38
rs750008948 Chr.8: 6924836-6924836 on Build GRCh38
rs555427579 Chr.8: 6924642-6924642 on Build GRCh38
rs374916917 Chr.8: 6924964-6924964 on Build GRCh38
rs954794683 Chr.8: 6924963-6924963 on Build GRCh38
rs866592808 Chr.8: 6924803-6924803 on Build GRCh38
rs1047486351 Chr.8: 6925061-6925061 on Build GRCh38
rs56023225 Chr.8: 6924688-6924688 on Build GRCh38
rs570049104 Chr.8: 6924931-6924931 on Build GRCh38
rs757012875 Chr.8: 6924807-6924807 on Build GRCh38
rs712276 Chr.8: 6924749-6924749 on Build GRCh38
rs56292948 Chr.8: 6924681-6924681 on Build GRCh38
rs1025470172 Chr.8: 6924879-6924879 on Build GRCh38
rs757073410 Chr.8: 6924925-6924925 on Build GRCh38
rs766752430 Chr.8: 6924899-6924899 on Build GRCh38
rs148628406 Chr.8: 6924887-6924887 on Build GRCh38
rs754089949 Chr.8: 6924813-6924813 on Build GRCh38
rs767987440 Chr.8: 6924884-6924884 on Build GRCh38
rs922315564 Chr.8: 6924680-6924680 on Build GRCh38
rs146685654 Chr.8: 6924993-6924993 on Build GRCh38
rs577437053 Chr.8: 6924967-6924967 on Build GRCh38
rs533406496 Chr.8: 6924869-6924869 on Build GRCh38
rs143254168 Chr.8: 6924968-6924968 on Build GRCh38

View Full Product Details