Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7828657-7829182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB106A
Entrez Gene ID
245909
Gene Name
defensin beta 106A
Gene Aliases
BD-6, DEFB-6, DEFB106
UniGene
Hs.535925
Gene Chromosome Location
Chr.8: 7825172-7829053 on Build GRCh38
Transcripts
NM_152251.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5763958 c.50-4G>A p.? Chr.8: 7828801-7828801 on Build GRCh38
7364052 c.141C>A p.C47* Chr.8: 7828896-7828896 on Build GRCh38
3951877 c.74A>C p.K25T Chr.8: 7828829-7828829 on Build GRCh38
5480290 c.120T>C p.N40N Chr.8: 7828875-7828875 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543897377 Chr.8: 7828891-7828891 on Build GRCh38
rs762612245 Chr.8: 7828916-7828916 on Build GRCh38
rs556344742 Chr.8: 7828706-7828706 on Build GRCh38
rs2979554 Chr.8: 7828678-7828678 on Build GRCh38
rs778039951 Chr.8: 7828877-7828877 on Build GRCh38
rs751253131 Chr.8: 7828862-7828862 on Build GRCh38
rs752595285 Chr.8: 7828794-7828794 on Build GRCh38
rs745946406 Chr.8: 7828899-7828899 on Build GRCh38
rs780992152 Chr.8: 7828815-7828816 on Build GRCh38
rs201085281 Chr.8: 7828801-7828801 on Build GRCh38
rs555761850 Chr.8: 7828736-7828736 on Build GRCh38
rs772408663 Chr.8: 7828771-7828771 on Build GRCh38
rs765696029 Chr.8: 7828955-7828955 on Build GRCh38
rs200114200 Chr.8: 7828759-7828759 on Build GRCh38
rs764943262 Chr.8: 7828754-7828754 on Build GRCh38
rs763804796 Chr.8: 7828796-7828796 on Build GRCh38
rs561848758 Chr.8: 7828978-7828978 on Build GRCh38
rs774751474 Chr.8: 7828770-7828770 on Build GRCh38
rs146195936 Chr.8: 7828875-7828875 on Build GRCh38
rs563695236 Chr.8: 7829126-7829126 on Build GRCh38
rs763600222 Chr.8: 7828854-7828854 on Build GRCh38
rs535791008 Chr.8: 7828728-7828728 on Build GRCh38
rs775354809 Chr.8: 7828847-7828847 on Build GRCh38
rs778081218 Chr.8: 7828815-7828815 on Build GRCh38
rs761425228 Chr.8: 7828863-7828863 on Build GRCh38
rs578149320 Chr.8: 7828788-7828788 on Build GRCh38
rs771012830 Chr.8: 7828817-7828817 on Build GRCh38
rs1055885510 Chr.8: 7829045-7829045 on Build GRCh38
rs779407458 Chr.8: 7828868-7828868 on Build GRCh38
rs372269010 Chr.8: 7828911-7828911 on Build GRCh38
rs755413890 Chr.8: 7828867-7828867 on Build GRCh38
rs753217606 Chr.8: 7828956-7828956 on Build GRCh38
rs542137561 Chr.8: 7828946-7828946 on Build GRCh38
rs2244098 Chr.8: 7828756-7828756 on Build GRCh38
rs868481065 Chr.8: 7829022-7829022 on Build GRCh38
rs748520036 Chr.8: 7828814-7828814 on Build GRCh38
rs374912581 Chr.8: 7828943-7828943 on Build GRCh38
rs532901826 Chr.8: 7829127-7829127 on Build GRCh38
rs773101426 Chr.8: 7828778-7828778 on Build GRCh38
rs755541997 Chr.8: 7828802-7828802 on Build GRCh38
rs549460664 Chr.8: 7829129-7829129 on Build GRCh38
rs572439615 Chr.8: 7828740-7828740 on Build GRCh38
rs868211353 Chr.8: 7829156-7829156 on Build GRCh38
rs767005789 Chr.8: 7828866-7828866 on Build GRCh38
rs769916753 Chr.8: 7828840-7828840 on Build GRCh38
rs867339000 Chr.8: 7828703-7828703 on Build GRCh38
rs936119641 Chr.8: 7828968-7828968 on Build GRCh38
rs866573390 Chr.8: 7828741-7828741 on Build GRCh38
rs569903827 Chr.8: 7828714-7828714 on Build GRCh38
rs570475597 Chr.8: 7828686-7828686 on Build GRCh38
rs539098974 Chr.8: 7828689-7828689 on Build GRCh38
rs375735859 Chr.8: 7828787-7828787 on Build GRCh38
rs747232056 Chr.8: 7828879-7828879 on Build GRCh38
rs771116346 Chr.8: 7828882-7828882 on Build GRCh38
rs867986701 Chr.8: 7829012-7829012 on Build GRCh38
rs761526983 Chr.8: 7828934-7828934 on Build GRCh38
rs760824755 Chr.8: 7828779-7828779 on Build GRCh38
rs367687834 Chr.8: 7828799-7828799 on Build GRCh38
rs756508999 Chr.8: 7828760-7828760 on Build GRCh38
rs759532256 Chr.8: 7828836-7828836 on Build GRCh38
rs749601925 Chr.8: 7828765-7828765 on Build GRCh38
rs547080236 Chr.8: 7829118-7829118 on Build GRCh38
rs766178290 Chr.8: 7828780-7828780 on Build GRCh38
rs150372518 Chr.8: 7828694-7828694 on Build GRCh38
rs534726274 Chr.8: 7828766-7828766 on Build GRCh38
rs745699552 Chr.8: 7828853-7828853 on Build GRCh38
rs774075286 Chr.8: 7828919-7828919 on Build GRCh38
rs569942542 Chr.8: 7829140-7829140 on Build GRCh38
rs776899335 Chr.8: 7828833-7828833 on Build GRCh38
rs769793915 Chr.8: 7828903-7828903 on Build GRCh38
rs762816989 Chr.8: 7828850-7828850 on Build GRCh38
rs768462948 Chr.8: 7828918-7828918 on Build GRCh38
rs751933509 Chr.8: 7828990-7828990 on Build GRCh38
rs758879050 Chr.8: 7828958-7828958 on Build GRCh38
rs779247935 Chr.8: 7828812-7828812 on Build GRCh38
rs558106657 Chr.8: 7828785-7828785 on Build GRCh38
rs757981636 Chr.8: 7828795-7828795 on Build GRCh38
rs867067367 Chr.8: 7829058-7829058 on Build GRCh38
rs765102425 Chr.8: 7828790-7828790 on Build GRCh38
rs780619673 Chr.8: 7828761-7828761 on Build GRCh38
rs758823117 Chr.8: 7828876-7828876 on Build GRCh38
rs750973928 Chr.8: 7828798-7828798 on Build GRCh38
rs867947124 Chr.8: 7829110-7829110 on Build GRCh38
rs527553382 Chr.8: 7829085-7829085 on Build GRCh38

View Full Product Details