Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7972380-7972881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66E
Entrez Gene ID
100132103
Gene Name
family with sequence similarity 66 member E
Gene Aliases
-
UniGene
Hs.694680
Gene Chromosome Location
Chr.8: 7955013-8008755 on Build GRCh38
Transcripts
NR_027424.1 NM_001256872.1

Gene Symbol
USP17L8
Entrez Gene ID
392188
Gene Name
ubiquitin specific peptidase 17-like family member 8
Gene Aliases
USP17L10
UniGene
Hs.741135
Gene Chromosome Location
Chr.8: 7971661-7973253 on Build GRCh38
Transcripts
NR_027424.1 NM_001256872.1

SNP Detail

SNP ID Chromosome Location
rs200618475 Chr.8: 7972810-7972810 on Build GRCh38
rs567019282 Chr.8: 7972423-7972423 on Build GRCh38
rs747399518 Chr.8: 7972554-7972554 on Build GRCh38
rs201612021 Chr.8: 7972775-7972775 on Build GRCh38
rs761730438 Chr.8: 7972848-7972848 on Build GRCh38
rs778144657 Chr.8: 7972507-7972507 on Build GRCh38
rs767460221 Chr.8: 7972856-7972856 on Build GRCh38
rs1005031588 Chr.8: 7972799-7972799 on Build GRCh38
rs775497047 Chr.8: 7972655-7972655 on Build GRCh38
rs779831273 Chr.8: 7972603-7972604 on Build GRCh38
rs192094999 Chr.8: 7972496-7972496 on Build GRCh38
rs372606266 Chr.8: 7972841-7972841 on Build GRCh38
rs781365523 Chr.8: 7972585-7972585 on Build GRCh38
rs371617071 Chr.8: 7972457-7972457 on Build GRCh38
rs187300358 Chr.8: 7972403-7972403 on Build GRCh38
rs769900770 Chr.8: 7972618-7972618 on Build GRCh38
rs757434420 Chr.8: 7972573-7972573 on Build GRCh38
rs546887922 Chr.8: 7972417-7972417 on Build GRCh38
rs749180205 Chr.8: 7972772-7972772 on Build GRCh38
rs375358016 Chr.8: 7972854-7972854 on Build GRCh38
rs774238038 Chr.8: 7972803-7972803 on Build GRCh38
rs199543709 Chr.8: 7972777-7972777 on Build GRCh38
rs746134425 Chr.8: 7972593-7972593 on Build GRCh38
rs758835975 Chr.8: 7972502-7972502 on Build GRCh38
rs368343535 Chr.8: 7972831-7972831 on Build GRCh38
rs753244398 Chr.8: 7972499-7972499 on Build GRCh38

View Full Product Details