Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7977514-7978022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66E
Entrez Gene ID
100132103
Gene Name
family with sequence similarity 66 member E
Gene Aliases
-
UniGene
Hs.694680
Gene Chromosome Location
Chr.8: 7955013-8008755 on Build GRCh38
Transcripts
NR_027424.1 NM_001256871.1

Gene Symbol
USP17L3
Entrez Gene ID
645836
Gene Name
ubiquitin specific peptidase 17-like family member 3
Gene Aliases
USP17B, USP17F
UniGene
Hs.741131
Gene Chromosome Location
Chr.8: 7976393-7977985 on Build GRCh38
Transcripts
NR_027424.1 NM_001256871.1

SNP Detail

SNP ID Chromosome Location
rs762001481 Chr.8: 7977542-7977542 on Build GRCh38
rs754969934 Chr.8: 7977915-7977915 on Build GRCh38
rs188014243 Chr.8: 7977977-7977977 on Build GRCh38
rs191361756 Chr.8: 7977535-7977535 on Build GRCh38
rs1036510049 Chr.8: 7977853-7977854 on Build GRCh38
rs199988858 Chr.8: 7977714-7977714 on Build GRCh38
rs777357524 Chr.8: 7977965-7977965 on Build GRCh38
rs201264829 Chr.8: 7977743-7977743 on Build GRCh38
rs548941570 Chr.8: 7977554-7977554 on Build GRCh38
rs201374304 Chr.8: 7977703-7977703 on Build GRCh38
rs182610233 Chr.8: 7977563-7977563 on Build GRCh38
rs202157266 Chr.8: 7977750-7977750 on Build GRCh38
rs202084160 Chr.8: 7977872-7977872 on Build GRCh38
rs771438536 Chr.8: 7977975-7977976 on Build GRCh38
rs751141756 Chr.8: 7977976-7977976 on Build GRCh38
rs772189886 Chr.8: 7977547-7977547 on Build GRCh38
rs527932174 Chr.8: 7977981-7977981 on Build GRCh38
rs752387672 Chr.8: 7977937-7977937 on Build GRCh38
rs200143156 Chr.8: 7977665-7977665 on Build GRCh38
rs200743959 Chr.8: 7977681-7977681 on Build GRCh38
rs780679617 Chr.8: 7977988-7977988 on Build GRCh38
rs760557502 Chr.8: 7977575-7977575 on Build GRCh38
rs370184970 Chr.8: 7977911-7977911 on Build GRCh38
rs777008214 Chr.8: 7977976-7977977 on Build GRCh38
rs200263085 Chr.8: 7977800-7977800 on Build GRCh38
rs200975552 Chr.8: 7977852-7977852 on Build GRCh38
rs201549590 Chr.8: 7977959-7977959 on Build GRCh38
rs200415368 Chr.8: 7977919-7977919 on Build GRCh38

View Full Product Details