Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11424036-11424541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
UniGene
Hs.583896
Gene Chromosome Location
Chr.8: 11368402-11438657 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643471 c.563C>A p.S188Y Chr.8: 11424455-11424455 on Build GRCh38
4749961 c.520C>T p.R174W Chr.8: 11424498-11424498 on Build GRCh38
3781362 c.534C>T p.A178A Chr.8: 11424484-11424484 on Build GRCh38
244371 c.601G>A p.V201M Chr.8: 11424417-11424417 on Build GRCh38
5890345 c.567C>T p.S189S Chr.8: 11424451-11424451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989481310 Chr.8: 11424269-11424269 on Build GRCh38
rs747371658 Chr.8: 11424369-11424369 on Build GRCh38
rs566542751 Chr.8: 11424165-11424165 on Build GRCh38
rs768510757 Chr.8: 11424454-11424454 on Build GRCh38
rs1030721261 Chr.8: 11424343-11424343 on Build GRCh38
rs112746311 Chr.8: 11424074-11424074 on Build GRCh38
rs755737011 Chr.8: 11424399-11424399 on Build GRCh38
rs752012096 Chr.8: 11424488-11424488 on Build GRCh38
rs773785540 Chr.8: 11424323-11424341 on Build GRCh38
rs963199025 Chr.8: 11424320-11424320 on Build GRCh38
rs905876284 Chr.8: 11424103-11424103 on Build GRCh38
rs963291153 Chr.8: 11424148-11424148 on Build GRCh38
rs950574406 Chr.8: 11424125-11424125 on Build GRCh38
rs757923888 Chr.8: 11424139-11424139 on Build GRCh38
rs761572595 Chr.8: 11424394-11424394 on Build GRCh38
rs547076790 Chr.8: 11424336-11424336 on Build GRCh38
rs570307661 Chr.8: 11424124-11424124 on Build GRCh38
rs368459570 Chr.8: 11424417-11424417 on Build GRCh38
rs984126198 Chr.8: 11424448-11424448 on Build GRCh38
rs750839979 Chr.8: 11424494-11424494 on Build GRCh38
rs764004616 Chr.8: 11424337-11424337 on Build GRCh38
rs202235860 Chr.8: 11424340-11424340 on Build GRCh38
rs759963618 Chr.8: 11424450-11424452 on Build GRCh38
rs947549133 Chr.8: 11424077-11424077 on Build GRCh38
rs764168254 Chr.8: 11424393-11424393 on Build GRCh38
rs376807405 Chr.8: 11424325-11424325 on Build GRCh38
rs1023001071 Chr.8: 11424096-11424096 on Build GRCh38
rs373564873 Chr.8: 11424339-11424339 on Build GRCh38
rs927414203 Chr.8: 11424290-11424290 on Build GRCh38
rs996008684 Chr.8: 11424321-11424321 on Build GRCh38
rs369090343 Chr.8: 11424497-11424497 on Build GRCh38
rs997889779 Chr.8: 11424217-11424217 on Build GRCh38
rs761747530 Chr.8: 11424464-11424464 on Build GRCh38
rs143743025 Chr.8: 11424421-11424421 on Build GRCh38
rs1053314727 Chr.8: 11424223-11424223 on Build GRCh38
rs566681543 Chr.8: 11424395-11424395 on Build GRCh38
rs139424835 Chr.8: 11424455-11424455 on Build GRCh38
rs537742836 Chr.8: 11424330-11424330 on Build GRCh38
rs561322004 Chr.8: 11424483-11424483 on Build GRCh38
rs905824381 Chr.8: 11424253-11424253 on Build GRCh38
rs146816358 Chr.8: 11424490-11424490 on Build GRCh38
rs779476502 Chr.8: 11424350-11424350 on Build GRCh38
rs923747374 Chr.8: 11424208-11424208 on Build GRCh38
rs200511231 Chr.8: 11424328-11424328 on Build GRCh38
rs987590751 Chr.8: 11424205-11424205 on Build GRCh38
rs754451449 Chr.8: 11424405-11424405 on Build GRCh38
rs765900981 Chr.8: 11424400-11424400 on Build GRCh38
rs1048276175 Chr.8: 11424149-11424149 on Build GRCh38
rs1016067086 Chr.8: 11424318-11424318 on Build GRCh38
rs141292162 Chr.8: 11424418-11424418 on Build GRCh38
rs771780754 Chr.8: 11424465-11424465 on Build GRCh38
rs901019403 Chr.8: 11424514-11424514 on Build GRCh38
rs372968845 Chr.8: 11424505-11424505 on Build GRCh38
rs1038192816 Chr.8: 11424440-11424440 on Build GRCh38
rs372350472 Chr.8: 11424473-11424473 on Build GRCh38
rs755649418 Chr.8: 11424347-11424347 on Build GRCh38
rs771433749 Chr.8: 11424378-11424381 on Build GRCh38
rs778137618 Chr.8: 11424367-11424367 on Build GRCh38
rs765620953 Chr.8: 11424515-11424522 on Build GRCh38
rs954379717 Chr.8: 11424198-11424198 on Build GRCh38
rs551503670 Chr.8: 11424144-11424144 on Build GRCh38
rs772010209 Chr.8: 11424508-11424508 on Build GRCh38
rs746092041 Chr.8: 11424429-11424429 on Build GRCh38
rs370672901 Chr.8: 11424314-11424314 on Build GRCh38
rs989648123 Chr.8: 11424381-11424381 on Build GRCh38
rs749519377 Chr.8: 11424498-11424498 on Build GRCh38
rs1057054458 Chr.8: 11424133-11424133 on Build GRCh38
rs921576687 Chr.8: 11424185-11424185 on Build GRCh38
rs753301707 Chr.8: 11424403-11424403 on Build GRCh38
rs757255814 Chr.8: 11424507-11424507 on Build GRCh38
rs900911506 Chr.8: 11424170-11424170 on Build GRCh38
rs199968652 Chr.8: 11424392-11424392 on Build GRCh38
rs370416023 Chr.8: 11424354-11424354 on Build GRCh38
rs567983296 Chr.8: 11424071-11424071 on Build GRCh38
rs149662282 Chr.8: 11424511-11424511 on Build GRCh38
rs556837921 Chr.8: 11424110-11424110 on Build GRCh38
rs942595478 Chr.8: 11424496-11424496 on Build GRCh38
rs777181659 Chr.8: 11424423-11424424 on Build GRCh38
rs368537961 Chr.8: 11424342-11424342 on Build GRCh38
rs775453862 Chr.8: 11424398-11424398 on Build GRCh38
rs572193573 Chr.8: 11424435-11424435 on Build GRCh38
rs770101595 Chr.8: 11424431-11424431 on Build GRCh38
rs934852357 Chr.8: 11424061-11424061 on Build GRCh38
rs768845937 Chr.8: 11424444-11424444 on Build GRCh38
rs1004756231 Chr.8: 11424134-11424134 on Build GRCh38
rs555254008 Chr.8: 11424277-11424277 on Build GRCh38
rs771301144 Chr.8: 11424375-11424375 on Build GRCh38
rs1022321826 Chr.8: 11424351-11424351 on Build GRCh38
rs760272883 Chr.8: 11424479-11424479 on Build GRCh38
rs373612696 Chr.8: 11424341-11424341 on Build GRCh38
rs775785936 Chr.8: 11424516-11424517 on Build GRCh38
rs757724438 Chr.8: 11424489-11424489 on Build GRCh38
rs374639232 Chr.8: 11424409-11424409 on Build GRCh38
rs927361717 Chr.8: 11424122-11424123 on Build GRCh38
rs1006705539 Chr.8: 11424168-11424168 on Build GRCh38
rs758850719 Chr.8: 11424355-11424355 on Build GRCh38
rs893427564 Chr.8: 11424073-11424073 on Build GRCh38
rs1044687619 Chr.8: 11424250-11424250 on Build GRCh38
rs759102263 Chr.8: 11424484-11424484 on Build GRCh38
rs536905756 Chr.8: 11424081-11424081 on Build GRCh38
rs769766563 Chr.8: 11424388-11424388 on Build GRCh38
rs534004738 Chr.8: 11424228-11424228 on Build GRCh38
rs747160964 Chr.8: 11424120-11424120 on Build GRCh38
rs368405922 Chr.8: 11424492-11424492 on Build GRCh38
rs947590249 Chr.8: 11424230-11424230 on Build GRCh38
rs934821857 Chr.8: 11424218-11424218 on Build GRCh38
rs760508594 Chr.8: 11424516-11424516 on Build GRCh38
rs746097006 Chr.8: 11424387-11424387 on Build GRCh38
rs1057461198 Chr.8: 11424276-11424276 on Build GRCh38
rs144838125 Chr.8: 11424072-11424072 on Build GRCh38
rs766056567 Chr.8: 11424481-11424481 on Build GRCh38
rs763754784 Chr.8: 11424343-11424344 on Build GRCh38
rs938768205 Chr.8: 11424301-11424301 on Build GRCh38
rs938712129 Chr.8: 11424123-11424123 on Build GRCh38
rs940760420 Chr.8: 11424434-11424434 on Build GRCh38
rs778299082 Chr.8: 11424408-11424408 on Build GRCh38
rs1002884082 Chr.8: 11424275-11424275 on Build GRCh38
rs761098784 Chr.8: 11424378-11424378 on Build GRCh38
rs1005107747 Chr.8: 11424284-11424284 on Build GRCh38

View Full Product Details